Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22097916-22098415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
UniGene
Hs.491223
Gene Chromosome Location
Chr.8: 22089150-22104380 on Build GRCh38
Transcripts
NM_022749.5 XM_005273602.3 XM_006716381.2 XM_006716385.3 XM_017013732.1 XM_017013733.1 XM_017013734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097992 c.724G>C p.E242Q Chr.8: 22098266-22098266 on Build GRCh38
4656027 c.527G>A p.G176D Chr.8: 22098069-22098069 on Build GRCh38
5184366 c.700G>A p.E234K Chr.8: 22098242-22098242 on Build GRCh38
1743298 c.712G>A p.G238S Chr.8: 22098254-22098254 on Build GRCh38
1097991 c.544A>G p.R182G Chr.8: 22098086-22098086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772236485 Chr.8: 22098298-22098298 on Build GRCh38
rs376369856 Chr.8: 22098088-22098088 on Build GRCh38
rs762336631 Chr.8: 22098338-22098338 on Build GRCh38
rs955748037 Chr.8: 22098340-22098340 on Build GRCh38
rs370489747 Chr.8: 22098167-22098167 on Build GRCh38
rs746102610 Chr.8: 22098217-22098217 on Build GRCh38
rs1000409059 Chr.8: 22098300-22098300 on Build GRCh38
rs774151994 Chr.8: 22098111-22098111 on Build GRCh38
rs532690794 Chr.8: 22097937-22097937 on Build GRCh38
rs754726784 Chr.8: 22098073-22098073 on Build GRCh38
rs536903609 Chr.8: 22097979-22097979 on Build GRCh38
rs1025807235 Chr.8: 22097975-22097975 on Build GRCh38
rs777641408 Chr.8: 22098262-22098262 on Build GRCh38
rs556491204 Chr.8: 22098099-22098099 on Build GRCh38
rs956810185 Chr.8: 22098033-22098033 on Build GRCh38
rs753837133 Chr.8: 22098270-22098270 on Build GRCh38
rs934404262 Chr.8: 22097946-22097946 on Build GRCh38
rs898250344 Chr.8: 22097940-22097940 on Build GRCh38
rs775878886 Chr.8: 22098126-22098126 on Build GRCh38
rs561649776 Chr.8: 22098102-22098102 on Build GRCh38
rs577309670 Chr.8: 22098059-22098059 on Build GRCh38
rs760569288 Chr.8: 22098316-22098316 on Build GRCh38
rs200255721 Chr.8: 22098161-22098161 on Build GRCh38
rs981331843 Chr.8: 22098114-22098114 on Build GRCh38
rs752489254 Chr.8: 22098255-22098255 on Build GRCh38
rs775762730 Chr.8: 22098226-22098226 on Build GRCh38
rs146497282 Chr.8: 22098359-22098359 on Build GRCh38
rs916959214 Chr.8: 22098231-22098231 on Build GRCh38
rs767138257 Chr.8: 22098376-22098376 on Build GRCh38
rs567319267 Chr.8: 22098009-22098009 on Build GRCh38
rs569323236 Chr.8: 22097972-22097972 on Build GRCh38
rs891272019 Chr.8: 22098178-22098178 on Build GRCh38
rs764558215 Chr.8: 22098142-22098142 on Build GRCh38
rs891326082 Chr.8: 22097994-22097994 on Build GRCh38
rs34061344 Chr.8: 22098004-22098005 on Build GRCh38
rs749442004 Chr.8: 22098194-22098194 on Build GRCh38
rs199793638 Chr.8: 22098190-22098190 on Build GRCh38
rs139434422 Chr.8: 22098254-22098254 on Build GRCh38
rs767482632 Chr.8: 22098233-22098253 on Build GRCh38
rs773782617 Chr.8: 22098345-22098345 on Build GRCh38
rs755965274 Chr.8: 22098259-22098259 on Build GRCh38
rs764069825 Chr.8: 22098386-22098386 on Build GRCh38
rs761005351 Chr.8: 22098236-22098236 on Build GRCh38
rs369587302 Chr.8: 22098267-22098267 on Build GRCh38
rs752395868 Chr.8: 22098380-22098380 on Build GRCh38
rs965587923 Chr.8: 22098090-22098090 on Build GRCh38
rs968982793 Chr.8: 22098287-22098287 on Build GRCh38
rs373199574 Chr.8: 22098362-22098362 on Build GRCh38
rs201195171 Chr.8: 22098242-22098242 on Build GRCh38
rs754608926 Chr.8: 22098184-22098184 on Build GRCh38
rs1028555787 Chr.8: 22098143-22098143 on Build GRCh38
rs1039901286 Chr.8: 22098199-22098199 on Build GRCh38
rs769014264 Chr.8: 22098318-22098318 on Build GRCh38
rs532651194 Chr.8: 22098191-22098191 on Build GRCh38
rs770303251 Chr.8: 22098343-22098343 on Build GRCh38
rs929732235 Chr.8: 22098164-22098164 on Build GRCh38
rs761120943 Chr.8: 22098140-22098140 on Build GRCh38
rs770307508 Chr.8: 22098018-22098018 on Build GRCh38
rs188321970 Chr.8: 22098368-22098368 on Build GRCh38
rs375766097 Chr.8: 22098192-22098192 on Build GRCh38
rs1973254 Chr.8: 22098041-22098041 on Build GRCh38
rs552781211 Chr.8: 22098058-22098058 on Build GRCh38
rs199544823 Chr.8: 22098087-22098087 on Build GRCh38
rs59684172 Chr.8: 22098100-22098100 on Build GRCh38
rs963986897 Chr.8: 22098001-22098001 on Build GRCh38
rs745933677 Chr.8: 22098283-22098283 on Build GRCh38
rs372267926 Chr.8: 22098209-22098209 on Build GRCh38
rs200264326 Chr.8: 22098079-22098079 on Build GRCh38
rs1041153040 Chr.8: 22098257-22098257 on Build GRCh38
rs749985467 Chr.8: 22098153-22098153 on Build GRCh38
rs747442134 Chr.8: 22098317-22098317 on Build GRCh38
rs372578475 Chr.8: 22098241-22098241 on Build GRCh38
rs542918588 Chr.8: 22098101-22098101 on Build GRCh38
rs867942517 Chr.8: 22098137-22098137 on Build GRCh38
rs971191297 Chr.8: 22098367-22098367 on Build GRCh38
rs761792801 Chr.8: 22098140-22098140 on Build GRCh38
rs146742522 Chr.8: 22098139-22098139 on Build GRCh38
rs1018501574 Chr.8: 22097997-22097997 on Build GRCh38
rs1047638532 Chr.8: 22098250-22098250 on Build GRCh38
rs1028441106 Chr.8: 22098279-22098279 on Build GRCh38
rs759424977 Chr.8: 22098361-22098361 on Build GRCh38
rs373853188 Chr.8: 22098094-22098094 on Build GRCh38
rs778894231 Chr.8: 22098277-22098277 on Build GRCh38
rs780310659 Chr.8: 22098310-22098310 on Build GRCh38
rs144079413 Chr.8: 22098273-22098273 on Build GRCh38
rs372814624 Chr.8: 22098253-22098253 on Build GRCh38
rs780878781 Chr.8: 22098188-22098188 on Build GRCh38
rs753147403 Chr.8: 22098032-22098032 on Build GRCh38
rs368245254 Chr.8: 22098205-22098205 on Build GRCh38
rs549272533 Chr.8: 22097982-22097982 on Build GRCh38
rs920427856 Chr.8: 22098371-22098371 on Build GRCh38
rs779659011 Chr.8: 22098068-22098068 on Build GRCh38
rs776924995 Chr.8: 22098334-22098334 on Build GRCh38
rs899061748 Chr.8: 22098013-22098013 on Build GRCh38
rs778421444 Chr.8: 22098049-22098049 on Build GRCh38
rs376449220 Chr.8: 22098307-22098307 on Build GRCh38
rs746881991 Chr.8: 22098069-22098069 on Build GRCh38
rs368525379 Chr.8: 22098233-22098233 on Build GRCh38
rs765894932 Chr.8: 22098251-22098251 on Build GRCh38
rs550988593 Chr.8: 22097942-22097942 on Build GRCh38
rs774165278 Chr.8: 22098058-22098059 on Build GRCh38
rs185486017 Chr.8: 22098163-22098163 on Build GRCh38
rs767790093 Chr.8: 22098105-22098105 on Build GRCh38
rs77609062 Chr.8: 22098383-22098383 on Build GRCh38
rs760343873 Chr.8: 22098385-22098385 on Build GRCh38
rs377271906 Chr.8: 22098220-22098220 on Build GRCh38
rs979467510 Chr.8: 22098011-22098011 on Build GRCh38
rs1008417774 Chr.8: 22097996-22097996 on Build GRCh38

View Full Product Details