Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22099183-22099673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
UniGene
Hs.491223
Gene Chromosome Location
Chr.8: 22089150-22104380 on Build GRCh38
Transcripts
NM_022749.5 XM_005273602.3 XM_006716381.2 XM_006716385.3 XM_017013732.1 XM_017013733.1 XM_017013734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097997 c.1094C>T p.A365V Chr.8: 22099303-22099303 on Build GRCh38
1097995 c.1093G>A p.A365T Chr.8: 22099302-22099302 on Build GRCh38
4423102 c.1119C>A p.F373L Chr.8: 22099328-22099328 on Build GRCh38
1097998 c.272C>T p.A91V Chr.8: 22099303-22099303 on Build GRCh38
1097996 c.271G>A p.A91T Chr.8: 22099302-22099302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766671478 Chr.8: 22099304-22099304 on Build GRCh38
rs928157703 Chr.8: 22099481-22099481 on Build GRCh38
rs901045053 Chr.8: 22099639-22099639 on Build GRCh38
rs763543931 Chr.8: 22099261-22099261 on Build GRCh38
rs745732311 Chr.8: 22099268-22099268 on Build GRCh38
rs764792621 Chr.8: 22099389-22099390 on Build GRCh38
rs74482324 Chr.8: 22099640-22099640 on Build GRCh38
rs772523527 Chr.8: 22099400-22099400 on Build GRCh38
rs71513865 Chr.8: 22099307-22099307 on Build GRCh38
rs907974432 Chr.8: 22099582-22099582 on Build GRCh38
rs758132943 Chr.8: 22099269-22099269 on Build GRCh38
rs920639989 Chr.8: 22099290-22099290 on Build GRCh38
rs373697515 Chr.8: 22099295-22099295 on Build GRCh38
rs779748306 Chr.8: 22099338-22099338 on Build GRCh38
rs930681600 Chr.8: 22099539-22099539 on Build GRCh38
rs1050033862 Chr.8: 22099651-22099651 on Build GRCh38
rs749899460 Chr.8: 22099409-22099409 on Build GRCh38
rs975055005 Chr.8: 22099207-22099207 on Build GRCh38
rs760001061 Chr.8: 22099315-22099315 on Build GRCh38
rs746792316 Chr.8: 22099345-22099345 on Build GRCh38
rs1048248965 Chr.8: 22099540-22099540 on Build GRCh38
rs777947840 Chr.8: 22099381-22099381 on Build GRCh38
rs756553229 Chr.8: 22099329-22099329 on Build GRCh38
rs539293067 Chr.8: 22099380-22099380 on Build GRCh38
rs749502726 Chr.8: 22099390-22099390 on Build GRCh38
rs937957165 Chr.8: 22099270-22099270 on Build GRCh38
rs761113401 Chr.8: 22099406-22099406 on Build GRCh38
rs371611700 Chr.8: 22099223-22099223 on Build GRCh38
rs886921285 Chr.8: 22099311-22099311 on Build GRCh38
rs529750579 Chr.8: 22099518-22099518 on Build GRCh38
rs1004017161 Chr.8: 22099317-22099317 on Build GRCh38
rs368759728 Chr.8: 22099452-22099454 on Build GRCh38
rs763205691 Chr.8: 22099303-22099303 on Build GRCh38
rs1055533612 Chr.8: 22099273-22099273 on Build GRCh38
rs376698794 Chr.8: 22099222-22099222 on Build GRCh38
rs756506497 Chr.8: 22099470-22099470 on Build GRCh38
rs368168499 Chr.8: 22099328-22099328 on Build GRCh38
rs756950679 Chr.8: 22099264-22099264 on Build GRCh38
rs989276254 Chr.8: 22099213-22099213 on Build GRCh38
rs202206983 Chr.8: 22099292-22099292 on Build GRCh38
rs763403818 Chr.8: 22099257-22099257 on Build GRCh38
rs749826155 Chr.8: 22099521-22099521 on Build GRCh38
rs779841753 Chr.8: 22099271-22099271 on Build GRCh38
rs371554448 Chr.8: 22099358-22099358 on Build GRCh38
rs569115468 Chr.8: 22099389-22099389 on Build GRCh38
rs755462688 Chr.8: 22099260-22099260 on Build GRCh38
rs1020719814 Chr.8: 22099425-22099425 on Build GRCh38
rs746946498 Chr.8: 22099288-22099288 on Build GRCh38
rs771153654 Chr.8: 22099393-22099393 on Build GRCh38
rs1028396291 Chr.8: 22099361-22099361 on Build GRCh38
rs957755688 Chr.8: 22099368-22099368 on Build GRCh38
rs369641176 Chr.8: 22099394-22099394 on Build GRCh38
rs372343548 Chr.8: 22099408-22099408 on Build GRCh38
rs753070144 Chr.8: 22099426-22099426 on Build GRCh38
rs759930523 Chr.8: 22099399-22099399 on Build GRCh38
rs199982834 Chr.8: 22099293-22099293 on Build GRCh38
rs773569831 Chr.8: 22099296-22099296 on Build GRCh38
rs751610973 Chr.8: 22099291-22099291 on Build GRCh38
rs376572772 Chr.8: 22099359-22099359 on Build GRCh38
rs1049689528 Chr.8: 22099647-22099647 on Build GRCh38
rs777943499 Chr.8: 22099508-22099508 on Build GRCh38
rs28429870 Chr.8: 22099464-22099464 on Build GRCh38
rs748028644 Chr.8: 22099363-22099363 on Build GRCh38
rs920576297 Chr.8: 22099534-22099534 on Build GRCh38
rs374658483 Chr.8: 22099254-22099254 on Build GRCh38
rs981457064 Chr.8: 22099549-22099549 on Build GRCh38
rs958925301 Chr.8: 22099550-22099550 on Build GRCh38
rs765528251 Chr.8: 22099236-22099236 on Build GRCh38
rs901525375 Chr.8: 22099224-22099224 on Build GRCh38
rs566430758 Chr.8: 22099591-22099591 on Build GRCh38
rs775982731 Chr.8: 22099401-22099401 on Build GRCh38
rs757797244 Chr.8: 22099544-22099544 on Build GRCh38
rs368750146 Chr.8: 22099267-22099267 on Build GRCh38
rs909350028 Chr.8: 22099585-22099585 on Build GRCh38
rs768043869 Chr.8: 22099320-22099320 on Build GRCh38
rs766696138 Chr.8: 22099258-22099258 on Build GRCh38
rs762557587 Chr.8: 22099410-22099410 on Build GRCh38
rs776632983 Chr.8: 22099212-22099212 on Build GRCh38
rs756756486 Chr.8: 22099322-22099322 on Build GRCh38
rs1011053149 Chr.8: 22099375-22099375 on Build GRCh38
rs774736068 Chr.8: 22099308-22099308 on Build GRCh38
rs753312831 Chr.8: 22099262-22099262 on Build GRCh38
rs752017375 Chr.8: 22099259-22099259 on Build GRCh38
rs552233996 Chr.8: 22099628-22099628 on Build GRCh38
rs901098829 Chr.8: 22099334-22099334 on Build GRCh38
rs533744687 Chr.8: 22099606-22099606 on Build GRCh38
rs764001273 Chr.8: 22099211-22099211 on Build GRCh38
rs753402658 Chr.8: 22099321-22099321 on Build GRCh38
rs547990397 Chr.8: 22099545-22099545 on Build GRCh38

View Full Product Details