Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22191967-22192460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP1
Entrez Gene ID
649
Gene Name
bone morphogenetic protein 1
Gene Aliases
OI13, PCOLC, PCP, PCP2, TLD
UniGene
Hs.1274
Gene Chromosome Location
Chr.8: 22165140-22212326 on Build GRCh38
Transcripts
NM_001199.3 NM_006129.4 NR_033403.1 NR_033404.1 XM_006716386.2 XM_011544617.1 XM_017013738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6342882 c.1112G>T p.R371L Chr.8: 22192083-22192083 on Build GRCh38
5954191 c.1087A>T p.N363Y Chr.8: 22192058-22192058 on Build GRCh38
1098049 c.1111C>T p.R371C Chr.8: 22192082-22192082 on Build GRCh38
5606351 c.1097C>T p.S366F Chr.8: 22192068-22192068 on Build GRCh38
4644852 c.1101G>A p.L367L Chr.8: 22192072-22192072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569211148 Chr.8: 22192352-22192352 on Build GRCh38
rs751032725 Chr.8: 22192034-22192034 on Build GRCh38
rs751139262 Chr.8: 22192109-22192109 on Build GRCh38
rs3924229 Chr.8: 22192379-22192379 on Build GRCh38
rs547186279 Chr.8: 22192169-22192169 on Build GRCh38
rs564392614 Chr.8: 22192102-22192102 on Build GRCh38
rs754163512 Chr.8: 22192098-22192098 on Build GRCh38
rs759048351 Chr.8: 22192203-22192203 on Build GRCh38
rs749096543 Chr.8: 22192073-22192073 on Build GRCh38
rs1052288647 Chr.8: 22192356-22192356 on Build GRCh38
rs372145084 Chr.8: 22192001-22192001 on Build GRCh38
rs756044980 Chr.8: 22192120-22192120 on Build GRCh38
rs774416710 Chr.8: 22192082-22192082 on Build GRCh38
rs76021885 Chr.8: 22192141-22192141 on Build GRCh38
rs746069533 Chr.8: 22192148-22192148 on Build GRCh38
rs73225860 Chr.8: 22192260-22192260 on Build GRCh38
rs559736255 Chr.8: 22192024-22192024 on Build GRCh38
rs3924230 Chr.8: 22192327-22192327 on Build GRCh38
rs749231859 Chr.8: 22192139-22192140 on Build GRCh38
rs1041472997 Chr.8: 22192137-22192137 on Build GRCh38
rs754422233 Chr.8: 22192035-22192035 on Build GRCh38
rs762421703 Chr.8: 22192164-22192164 on Build GRCh38
rs765681002 Chr.8: 22192105-22192105 on Build GRCh38
rs370077045 Chr.8: 22192168-22192168 on Build GRCh38
rs1022174394 Chr.8: 22192417-22192417 on Build GRCh38
rs777102639 Chr.8: 22192160-22192160 on Build GRCh38
rs771141447 Chr.8: 22192075-22192075 on Build GRCh38
rs774419621 Chr.8: 22192006-22192006 on Build GRCh38
rs777882480 Chr.8: 22192138-22192138 on Build GRCh38
rs555586276 Chr.8: 22192393-22192393 on Build GRCh38
rs3924231 Chr.8: 22192267-22192267 on Build GRCh38
rs757978831 Chr.8: 22192025-22192025 on Build GRCh38
rs1008515972 Chr.8: 22192404-22192404 on Build GRCh38
rs747520920 Chr.8: 22192072-22192072 on Build GRCh38
rs375180557 Chr.8: 22192089-22192089 on Build GRCh38
rs774387209 Chr.8: 22192147-22192148 on Build GRCh38
rs1026031602 Chr.8: 22192004-22192004 on Build GRCh38
rs945755549 Chr.8: 22192103-22192103 on Build GRCh38
rs990427456 Chr.8: 22192416-22192416 on Build GRCh38
rs370226855 Chr.8: 22192005-22192005 on Build GRCh38
rs145284541 Chr.8: 22192083-22192083 on Build GRCh38
rs759563496 Chr.8: 22192008-22192008 on Build GRCh38
rs112001602 Chr.8: 22192012-22192012 on Build GRCh38
rs756685154 Chr.8: 22192018-22192018 on Build GRCh38
rs879092987 Chr.8: 22192110-22192110 on Build GRCh38
rs187520032 Chr.8: 22192431-22192431 on Build GRCh38
rs775608544 Chr.8: 22192088-22192088 on Build GRCh38
rs768695286 Chr.8: 22192362-22192362 on Build GRCh38
rs776642777 Chr.8: 22192108-22192108 on Build GRCh38
rs1037909422 Chr.8: 22192250-22192250 on Build GRCh38
rs748093647 Chr.8: 22192065-22192065 on Build GRCh38
rs772355766 Chr.8: 22192149-22192149 on Build GRCh38
rs779606580 Chr.8: 22192032-22192032 on Build GRCh38
rs370731778 Chr.8: 22192192-22192192 on Build GRCh38
rs780127226 Chr.8: 22192326-22192326 on Build GRCh38
rs941970276 Chr.8: 22192206-22192206 on Build GRCh38
rs749040090 Chr.8: 22192140-22192140 on Build GRCh38
rs754727375 Chr.8: 22192118-22192118 on Build GRCh38
rs769104277 Chr.8: 22192159-22192159 on Build GRCh38
rs1055623945 Chr.8: 22192184-22192184 on Build GRCh38
rs767687661 Chr.8: 22192011-22192011 on Build GRCh38
rs116922460 Chr.8: 22192036-22192036 on Build GRCh38
rs546434435 Chr.8: 22192094-22192094 on Build GRCh38
rs550032501 Chr.8: 22192142-22192142 on Build GRCh38
rs771932751 Chr.8: 22192190-22192190 on Build GRCh38
rs772044431 Chr.8: 22192086-22192086 on Build GRCh38
rs535579955 Chr.8: 22192157-22192157 on Build GRCh38
rs557995282 Chr.8: 22192307-22192307 on Build GRCh38
rs368266318 Chr.8: 22192129-22192129 on Build GRCh38
rs370391413 Chr.8: 22192066-22192066 on Build GRCh38
rs768657002 Chr.8: 22192142-22192143 on Build GRCh38
rs752265892 Chr.8: 22192119-22192119 on Build GRCh38
rs28362095 Chr.8: 22192156-22192156 on Build GRCh38
rs764854135 Chr.8: 22192379-22192379 on Build GRCh38

View Full Product Details