Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22621031-22621573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
-
UniGene
Hs.433722
Gene Chromosome Location
Chr.8: 22620418-22669148 on Build GRCh38
Transcripts
NM_018688.4 XM_005273573.3 XM_011544586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1456118 c.688G>A p.D230N Chr.8: 22621496-22621496 on Build GRCh38
7201670 c.719A>C p.K240T Chr.8: 22621465-22621465 on Build GRCh38
3698712 c.623A>G p.Y208C Chr.8: 22621561-22621561 on Build GRCh38
1098144 c.741C>T p.L247L Chr.8: 22621443-22621443 on Build GRCh38
6020604 c.664C>T p.Q222* Chr.8: 22621520-22621520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963389537 Chr.8: 22621413-22621413 on Build GRCh38
rs779840454 Chr.8: 22621458-22621458 on Build GRCh38
rs994567343 Chr.8: 22621430-22621431 on Build GRCh38
rs1047360499 Chr.8: 22621157-22621157 on Build GRCh38
rs114738244 Chr.8: 22621304-22621304 on Build GRCh38
rs551008362 Chr.8: 22621522-22621522 on Build GRCh38
rs1011821298 Chr.8: 22621212-22621212 on Build GRCh38
rs945506665 Chr.8: 22621196-22621196 on Build GRCh38
rs756507630 Chr.8: 22621504-22621504 on Build GRCh38
rs199884901 Chr.8: 22621446-22621446 on Build GRCh38
rs553792477 Chr.8: 22621243-22621245 on Build GRCh38
rs748885813 Chr.8: 22621382-22621382 on Build GRCh38
rs367825147 Chr.8: 22621398-22621398 on Build GRCh38
rs536823242 Chr.8: 22621169-22621169 on Build GRCh38
rs774303843 Chr.8: 22621486-22621486 on Build GRCh38
rs747619153 Chr.8: 22621454-22621454 on Build GRCh38
rs1025047455 Chr.8: 22621071-22621071 on Build GRCh38
rs1040178056 Chr.8: 22621193-22621193 on Build GRCh38
rs530633458 Chr.8: 22621309-22621309 on Build GRCh38
rs974308689 Chr.8: 22621133-22621133 on Build GRCh38
rs750146463 Chr.8: 22621513-22621513 on Build GRCh38
rs575499906 Chr.8: 22621440-22621440 on Build GRCh38
rs778350247 Chr.8: 22621391-22621391 on Build GRCh38
rs183928789 Chr.8: 22621303-22621303 on Build GRCh38
rs774267367 Chr.8: 22621109-22621109 on Build GRCh38
rs899832795 Chr.8: 22621311-22621311 on Build GRCh38
rs965116144 Chr.8: 22621335-22621335 on Build GRCh38
rs754894198 Chr.8: 22621532-22621533 on Build GRCh38
rs945537606 Chr.8: 22621415-22621415 on Build GRCh38
rs769767366 Chr.8: 22621525-22621525 on Build GRCh38
rs973159375 Chr.8: 22621254-22621254 on Build GRCh38
rs763135160 Chr.8: 22621430-22621430 on Build GRCh38
rs144962194 Chr.8: 22621077-22621077 on Build GRCh38
rs540265828 Chr.8: 22621188-22621188 on Build GRCh38
rs752169219 Chr.8: 22621434-22621434 on Build GRCh38
rs754276468 Chr.8: 22621365-22621365 on Build GRCh38
rs912290032 Chr.8: 22621141-22621141 on Build GRCh38
rs386723478 Chr.8: 22621078-22621079 on Build GRCh38
rs762087866 Chr.8: 22621417-22621417 on Build GRCh38
rs557632496 Chr.8: 22621149-22621149 on Build GRCh38
rs753873761 Chr.8: 22621485-22621490 on Build GRCh38
rs747950725 Chr.8: 22621524-22621524 on Build GRCh38
rs767672797 Chr.8: 22621476-22621481 on Build GRCh38
rs745336812 Chr.8: 22621451-22621451 on Build GRCh38
rs750509864 Chr.8: 22621428-22621428 on Build GRCh38
rs1033726230 Chr.8: 22621247-22621247 on Build GRCh38
rs371797360 Chr.8: 22621445-22621445 on Build GRCh38
rs951204540 Chr.8: 22621118-22621118 on Build GRCh38
rs765579313 Chr.8: 22621425-22621425 on Build GRCh38
rs973959813 Chr.8: 22621110-22621110 on Build GRCh38
rs776519160 Chr.8: 22621472-22621472 on Build GRCh38
rs757867694 Chr.8: 22621480-22621480 on Build GRCh38
rs202037271 Chr.8: 22621279-22621281 on Build GRCh38
rs943715051 Chr.8: 22621142-22621142 on Build GRCh38
rs7001041 Chr.8: 22621172-22621172 on Build GRCh38
rs771677112 Chr.8: 22621452-22621452 on Build GRCh38
rs1046932813 Chr.8: 22621200-22621200 on Build GRCh38
rs770111674 Chr.8: 22621476-22621476 on Build GRCh38
rs779548998 Chr.8: 22621517-22621517 on Build GRCh38
rs7008429 Chr.8: 22621079-22621079 on Build GRCh38
rs767113089 Chr.8: 22621081-22621081 on Build GRCh38
rs113002231 Chr.8: 22621065-22621065 on Build GRCh38
rs377103262 Chr.8: 22621475-22621475 on Build GRCh38
rs1009687155 Chr.8: 22621056-22621056 on Build GRCh38
rs368026258 Chr.8: 22621460-22621460 on Build GRCh38
rs975563911 Chr.8: 22621340-22621340 on Build GRCh38
rs913890052 Chr.8: 22621510-22621511 on Build GRCh38
rs982518775 Chr.8: 22621136-22621136 on Build GRCh38
rs746437989 Chr.8: 22621487-22621487 on Build GRCh38
rs777281096 Chr.8: 22621381-22621381 on Build GRCh38
rs765925631 Chr.8: 22621173-22621173 on Build GRCh38
rs573268922 Chr.8: 22621180-22621180 on Build GRCh38
rs989751475 Chr.8: 22621373-22621373 on Build GRCh38
rs991622349 Chr.8: 22621106-22621106 on Build GRCh38
rs549133253 Chr.8: 22621314-22621314 on Build GRCh38
rs771085784 Chr.8: 22621530-22621530 on Build GRCh38
rs201299945 Chr.8: 22621390-22621390 on Build GRCh38
rs767983651 Chr.8: 22621545-22621545 on Build GRCh38
rs781688409 Chr.8: 22621362-22621362 on Build GRCh38
rs1035564550 Chr.8: 22621363-22621363 on Build GRCh38
rs995412507 Chr.8: 22621281-22621281 on Build GRCh38
rs1002619845 Chr.8: 22621245-22621245 on Build GRCh38
rs930148626 Chr.8: 22621266-22621266 on Build GRCh38
rs540896817 Chr.8: 22621371-22621371 on Build GRCh38
rs759608287 Chr.8: 22621124-22621124 on Build GRCh38
rs1011435350 Chr.8: 22621076-22621076 on Build GRCh38
rs774444271 Chr.8: 22621532-22621532 on Build GRCh38
rs779546029 Chr.8: 22621496-22621496 on Build GRCh38
rs371953031 Chr.8: 22621473-22621473 on Build GRCh38
rs775535509 Chr.8: 22621400-22621400 on Build GRCh38
rs965617197 Chr.8: 22621471-22621471 on Build GRCh38
rs746906528 Chr.8: 22621459-22621459 on Build GRCh38
rs778582977 Chr.8: 22621450-22621450 on Build GRCh38
rs1021374296 Chr.8: 22621219-22621219 on Build GRCh38
rs759588067 Chr.8: 22621536-22621536 on Build GRCh38
rs757999867 Chr.8: 22621514-22621514 on Build GRCh38
rs936305858 Chr.8: 22621367-22621367 on Build GRCh38
rs1043135079 Chr.8: 22621327-22621328 on Build GRCh38
rs138098724 Chr.8: 22621150-22621150 on Build GRCh38
rs539327444 Chr.8: 22621096-22621096 on Build GRCh38
rs571889678 Chr.8: 22621244-22621244 on Build GRCh38
rs1026809839 Chr.8: 22621111-22621111 on Build GRCh38
rs577564671 Chr.8: 22621260-22621260 on Build GRCh38
rs762049020 Chr.8: 22621455-22621458 on Build GRCh38
rs569508767 Chr.8: 22621527-22621527 on Build GRCh38
rs975071881 Chr.8: 22621147-22621147 on Build GRCh38
rs763306549 Chr.8: 22621391-22621392 on Build GRCh38
rs7000993 Chr.8: 22621078-22621078 on Build GRCh38
rs963122480 Chr.8: 22621254-22621255 on Build GRCh38
rs982549901 Chr.8: 22621305-22621305 on Build GRCh38
rs998699597 Chr.8: 22621221-22621221 on Build GRCh38
rs764862873 Chr.8: 22621507-22621507 on Build GRCh38
rs528176697 Chr.8: 22621416-22621416 on Build GRCh38
rs754694799 Chr.8: 22621257-22621257 on Build GRCh38
rs369519533 Chr.8: 22621481-22621481 on Build GRCh38
rs201665640 Chr.8: 22621408-22621408 on Build GRCh38
rs751815160 Chr.8: 22621415-22621416 on Build GRCh38
rs755457330 Chr.8: 22621435-22621435 on Build GRCh38
rs746946195 Chr.8: 22621534-22621534 on Build GRCh38
rs959745989 Chr.8: 22621089-22621089 on Build GRCh38
rs764232507 Chr.8: 22621416-22621416 on Build GRCh38
rs376718168 Chr.8: 22621529-22621529 on Build GRCh38
rs766793980 Chr.8: 22621431-22621431 on Build GRCh38
rs760381282 Chr.8: 22621402-22621402 on Build GRCh38
rs187678910 Chr.8: 22621497-22621497 on Build GRCh38
rs1019789946 Chr.8: 22621312-22621312 on Build GRCh38
rs766426655 Chr.8: 22621485-22621485 on Build GRCh38

View Full Product Details