Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22630300-22630799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
-
UniGene
Hs.433722
Gene Chromosome Location
Chr.8: 22620418-22669148 on Build GRCh38
Transcripts
NM_018688.4 XM_005273573.3 XM_011544586.2 XM_011544587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297667 c.271C>A p.R91R Chr.8: 22630468-22630468 on Build GRCh38
3995980 c.235C>T p.L79L Chr.8: 22630504-22630504 on Build GRCh38
750065 c.177C>T p.A59A Chr.8: 22630562-22630562 on Build GRCh38
1623862 c.171A>G p.K57K Chr.8: 22630568-22630568 on Build GRCh38
238965 c.197_198insA p.L67fs*7 Chr.8: 22630541-22630542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924139171 Chr.8: 22630448-22630448 on Build GRCh38
rs180986552 Chr.8: 22630703-22630703 on Build GRCh38
rs887444339 Chr.8: 22630668-22630668 on Build GRCh38
rs554733472 Chr.8: 22630358-22630358 on Build GRCh38
rs536424714 Chr.8: 22630355-22630355 on Build GRCh38
rs574780680 Chr.8: 22630711-22630711 on Build GRCh38
rs761087568 Chr.8: 22630422-22630422 on Build GRCh38
rs773968128 Chr.8: 22630440-22630440 on Build GRCh38
rs370739500 Chr.8: 22630490-22630490 on Build GRCh38
rs575526587 Chr.8: 22630354-22630354 on Build GRCh38
rs774842689 Chr.8: 22630561-22630561 on Build GRCh38
rs1029815259 Chr.8: 22630502-22630502 on Build GRCh38
rs749722833 Chr.8: 22630618-22630618 on Build GRCh38
rs143121934 Chr.8: 22630635-22630635 on Build GRCh38
rs1048930651 Chr.8: 22630667-22630667 on Build GRCh38
rs771177103 Chr.8: 22630397-22630397 on Build GRCh38
rs373245133 Chr.8: 22630454-22630454 on Build GRCh38
rs757994252 Chr.8: 22630343-22630343 on Build GRCh38
rs372191525 Chr.8: 22630576-22630576 on Build GRCh38
rs897091899 Chr.8: 22630715-22630715 on Build GRCh38
rs560525260 Chr.8: 22630773-22630773 on Build GRCh38
rs374643435 Chr.8: 22630417-22630417 on Build GRCh38
rs189374699 Chr.8: 22630699-22630699 on Build GRCh38
rs750251527 Chr.8: 22630585-22630585 on Build GRCh38
rs374423428 Chr.8: 22630526-22630526 on Build GRCh38
rs181817464 Chr.8: 22630467-22630467 on Build GRCh38
rs922383503 Chr.8: 22630395-22630395 on Build GRCh38
rs773712734 Chr.8: 22630543-22630543 on Build GRCh38
rs747408772 Chr.8: 22630478-22630478 on Build GRCh38
rs372090933 Chr.8: 22630499-22630499 on Build GRCh38
rs558586590 Chr.8: 22630698-22630698 on Build GRCh38
rs779149953 Chr.8: 22630468-22630468 on Build GRCh38
rs367655750 Chr.8: 22630562-22630562 on Build GRCh38
rs550917215 Chr.8: 22630597-22630597 on Build GRCh38
rs746001140 Chr.8: 22630406-22630406 on Build GRCh38
rs937271368 Chr.8: 22630666-22630666 on Build GRCh38
rs758626086 Chr.8: 22630530-22630530 on Build GRCh38
rs763759647 Chr.8: 22630466-22630466 on Build GRCh38
rs1004541733 Chr.8: 22630673-22630673 on Build GRCh38
rs753183991 Chr.8: 22630608-22630608 on Build GRCh38
rs533659223 Chr.8: 22630656-22630656 on Build GRCh38
rs566615423 Chr.8: 22630648-22630648 on Build GRCh38
rs983138370 Chr.8: 22630325-22630325 on Build GRCh38
rs762463420 Chr.8: 22630429-22630429 on Build GRCh38
rs1045997111 Chr.8: 22630658-22630658 on Build GRCh38
rs527782919 Chr.8: 22630536-22630536 on Build GRCh38
rs770222198 Chr.8: 22630542-22630542 on Build GRCh38
rs369708307 Chr.8: 22630604-22630604 on Build GRCh38
rs777059512 Chr.8: 22630426-22630426 on Build GRCh38
rs774200897 Chr.8: 22630489-22630489 on Build GRCh38
rs572835721 Chr.8: 22630359-22630359 on Build GRCh38
rs542000407 Chr.8: 22630514-22630514 on Build GRCh38
rs764946714 Chr.8: 22630577-22630577 on Build GRCh38
rs889587116 Chr.8: 22630749-22630749 on Build GRCh38
rs544518387 Chr.8: 22630470-22630470 on Build GRCh38
rs1030258098 Chr.8: 22630709-22630709 on Build GRCh38
rs772539761 Chr.8: 22630416-22630416 on Build GRCh38
rs769057763 Chr.8: 22630475-22630486 on Build GRCh38
rs755406356 Chr.8: 22630535-22630535 on Build GRCh38
rs562812650 Chr.8: 22630487-22630487 on Build GRCh38
rs905671866 Chr.8: 22630662-22630662 on Build GRCh38
rs1042588299 Chr.8: 22630441-22630441 on Build GRCh38
rs373934216 Chr.8: 22630474-22630474 on Build GRCh38
rs772792835 Chr.8: 22630720-22630720 on Build GRCh38
rs762042803 Chr.8: 22630480-22630480 on Build GRCh38
rs779827419 Chr.8: 22630628-22630628 on Build GRCh38
rs961807379 Chr.8: 22630348-22630348 on Build GRCh38
rs368195300 Chr.8: 22630533-22630533 on Build GRCh38
rs34841678 Chr.8: 22630717-22630718 on Build GRCh38
rs569151863 Chr.8: 22630616-22630616 on Build GRCh38
rs939612543 Chr.8: 22630649-22630649 on Build GRCh38
rs988839378 Chr.8: 22630765-22630765 on Build GRCh38
rs768477513 Chr.8: 22630570-22630570 on Build GRCh38
rs976531514 Chr.8: 22630385-22630385 on Build GRCh38
rs1012294318 Chr.8: 22630327-22630327 on Build GRCh38
rs977356267 Chr.8: 22630500-22630500 on Build GRCh38
rs187399704 Chr.8: 22630598-22630598 on Build GRCh38
rs77509180 Chr.8: 22630437-22630437 on Build GRCh38
rs978371577 Chr.8: 22630612-22630612 on Build GRCh38
rs779958916 Chr.8: 22630531-22630531 on Build GRCh38
rs542067788 Chr.8: 22630736-22630736 on Build GRCh38
rs147639286 Chr.8: 22630361-22630361 on Build GRCh38
rs1015446247 Chr.8: 22630367-22630367 on Build GRCh38
rs776836042 Chr.8: 22630479-22630479 on Build GRCh38
rs745475111 Chr.8: 22630549-22630549 on Build GRCh38
rs368613631 Chr.8: 22630481-22630481 on Build GRCh38
rs769092968 Chr.8: 22630364-22630364 on Build GRCh38
rs554463760 Chr.8: 22630636-22630636 on Build GRCh38
rs73671219 Chr.8: 22630491-22630491 on Build GRCh38
rs746727116 Chr.8: 22630387-22630387 on Build GRCh38
rs916737423 Chr.8: 22630670-22630670 on Build GRCh38
rs543781590 Chr.8: 22630702-22630702 on Build GRCh38
rs777138191 Chr.8: 22630357-22630357 on Build GRCh38
rs757114130 Chr.8: 22630520-22630520 on Build GRCh38
rs750586585 Chr.8: 22630529-22630529 on Build GRCh38
rs925922729 Chr.8: 22630390-22630390 on Build GRCh38
rs770146937 Chr.8: 22630672-22630672 on Build GRCh38
rs376077176 Chr.8: 22630463-22630463 on Build GRCh38
rs780198309 Chr.8: 22630476-22630476 on Build GRCh38
rs570768198 Chr.8: 22630572-22630572 on Build GRCh38
rs771775272 Chr.8: 22630553-22630553 on Build GRCh38
rs1036357643 Chr.8: 22630712-22630712 on Build GRCh38
rs967650790 Chr.8: 22630610-22630610 on Build GRCh38
rs753550217 Chr.8: 22630519-22630519 on Build GRCh38
rs369523953 Chr.8: 22630404-22630404 on Build GRCh38
rs748508920 Chr.8: 22630538-22630538 on Build GRCh38
rs759183445 Chr.8: 22630452-22630452 on Build GRCh38
rs1029926970 Chr.8: 22630729-22630729 on Build GRCh38
rs961474098 Chr.8: 22630371-22630371 on Build GRCh38
rs777216711 Chr.8: 22630427-22630427 on Build GRCh38
rs756718303 Chr.8: 22630609-22630609 on Build GRCh38
rs377717883 Chr.8: 22630423-22630423 on Build GRCh38
rs954462748 Chr.8: 22630532-22630532 on Build GRCh38

View Full Product Details