Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26643359-26643857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL2
Entrez Gene ID
1808
Gene Name
dihydropyrimidinase like 2
Gene Aliases
CRMP-2, CRMP2, DHPRP2, DRP-2, DRP2, N2A3, ULIP-2, ULIP2
UniGene
Hs.593187
Gene Chromosome Location
Chr.8: 26514193-26658177 on Build GRCh38
Transcripts
NM_001197293.2 NM_001244604.1 NM_001386.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463016 c.956C>T p.S319F Chr.8: 26643583-26643583 on Build GRCh38
5946677 c.831G>A p.E277E Chr.8: 26643458-26643458 on Build GRCh38
6484325 c.879G>C p.K293N Chr.8: 26643506-26643506 on Build GRCh38
5932218 c.914C>T p.P305L Chr.8: 26643541-26643541 on Build GRCh38
4462124 c.918C>T p.P306P Chr.8: 26643545-26643545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756229425 Chr.8: 26643519-26643519 on Build GRCh38
rs753361864 Chr.8: 26643430-26643430 on Build GRCh38
rs564872054 Chr.8: 26643643-26643643 on Build GRCh38
rs919343738 Chr.8: 26643775-26643775 on Build GRCh38
rs559093954 Chr.8: 26643436-26643436 on Build GRCh38
rs750802287 Chr.8: 26643397-26643397 on Build GRCh38
rs371024991 Chr.8: 26643420-26643420 on Build GRCh38
rs374871574 Chr.8: 26643624-26643624 on Build GRCh38
rs747717094 Chr.8: 26643465-26643465 on Build GRCh38
rs958407363 Chr.8: 26643479-26643479 on Build GRCh38
rs563202767 Chr.8: 26643608-26643608 on Build GRCh38
rs777753654 Chr.8: 26643562-26643562 on Build GRCh38
rs767986872 Chr.8: 26643455-26643455 on Build GRCh38
rs375095984 Chr.8: 26643534-26643534 on Build GRCh38
rs776011503 Chr.8: 26643638-26643638 on Build GRCh38
rs1002467961 Chr.8: 26643675-26643675 on Build GRCh38
rs776278802 Chr.8: 26643461-26643461 on Build GRCh38
rs187707246 Chr.8: 26643404-26643404 on Build GRCh38
rs1002411548 Chr.8: 26643672-26643672 on Build GRCh38
rs746848520 Chr.8: 26643587-26643587 on Build GRCh38
rs993537933 Chr.8: 26643737-26643737 on Build GRCh38
rs1013991435 Chr.8: 26643705-26643705 on Build GRCh38
rs138088405 Chr.8: 26643485-26643485 on Build GRCh38
rs377086745 Chr.8: 26643399-26643399 on Build GRCh38
rs966296600 Chr.8: 26643805-26643805 on Build GRCh38
rs774770456 Chr.8: 26643620-26643620 on Build GRCh38
rs762658062 Chr.8: 26643743-26643743 on Build GRCh38
rs200846358 Chr.8: 26643482-26643482 on Build GRCh38
rs781260302 Chr.8: 26643406-26643406 on Build GRCh38
rs137931500 Chr.8: 26643730-26643730 on Build GRCh38
rs765051202 Chr.8: 26643396-26643396 on Build GRCh38
rs975960521 Chr.8: 26643819-26643819 on Build GRCh38
rs972501088 Chr.8: 26643733-26643733 on Build GRCh38
rs566891679 Chr.8: 26643783-26643783 on Build GRCh38
rs190693812 Chr.8: 26643410-26643410 on Build GRCh38
rs1040889867 Chr.8: 26643625-26643625 on Build GRCh38
rs754954889 Chr.8: 26643592-26643592 on Build GRCh38
rs910387418 Chr.8: 26643792-26643792 on Build GRCh38
rs7841384 Chr.8: 26643835-26643835 on Build GRCh38
rs771130921 Chr.8: 26643618-26643618 on Build GRCh38
rs747628014 Chr.8: 26643607-26643607 on Build GRCh38
rs746980657 Chr.8: 26643444-26643444 on Build GRCh38
rs752846351 Chr.8: 26643407-26643407 on Build GRCh38
rs1053236613 Chr.8: 26643836-26643836 on Build GRCh38
rs904077604 Chr.8: 26643663-26643663 on Build GRCh38
rs1055218217 Chr.8: 26643684-26643684 on Build GRCh38
rs945344314 Chr.8: 26643661-26643661 on Build GRCh38
rs368393366 Chr.8: 26643656-26643656 on Build GRCh38
rs149525532 Chr.8: 26643492-26643492 on Build GRCh38
rs779903275 Chr.8: 26643584-26643584 on Build GRCh38
rs767983441 Chr.8: 26643634-26643634 on Build GRCh38
rs757320445 Chr.8: 26643676-26643676 on Build GRCh38
rs199869101 Chr.8: 26643481-26643481 on Build GRCh38
rs764265785 Chr.8: 26643527-26643527 on Build GRCh38
rs563371941 Chr.8: 26643403-26643403 on Build GRCh38
rs752754362 Chr.8: 26643518-26643518 on Build GRCh38
rs775801065 Chr.8: 26643831-26643831 on Build GRCh38
rs372408415 Chr.8: 26643502-26643502 on Build GRCh38
rs192797599 Chr.8: 26643791-26643791 on Build GRCh38
rs896680961 Chr.8: 26643691-26643691 on Build GRCh38
rs1022995013 Chr.8: 26643401-26643401 on Build GRCh38
rs980728856 Chr.8: 26643393-26643393 on Build GRCh38
rs758071708 Chr.8: 26643563-26643563 on Build GRCh38
rs184841156 Chr.8: 26643828-26643828 on Build GRCh38
rs972158643 Chr.8: 26643423-26643423 on Build GRCh38
rs187445527 Chr.8: 26643653-26643653 on Build GRCh38
rs146185684 Chr.8: 26643725-26643725 on Build GRCh38
rs748979344 Chr.8: 26643613-26643613 on Build GRCh38
rs112393878 Chr.8: 26643437-26643437 on Build GRCh38
rs969594149 Chr.8: 26643662-26643662 on Build GRCh38
rs778021084 Chr.8: 26643417-26643417 on Build GRCh38
rs943261359 Chr.8: 26643798-26643798 on Build GRCh38
rs536359115 Chr.8: 26643740-26643740 on Build GRCh38
rs551279053 Chr.8: 26643603-26643603 on Build GRCh38
rs768077249 Chr.8: 26643503-26643503 on Build GRCh38
rs970082230 Chr.8: 26643380-26643380 on Build GRCh38
rs925897462 Chr.8: 26643832-26643832 on Build GRCh38
rs546928356 Chr.8: 26643767-26643767 on Build GRCh38
rs766765018 Chr.8: 26643400-26643400 on Build GRCh38

View Full Product Details