Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144356901-144357403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5672962 c.693C>T p.H231H Chr.8: 144357068-144357068 on Build GRCh38
6661981 c.640-4G>A p.? Chr.8: 144357125-144357125 on Build GRCh38
3898188 c.696T>C p.G232G Chr.8: 144357065-144357065 on Build GRCh38
5531100 c.745C>T p.H249Y Chr.8: 144357016-144357016 on Build GRCh38
6050112 c.662G>T p.R221L Chr.8: 144357099-144357099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782070332 Chr.8: 144356953-144356953 on Build GRCh38
rs782383000 Chr.8: 144357044-144357044 on Build GRCh38
rs750016818 Chr.8: 144357322-144357322 on Build GRCh38
rs376850707 Chr.8: 144357124-144357124 on Build GRCh38
rs530584730 Chr.8: 144357269-144357269 on Build GRCh38
rs782155174 Chr.8: 144357052-144357052 on Build GRCh38
rs782189197 Chr.8: 144357106-144357106 on Build GRCh38
rs782653376 Chr.8: 144357194-144357194 on Build GRCh38
rs782480866 Chr.8: 144357004-144357004 on Build GRCh38
rs890712361 Chr.8: 144357238-144357238 on Build GRCh38
rs150030093 Chr.8: 144357096-144357096 on Build GRCh38
rs782111680 Chr.8: 144356956-144356957 on Build GRCh38
rs782559616 Chr.8: 144357086-144357086 on Build GRCh38
rs559480601 Chr.8: 144357216-144357216 on Build GRCh38
rs782222157 Chr.8: 144356934-144356934 on Build GRCh38
rs139330417 Chr.8: 144357076-144357076 on Build GRCh38
rs782776424 Chr.8: 144356982-144356982 on Build GRCh38
rs1031898859 Chr.8: 144356999-144356999 on Build GRCh38
rs369704521 Chr.8: 144356927-144356927 on Build GRCh38
rs200079529 Chr.8: 144357011-144357011 on Build GRCh38
rs373298729 Chr.8: 144357068-144357068 on Build GRCh38
rs782662505 Chr.8: 144356928-144356928 on Build GRCh38
rs1053807394 Chr.8: 144357231-144357231 on Build GRCh38
rs115001185 Chr.8: 144357345-144357345 on Build GRCh38
rs782173457 Chr.8: 144357020-144357020 on Build GRCh38
rs782464019 Chr.8: 144357024-144357025 on Build GRCh38
rs1009525003 Chr.8: 144357292-144357292 on Build GRCh38
rs375647211 Chr.8: 144356925-144356925 on Build GRCh38
rs530145500 Chr.8: 144357221-144357221 on Build GRCh38
rs782262513 Chr.8: 144357008-144357008 on Build GRCh38
rs2272666 Chr.8: 144357324-144357324 on Build GRCh38
rs782104865 Chr.8: 144357129-144357129 on Build GRCh38
rs376025805 Chr.8: 144356983-144356983 on Build GRCh38
rs782326012 Chr.8: 144357033-144357033 on Build GRCh38
rs781905056 Chr.8: 144357080-144357080 on Build GRCh38
rs782625066 Chr.8: 144356924-144356924 on Build GRCh38
rs781803849 Chr.8: 144357148-144357148 on Build GRCh38
rs149208512 Chr.8: 144357100-144357100 on Build GRCh38
rs372382267 Chr.8: 144356980-144356980 on Build GRCh38
rs781839444 Chr.8: 144357043-144357043 on Build GRCh38
rs368962350 Chr.8: 144356970-144356970 on Build GRCh38
rs368173186 Chr.8: 144357172-144357172 on Build GRCh38
rs146812670 Chr.8: 144357055-144357055 on Build GRCh38
rs781922743 Chr.8: 144356951-144356951 on Build GRCh38
rs782785721 Chr.8: 144357131-144357131 on Build GRCh38
rs1009060977 Chr.8: 144357039-144357039 on Build GRCh38
rs955068843 Chr.8: 144357009-144357009 on Build GRCh38
rs139706818 Chr.8: 144356943-144356943 on Build GRCh38
rs552672053 Chr.8: 144356941-144356941 on Build GRCh38
rs920276321 Chr.8: 144357074-144357074 on Build GRCh38
rs782173327 Chr.8: 144356931-144356931 on Build GRCh38
rs781887898 Chr.8: 144356992-144356993 on Build GRCh38
rs782011297 Chr.8: 144357051-144357051 on Build GRCh38
rs782589898 Chr.8: 144356990-144356990 on Build GRCh38
rs370978282 Chr.8: 144357191-144357191 on Build GRCh38
rs782565511 Chr.8: 144357165-144357166 on Build GRCh38
rs377254762 Chr.8: 144357348-144357348 on Build GRCh38
rs782433614 Chr.8: 144357161-144357161 on Build GRCh38
rs782519875 Chr.8: 144357164-144357164 on Build GRCh38
rs781816444 Chr.8: 144356994-144356994 on Build GRCh38
rs569537166 Chr.8: 144357257-144357257 on Build GRCh38
rs541935911 Chr.8: 144356948-144356948 on Build GRCh38
rs541464341 Chr.8: 144357163-144357163 on Build GRCh38
rs782303523 Chr.8: 144357167-144357167 on Build GRCh38
rs781930248 Chr.8: 144357059-144357059 on Build GRCh38
rs782526062 Chr.8: 144357102-144357102 on Build GRCh38
rs548231491 Chr.8: 144357252-144357252 on Build GRCh38
rs782443480 Chr.8: 144357169-144357169 on Build GRCh38
rs1138056 Chr.8: 144357121-144357121 on Build GRCh38
rs782730125 Chr.8: 144356957-144356957 on Build GRCh38
rs201162690 Chr.8: 144357083-144357083 on Build GRCh38
rs904490383 Chr.8: 144356987-144356987 on Build GRCh38
rs782820063 Chr.8: 144356957-144356957 on Build GRCh38
rs782628611 Chr.8: 144357006-144357006 on Build GRCh38
rs781956596 Chr.8: 144357040-144357040 on Build GRCh38
rs782334349 Chr.8: 144357118-144357118 on Build GRCh38
rs373543815 Chr.8: 144356935-144356935 on Build GRCh38
rs116687799 Chr.8: 144357364-144357364 on Build GRCh38
rs371148890 Chr.8: 144357146-144357146 on Build GRCh38
rs782672318 Chr.8: 144357165-144357165 on Build GRCh38
rs904542638 Chr.8: 144357183-144357183 on Build GRCh38
rs111653799 Chr.8: 144357327-144357327 on Build GRCh38
rs782076645 Chr.8: 144357067-144357067 on Build GRCh38
rs994566678 Chr.8: 144357333-144357333 on Build GRCh38
rs782596489 Chr.8: 144357170-144357170 on Build GRCh38
rs564523873 Chr.8: 144356949-144356949 on Build GRCh38
rs143646016 Chr.8: 144357015-144357015 on Build GRCh38

View Full Product Details