Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144357849-144358340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6661982 c.361G>A p.V121M Chr.8: 144358087-144358087 on Build GRCh38
6393877 c.286G>A p.A96T Chr.8: 144358162-144358162 on Build GRCh38
6193835 c.305C>A p.T102K Chr.8: 144358143-144358143 on Build GRCh38
6361817 c.251G>T p.G84V Chr.8: 144358197-144358197 on Build GRCh38
3785909 c.109T>G p.Y37D Chr.8: 144358339-144358339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535349860 Chr.8: 144357950-144357950 on Build GRCh38
rs782239102 Chr.8: 144358053-144358053 on Build GRCh38
rs782336300 Chr.8: 144358163-144358163 on Build GRCh38
rs782607738 Chr.8: 144358052-144358052 on Build GRCh38
rs782742094 Chr.8: 144358116-144358116 on Build GRCh38
rs372626675 Chr.8: 144357972-144357972 on Build GRCh38
rs533637751 Chr.8: 144357896-144357896 on Build GRCh38
rs782805336 Chr.8: 144358297-144358297 on Build GRCh38
rs930902831 Chr.8: 144357998-144357998 on Build GRCh38
rs35266279 Chr.8: 144358260-144358260 on Build GRCh38
rs782743551 Chr.8: 144358282-144358282 on Build GRCh38
rs375550621 Chr.8: 144358140-144358140 on Build GRCh38
rs781782800 Chr.8: 144358314-144358314 on Build GRCh38
rs770883571 Chr.8: 144358119-144358119 on Build GRCh38
rs34763875 Chr.8: 144358292-144358292 on Build GRCh38
rs781873755 Chr.8: 144358001-144358001 on Build GRCh38
rs782180781 Chr.8: 144358000-144358001 on Build GRCh38
rs547331299 Chr.8: 144358042-144358042 on Build GRCh38
rs112549558 Chr.8: 144358179-144358179 on Build GRCh38
rs782274379 Chr.8: 144358151-144358151 on Build GRCh38
rs200282665 Chr.8: 144358290-144358290 on Build GRCh38
rs564308034 Chr.8: 144358171-144358171 on Build GRCh38
rs782526479 Chr.8: 144358291-144358291 on Build GRCh38
rs781903714 Chr.8: 144358135-144358135 on Build GRCh38
rs956722175 Chr.8: 144357899-144357899 on Build GRCh38
rs781826346 Chr.8: 144358142-144358142 on Build GRCh38
rs782132446 Chr.8: 144358125-144358125 on Build GRCh38
rs372596407 Chr.8: 144358059-144358059 on Build GRCh38
rs113885618 Chr.8: 144358155-144358155 on Build GRCh38
rs528243989 Chr.8: 144358204-144358204 on Build GRCh38
rs574776872 Chr.8: 144358038-144358038 on Build GRCh38
rs782597001 Chr.8: 144358320-144358320 on Build GRCh38
rs563338259 Chr.8: 144358149-144358149 on Build GRCh38
rs535421721 Chr.8: 144358040-144358041 on Build GRCh38
rs782187802 Chr.8: 144358159-144358159 on Build GRCh38
rs782643057 Chr.8: 144358150-144358150 on Build GRCh38
rs921896561 Chr.8: 144357985-144357985 on Build GRCh38
rs782381747 Chr.8: 144358054-144358054 on Build GRCh38
rs553219080 Chr.8: 144357964-144357964 on Build GRCh38
rs1049746879 Chr.8: 144358002-144358002 on Build GRCh38
rs782152530 Chr.8: 144358086-144358086 on Build GRCh38
rs782011787 Chr.8: 144358062-144358062 on Build GRCh38
rs200381094 Chr.8: 144358158-144358158 on Build GRCh38
rs782546539 Chr.8: 144358015-144358015 on Build GRCh38
rs782240396 Chr.8: 144358169-144358169 on Build GRCh38
rs782730397 Chr.8: 144358141-144358141 on Build GRCh38
rs368851769 Chr.8: 144357960-144357960 on Build GRCh38
rs781930050 Chr.8: 144358108-144358108 on Build GRCh38
rs782168336 Chr.8: 144358178-144358178 on Build GRCh38
rs782149168 Chr.8: 144358166-144358167 on Build GRCh38
rs782500969 Chr.8: 144358143-144358143 on Build GRCh38
rs781967081 Chr.8: 144358164-144358164 on Build GRCh38
rs988936837 Chr.8: 144357940-144357940 on Build GRCh38
rs782364876 Chr.8: 144358138-144358139 on Build GRCh38
rs782179596 Chr.8: 144358022-144358022 on Build GRCh38
rs374602988 Chr.8: 144357891-144357891 on Build GRCh38
rs940836285 Chr.8: 144358073-144358073 on Build GRCh38
rs967961452 Chr.8: 144357890-144357890 on Build GRCh38
rs541849271 Chr.8: 144358091-144358091 on Build GRCh38
rs782087192 Chr.8: 144358261-144358261 on Build GRCh38
rs782448687 Chr.8: 144358315-144358315 on Build GRCh38
rs116363663 Chr.8: 144357888-144357888 on Build GRCh38
rs545935518 Chr.8: 144358170-144358170 on Build GRCh38
rs369400816 Chr.8: 144358120-144358120 on Build GRCh38
rs782795427 Chr.8: 144358127-144358127 on Build GRCh38
rs782078974 Chr.8: 144358111-144358111 on Build GRCh38
rs2280840 Chr.8: 144357897-144357897 on Build GRCh38
rs782375884 Chr.8: 144358151-144358152 on Build GRCh38
rs782703374 Chr.8: 144358186-144358186 on Build GRCh38

View Full Product Details