Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144358364-144358858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5 NM_001253815.1 NM_001253816.1 NM_024531.4 NR_045600.1 XM_006716658.2 XM_006716659.2 XM_006716660.2 XM_017013819.1 XM_017013820.1 XM_017013821.1 XM_017013822.1

Gene Symbol
SLC52A2
Entrez Gene ID
79581
Gene Name
solute carrier family 52 member 2
Gene Aliases
BVVLS2, D15Ertd747e, GPCR41, GPR172A, PAR1, RFT3, RFVT2, hRFT3
UniGene
Hs.6459
Gene Chromosome Location
Chr.8: 144358547-144361286 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5 NM_001253815.1 NM_001253816.1 NM_024531.4 NR_045600.1 XM_006716658.2 XM_006716659.2 XM_006716660.2 XM_017013819.1 XM_017013820.1 XM_017013821.1 XM_017013822.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252245 c.1A>G p.M1V Chr.8: 144358447-144358447 on Build GRCh38
5626328 c.55C>T p.P19S Chr.8: 144358393-144358393 on Build GRCh38
4646205 c.9C>T p.A3A Chr.8: 144358439-144358439 on Build GRCh38
6567658 c.17C>T p.S6F Chr.8: 144358431-144358431 on Build GRCh38
5976744 c.84C>T p.D28D Chr.8: 144358364-144358364 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs952021230 Chr.8: 144358479-144358479 on Build GRCh38
rs551163570 Chr.8: 144358519-144358519 on Build GRCh38
rs113552670 Chr.8: 144358428-144358428 on Build GRCh38
rs540079494 Chr.8: 144358715-144358715 on Build GRCh38
rs947799880 Chr.8: 144358515-144358515 on Build GRCh38
rs782280291 Chr.8: 144358439-144358439 on Build GRCh38
rs782527871 Chr.8: 144358407-144358407 on Build GRCh38
rs782602801 Chr.8: 144358427-144358427 on Build GRCh38
rs936392675 Chr.8: 144358645-144358645 on Build GRCh38
rs782690748 Chr.8: 144358408-144358408 on Build GRCh38
rs887858910 Chr.8: 144358738-144358738 on Build GRCh38
rs782079584 Chr.8: 144358451-144358451 on Build GRCh38
rs1052399633 Chr.8: 144358537-144358537 on Build GRCh38
rs918057011 Chr.8: 144358508-144358508 on Build GRCh38
rs899697444 Chr.8: 144358708-144358708 on Build GRCh38
rs962047959 Chr.8: 144358833-144358833 on Build GRCh38
rs782053106 Chr.8: 144358477-144358477 on Build GRCh38
rs573270833 Chr.8: 144358662-144358662 on Build GRCh38
rs369644400 Chr.8: 144358727-144358727 on Build GRCh38
rs782750055 Chr.8: 144358398-144358398 on Build GRCh38
rs568462733 Chr.8: 144358611-144358611 on Build GRCh38
rs561859257 Chr.8: 144358774-144358774 on Build GRCh38
rs550638473 Chr.8: 144358434-144358434 on Build GRCh38
rs115940535 Chr.8: 144358634-144358634 on Build GRCh38
rs1029067767 Chr.8: 144358725-144358725 on Build GRCh38
rs1019069468 Chr.8: 144358465-144358465 on Build GRCh38
rs1015353494 Chr.8: 144358778-144358778 on Build GRCh38
rs13265792 Chr.8: 144358400-144358400 on Build GRCh38
rs183286864 Chr.8: 144358462-144358462 on Build GRCh38
rs1052535016 Chr.8: 144358649-144358649 on Build GRCh38
rs925018292 Chr.8: 144358642-144358642 on Build GRCh38
rs1015214204 Chr.8: 144358615-144358615 on Build GRCh38
rs962609844 Chr.8: 144358621-144358621 on Build GRCh38
rs782130850 Chr.8: 144358459-144358459 on Build GRCh38
rs903545945 Chr.8: 144358522-144358522 on Build GRCh38
rs575502265 Chr.8: 144358633-144358633 on Build GRCh38
rs896214929 Chr.8: 144358406-144358406 on Build GRCh38
rs899646531 Chr.8: 144358579-144358579 on Build GRCh38
rs782300700 Chr.8: 144358446-144358446 on Build GRCh38
rs903493584 Chr.8: 144358423-144358423 on Build GRCh38
rs974762470 Chr.8: 144358471-144358471 on Build GRCh38
rs782648310 Chr.8: 144358435-144358435 on Build GRCh38
rs782237825 Chr.8: 144358429-144358429 on Build GRCh38
rs144713518 Chr.8: 144358588-144358588 on Build GRCh38
rs781790906 Chr.8: 144358411-144358411 on Build GRCh38
rs1029013775 Chr.8: 144358598-144358598 on Build GRCh38
rs1057031728 Chr.8: 144358506-144358506 on Build GRCh38
rs782459581 Chr.8: 144358413-144358413 on Build GRCh38
rs113099131 Chr.8: 144358585-144358585 on Build GRCh38
rs1040914976 Chr.8: 144358699-144358699 on Build GRCh38
rs11546260 Chr.8: 144358747-144358747 on Build GRCh38
rs985084225 Chr.8: 144358483-144358483 on Build GRCh38
rs985188227 Chr.8: 144358610-144358610 on Build GRCh38
rs187464121 Chr.8: 144358622-144358622 on Build GRCh38
rs781900084 Chr.8: 144358467-144358467 on Build GRCh38
rs571040659 Chr.8: 144358559-144358559 on Build GRCh38
rs575557443 Chr.8: 144358639-144358639 on Build GRCh38
rs782355251 Chr.8: 144358454-144358454 on Build GRCh38
rs534941041 Chr.8: 144358560-144358560 on Build GRCh38
rs781980167 Chr.8: 144358456-144358456 on Build GRCh38
rs535737457 Chr.8: 144358616-144358616 on Build GRCh38
rs779099074 Chr.8: 144358496-144358496 on Build GRCh38
rs943964555 Chr.8: 144358689-144358689 on Build GRCh38
rs952075170 Chr.8: 144358606-144358606 on Build GRCh38
rs936310719 Chr.8: 144358534-144358534 on Build GRCh38
rs782707898 Chr.8: 144358488-144358488 on Build GRCh38
rs940902751 Chr.8: 144358505-144358505 on Build GRCh38
rs892098486 Chr.8: 144358682-144358682 on Build GRCh38
rs772404439 Chr.8: 144358575-144358575 on Build GRCh38
rs568833806 Chr.8: 144358450-144358450 on Build GRCh38

View Full Product Details