Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144526457-144526945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf82
Entrez Gene ID
414919
Gene Name
chromosome 8 open reading frame 82
Gene Aliases
-
UniGene
Hs.105685
Gene Chromosome Location
Chr.8: 144526219-144529074 on Build GRCh38
Transcripts
NM_001001795.1 NM_001024678.3

Gene Symbol
LRRC24
Entrez Gene ID
441381
Gene Name
leucine rich repeat containing 24
Gene Aliases
LRRC14OS
UniGene
Hs.740859
Gene Chromosome Location
Chr.8: 144522377-144527032 on Build GRCh38
Transcripts
NM_001001795.1 NM_001024678.3

SNP Detail

SNP ID Chromosome Location
rs945830105 Chr.8: 144526638-144526639 on Build GRCh38
rs116793122 Chr.8: 144526903-144526903 on Build GRCh38
rs189120848 Chr.8: 144526499-144526499 on Build GRCh38
rs1004876247 Chr.8: 144526892-144526892 on Build GRCh38
rs960031292 Chr.8: 144526799-144526799 on Build GRCh38
rs1015885265 Chr.8: 144526895-144526895 on Build GRCh38
rs533477089 Chr.8: 144526804-144526804 on Build GRCh38
rs560648803 Chr.8: 144526745-144526745 on Build GRCh38
rs552066538 Chr.8: 144526814-144526814 on Build GRCh38
rs940892447 Chr.8: 144526880-144526880 on Build GRCh38
rs536932755 Chr.8: 144526752-144526752 on Build GRCh38
rs913062328 Chr.8: 144526636-144526636 on Build GRCh38
rs60264048 Chr.8: 144526771-144526771 on Build GRCh38
rs987214260 Chr.8: 144526524-144526524 on Build GRCh38
rs777505239 Chr.8: 144526641-144526642 on Build GRCh38
rs760425287 Chr.8: 144526660-144526660 on Build GRCh38
rs753444713 Chr.8: 144526833-144526833 on Build GRCh38
rs1046800191 Chr.8: 144526875-144526875 on Build GRCh38
rs942373503 Chr.8: 144526652-144526652 on Build GRCh38
rs879675486 Chr.8: 144526885-144526885 on Build GRCh38
rs980099111 Chr.8: 144526545-144526546 on Build GRCh38
rs890367263 Chr.8: 144526834-144526834 on Build GRCh38
rs894313660 Chr.8: 144526659-144526659 on Build GRCh38
rs925836151 Chr.8: 144526645-144526645 on Build GRCh38
rs1019807418 Chr.8: 144526525-144526525 on Build GRCh38
rs1027136261 Chr.8: 144526777-144526777 on Build GRCh38
rs763622754 Chr.8: 144526807-144526807 on Build GRCh38
rs560504151 Chr.8: 144526831-144526831 on Build GRCh38
rs952878051 Chr.8: 144526888-144526888 on Build GRCh38
rs931568793 Chr.8: 144526580-144526580 on Build GRCh38
rs1034879512 Chr.8: 144526561-144526561 on Build GRCh38
rs556866137 Chr.8: 144526850-144526850 on Build GRCh38
rs904465456 Chr.8: 144526722-144526722 on Build GRCh38
rs983028240 Chr.8: 144526787-144526787 on Build GRCh38
rs908518395 Chr.8: 144526599-144526599 on Build GRCh38
rs537640524 Chr.8: 144526494-144526494 on Build GRCh38
rs953097875 Chr.8: 144526784-144526784 on Build GRCh38
rs908546269 Chr.8: 144526791-144526791 on Build GRCh38
rs964170311 Chr.8: 144526538-144526538 on Build GRCh38
rs983213705 Chr.8: 144526583-144526583 on Build GRCh38
rs890098438 Chr.8: 144526747-144526747 on Build GRCh38
rs1034082378 Chr.8: 144526755-144526755 on Build GRCh38
rs1039357970 Chr.8: 144526654-144526654 on Build GRCh38
rs1008603087 Chr.8: 144526842-144526842 on Build GRCh38
rs566911144 Chr.8: 144526559-144526559 on Build GRCh38
rs900941845 Chr.8: 144526667-144526667 on Build GRCh38
rs964450304 Chr.8: 144526770-144526770 on Build GRCh38
rs933750917 Chr.8: 144526682-144526682 on Build GRCh38
rs992784258 Chr.8: 144526801-144526801 on Build GRCh38
rs1042860563 Chr.8: 144526702-144526702 on Build GRCh38
rs992700963 Chr.8: 144526603-144526603 on Build GRCh38
rs922282357 Chr.8: 144526869-144526869 on Build GRCh38
rs1017406213 Chr.8: 144526753-144526753 on Build GRCh38
rs967220993 Chr.8: 144526825-144526825 on Build GRCh38
rs571693851 Chr.8: 144526606-144526606 on Build GRCh38
rs975521879 Chr.8: 144526857-144526857 on Build GRCh38
rs370390447 Chr.8: 144526523-144526523 on Build GRCh38
rs920208898 Chr.8: 144526557-144526557 on Build GRCh38
rs886728495 Chr.8: 144526509-144526509 on Build GRCh38
rs900276971 Chr.8: 144526853-144526853 on Build GRCh38
rs116905643 Chr.8: 144526746-144526746 on Build GRCh38
rs539334962 Chr.8: 144526712-144526712 on Build GRCh38
rs1016485717 Chr.8: 144526760-144526760 on Build GRCh38
rs994567396 Chr.8: 144526874-144526874 on Build GRCh38
rs181436434 Chr.8: 144526505-144526505 on Build GRCh38
rs549545574 Chr.8: 144526891-144526891 on Build GRCh38
rs553368528 Chr.8: 144526552-144526552 on Build GRCh38
rs915892771 Chr.8: 144526920-144526920 on Build GRCh38
rs886615592 Chr.8: 144526748-144526748 on Build GRCh38
rs934585470 Chr.8: 144526832-144526832 on Build GRCh38
rs542344513 Chr.8: 144526644-144526644 on Build GRCh38
rs567644435 Chr.8: 144526897-144526897 on Build GRCh38
rs1027582319 Chr.8: 144526878-144526878 on Build GRCh38
rs963728827 Chr.8: 144526536-144526536 on Build GRCh38

View Full Product Details