Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144527093-144527615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf82
Entrez Gene ID
414919
Gene Name
chromosome 8 open reading frame 82
Gene Aliases
-
UniGene
Hs.105685
Gene Chromosome Location
Chr.8: 144526219-144529074 on Build GRCh38
Transcripts
NM_001001795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272388 c.453C>A p.N151K Chr.8: 144527540-144527540 on Build GRCh38
6418335 c.421G>C p.E141Q Chr.8: 144527572-144527572 on Build GRCh38
5472548 c.483T>G p.R161R Chr.8: 144527510-144527510 on Build GRCh38
1199049 c.445G>A p.A149T Chr.8: 144527548-144527548 on Build GRCh38
5773149 c.430C>T p.R144C Chr.8: 144527563-144527563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562954258 Chr.8: 144527241-144527241 on Build GRCh38
rs543474208 Chr.8: 144527235-144527235 on Build GRCh38
rs959365000 Chr.8: 144527369-144527369 on Build GRCh38
rs962373325 Chr.8: 144527336-144527336 on Build GRCh38
rs1051096683 Chr.8: 144527497-144527497 on Build GRCh38
rs903190174 Chr.8: 144527281-144527281 on Build GRCh38
rs554511706 Chr.8: 144527208-144527208 on Build GRCh38
rs899884791 Chr.8: 144527296-144527296 on Build GRCh38
rs111869320 Chr.8: 144527588-144527588 on Build GRCh38
rs936026999 Chr.8: 144527429-144527429 on Build GRCh38
rs1033760923 Chr.8: 144527528-144527528 on Build GRCh38
rs992052587 Chr.8: 144527231-144527231 on Build GRCh38
rs1015426448 Chr.8: 144527328-144527328 on Build GRCh38
rs374358931 Chr.8: 144527589-144527589 on Build GRCh38
rs1004094189 Chr.8: 144527312-144527312 on Build GRCh38
rs950161250 Chr.8: 144527413-144527413 on Build GRCh38
rs917527456 Chr.8: 144527233-144527233 on Build GRCh38
rs982454758 Chr.8: 144527211-144527211 on Build GRCh38
rs962319060 Chr.8: 144527213-144527213 on Build GRCh38
rs186071534 Chr.8: 144527143-144527143 on Build GRCh38
rs903307527 Chr.8: 144527133-144527133 on Build GRCh38
rs753686174 Chr.8: 144527581-144527581 on Build GRCh38
rs1041774411 Chr.8: 144527257-144527257 on Build GRCh38
rs997059744 Chr.8: 144527204-144527204 on Build GRCh38
rs1054561755 Chr.8: 144527295-144527295 on Build GRCh38
rs924831675 Chr.8: 144527420-144527420 on Build GRCh38
rs991710605 Chr.8: 144527373-144527373 on Build GRCh38
rs878906328 Chr.8: 144527523-144527523 on Build GRCh38
rs917414410 Chr.8: 144527409-144527409 on Build GRCh38
rs865995097 Chr.8: 144527212-144527212 on Build GRCh38
rs977670700 Chr.8: 144527418-144527418 on Build GRCh38
rs959186378 Chr.8: 144527540-144527540 on Build GRCh38
rs578110481 Chr.8: 144527345-144527345 on Build GRCh38
rs937538849 Chr.8: 144527227-144527227 on Build GRCh38
rs571757742 Chr.8: 144527119-144527119 on Build GRCh38
rs1033462877 Chr.8: 144527352-144527352 on Build GRCh38
rs767068387 Chr.8: 144527508-144527508 on Build GRCh38
rs545746232 Chr.8: 144527395-144527395 on Build GRCh38
rs1014293288 Chr.8: 144527118-144527118 on Build GRCh38
rs1015480192 Chr.8: 144527522-144527522 on Build GRCh38
rs1054846713 Chr.8: 144527460-144527460 on Build GRCh38
rs935958869 Chr.8: 144527293-144527293 on Build GRCh38
rs891078885 Chr.8: 144527301-144527301 on Build GRCh38
rs1036588597 Chr.8: 144527136-144527158 on Build GRCh38
rs560764443 Chr.8: 144527470-144527470 on Build GRCh38
rs761338156 Chr.8: 144527498-144527498 on Build GRCh38
rs779683050 Chr.8: 144527579-144527579 on Build GRCh38
rs751107392 Chr.8: 144527482-144527482 on Build GRCh38
rs963913101 Chr.8: 144527167-144527167 on Build GRCh38
rs1029610771 Chr.8: 144527206-144527206 on Build GRCh38
rs113225732 Chr.8: 144527424-144527424 on Build GRCh38
rs11557085 Chr.8: 144527516-144527516 on Build GRCh38
rs926342611 Chr.8: 144527226-144527226 on Build GRCh38
rs576057161 Chr.8: 144527222-144527222 on Build GRCh38
rs1029909163 Chr.8: 144527300-144527300 on Build GRCh38
rs932721829 Chr.8: 144527478-144527478 on Build GRCh38
rs890960902 Chr.8: 144527503-144527503 on Build GRCh38
rs549408186 Chr.8: 144527582-144527582 on Build GRCh38
rs755822593 Chr.8: 144527564-144527564 on Build GRCh38

View Full Product Details