Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173824445-173824954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPL
Entrez Gene ID
91687
Gene Name
centromere protein L
Gene Aliases
C1orf155, CENP-L, dJ383J4.3
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173799550-173824639 on Build GRCh38
Transcripts
NM_001127181.2 NM_033319.3 NM_018122.4 XM_006711427.3 XM_011509711.2

Gene Symbol
DARS2
Entrez Gene ID
55157
Gene Name
aspartyl-tRNA synthetase 2, mitochondrial
Gene Aliases
ASPRS, LBSL, MT-ASPRS
UniGene
Hs.647707
Gene Chromosome Location
Chr.1: 173824659-173858544 on Build GRCh38
Transcripts
NM_001127181.2 NM_033319.3 NM_018122.4 XM_006711427.3 XM_011509711.2

SNP Detail

SNP ID Chromosome Location
rs1010199696 Chr.1: 173824859-173824859 on Build GRCh38
rs985191410 Chr.1: 173824799-173824799 on Build GRCh38
rs1054965729 Chr.1: 173824854-173824854 on Build GRCh38
rs917752868 Chr.1: 173824596-173824596 on Build GRCh38
rs1007816262 Chr.1: 173824700-173824700 on Build GRCh38
rs1033514410 Chr.1: 173824666-173824666 on Build GRCh38
rs950859424 Chr.1: 173824752-173824752 on Build GRCh38
rs377421881 Chr.1: 173824471-173824471 on Build GRCh38
rs544280201 Chr.1: 173824642-173824642 on Build GRCh38
rs1009461490 Chr.1: 173824588-173824588 on Build GRCh38
rs575882266 Chr.1: 173824876-173824876 on Build GRCh38
rs889874796 Chr.1: 173824539-173824539 on Build GRCh38
rs1049808384 Chr.1: 173824517-173824517 on Build GRCh38
rs575202140 Chr.1: 173824578-173824578 on Build GRCh38
rs1020839481 Chr.1: 173824628-173824628 on Build GRCh38
rs867647313 Chr.1: 173824711-173824711 on Build GRCh38
rs1042547236 Chr.1: 173824656-173824656 on Build GRCh38
rs191506182 Chr.1: 173824792-173824792 on Build GRCh38
rs976174210 Chr.1: 173824516-173824516 on Build GRCh38
rs1000722333 Chr.1: 173824563-173824563 on Build GRCh38
rs1020786960 Chr.1: 173824860-173824860 on Build GRCh38
rs997579129 Chr.1: 173824743-173824743 on Build GRCh38
rs752501776 Chr.1: 173824583-173824583 on Build GRCh38
rs972952504 Chr.1: 173824773-173824773 on Build GRCh38
rs902281167 Chr.1: 173824670-173824670 on Build GRCh38
rs886045585 Chr.1: 173824708-173824708 on Build GRCh38
rs956258212 Chr.1: 173824585-173824585 on Build GRCh38
rs984832770 Chr.1: 173824537-173824537 on Build GRCh38
rs1049913930 Chr.1: 173824836-173824836 on Build GRCh38
rs939700965 Chr.1: 173824748-173824748 on Build GRCh38
rs777622082 Chr.1: 173824898-173824898 on Build GRCh38
rs117902554 Chr.1: 173824540-173824540 on Build GRCh38
rs533128814 Chr.1: 173824697-173824697 on Build GRCh38
rs936318556 Chr.1: 173824687-173824687 on Build GRCh38
rs147279823 Chr.1: 173824931-173824931 on Build GRCh38
rs981910873 Chr.1: 173824662-173824662 on Build GRCh38
rs936080765 Chr.1: 173824852-173824852 on Build GRCh38
rs959244556 Chr.1: 173824904-173824904 on Build GRCh38
rs950795823 Chr.1: 173824528-173824528 on Build GRCh38
rs909112102 Chr.1: 173824543-173824543 on Build GRCh38
rs1026799059 Chr.1: 173824747-173824747 on Build GRCh38
rs937966562 Chr.1: 173824552-173824552 on Build GRCh38
rs934109253 Chr.1: 173824803-173824803 on Build GRCh38
rs560324631 Chr.1: 173824843-173824843 on Build GRCh38
rs564192663 Chr.1: 173824689-173824689 on Build GRCh38
rs949109467 Chr.1: 173824649-173824649 on Build GRCh38
rs1017751784 Chr.1: 173824701-173824701 on Build GRCh38
rs970498204 Chr.1: 173824637-173824637 on Build GRCh38
rs548385789 Chr.1: 173824534-173824534 on Build GRCh38
rs1026286241 Chr.1: 173824526-173824526 on Build GRCh38
rs879215879 Chr.1: 173824685-173824685 on Build GRCh38
rs778614831 Chr.1: 173824495-173824495 on Build GRCh38
rs922858050 Chr.1: 173824790-173824790 on Build GRCh38
rs529170828 Chr.1: 173824886-173824886 on Build GRCh38
rs758182518 Chr.1: 173824698-173824698 on Build GRCh38
rs1016308861 Chr.1: 173824901-173824901 on Build GRCh38
rs879324961 Chr.1: 173824857-173824859 on Build GRCh38

View Full Product Details