Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175192491-175192994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0040
Entrez Gene ID
9674
Gene Name
KIAA0040
Gene Aliases
-
UniGene
Hs.518138
Gene Chromosome Location
Chr.1: 175156986-175193300 on Build GRCh38
Transcripts
NM_001162893.1 NM_001162894.1 NM_001162895.1 NM_001319230.1 NM_001319231.1 NM_014656.2 XM_017002909.1 XM_017002910.1 XM_017002911.1

SNP Detail

SNP ID Chromosome Location
rs928816804 Chr.1: 175192843-175192843 on Build GRCh38
rs907192304 Chr.1: 175192788-175192788 on Build GRCh38
rs527703069 Chr.1: 175192516-175192516 on Build GRCh38
rs145358210 Chr.1: 175192726-175192726 on Build GRCh38
rs918512140 Chr.1: 175192895-175192895 on Build GRCh38
rs201321560 Chr.1: 175192663-175192664 on Build GRCh38
rs541473295 Chr.1: 175192518-175192518 on Build GRCh38
rs979176316 Chr.1: 175192865-175192865 on Build GRCh38
rs986178959 Chr.1: 175192905-175192905 on Build GRCh38
rs961874105 Chr.1: 175192887-175192887 on Build GRCh38
rs530464633 Chr.1: 175192645-175192645 on Build GRCh38
rs1010505428 Chr.1: 175192539-175192539 on Build GRCh38
rs953499215 Chr.1: 175192873-175192873 on Build GRCh38
rs551202481 Chr.1: 175192742-175192742 on Build GRCh38
rs888197408 Chr.1: 175192749-175192749 on Build GRCh38
rs1055576071 Chr.1: 175192913-175192913 on Build GRCh38
rs550408736 Chr.1: 175192659-175192659 on Build GRCh38
rs975998516 Chr.1: 175192668-175192668 on Build GRCh38
rs61359813 Chr.1: 175192842-175192843 on Build GRCh38
rs566911634 Chr.1: 175192909-175192909 on Build GRCh38
rs891967684 Chr.1: 175192941-175192941 on Build GRCh38
rs984550038 Chr.1: 175192639-175192639 on Build GRCh38
rs976019330 Chr.1: 175192889-175192889 on Build GRCh38
rs1034505453 Chr.1: 175192779-175192779 on Build GRCh38
rs764527794 Chr.1: 175192703-175192703 on Build GRCh38
rs931922629 Chr.1: 175192899-175192900 on Build GRCh38
rs971827967 Chr.1: 175192574-175192574 on Build GRCh38
rs976527993 Chr.1: 175192894-175192894 on Build GRCh38
rs115368522 Chr.1: 175192938-175192938 on Build GRCh38
rs895544310 Chr.1: 175192916-175192916 on Build GRCh38
rs936989162 Chr.1: 175192910-175192910 on Build GRCh38
rs952104346 Chr.1: 175192807-175192807 on Build GRCh38
rs903436988 Chr.1: 175192940-175192940 on Build GRCh38
rs186239689 Chr.1: 175192565-175192565 on Build GRCh38
rs1001906685 Chr.1: 175192790-175192790 on Build GRCh38
rs865951281 Chr.1: 175192826-175192826 on Build GRCh38
rs1052789365 Chr.1: 175192858-175192858 on Build GRCh38
rs1000490125 Chr.1: 175192971-175192971 on Build GRCh38
rs111922882 Chr.1: 175192693-175192693 on Build GRCh38
rs1044992534 Chr.1: 175192836-175192836 on Build GRCh38
rs967591472 Chr.1: 175192864-175192864 on Build GRCh38
rs754362769 Chr.1: 175192721-175192721 on Build GRCh38
rs927402980 Chr.1: 175192600-175192600 on Build GRCh38
rs891998778 Chr.1: 175192731-175192731 on Build GRCh38
rs371658604 Chr.1: 175192874-175192874 on Build GRCh38
rs948923270 Chr.1: 175192831-175192831 on Build GRCh38
rs984851497 Chr.1: 175192847-175192847 on Build GRCh38
rs943862173 Chr.1: 175192696-175192696 on Build GRCh38
rs980207115 Chr.1: 175192578-175192578 on Build GRCh38
rs1025016496 Chr.1: 175192741-175192741 on Build GRCh38
rs936425034 Chr.1: 175192711-175192711 on Build GRCh38
rs911090111 Chr.1: 175192695-175192695 on Build GRCh38
rs943894709 Chr.1: 175192883-175192883 on Build GRCh38
rs533617046 Chr.1: 175192798-175192798 on Build GRCh38
rs946230818 Chr.1: 175192944-175192944 on Build GRCh38
rs1048025299 Chr.1: 175192739-175192739 on Build GRCh38
rs985698518 Chr.1: 175192613-175192613 on Build GRCh38
rs903563277 Chr.1: 175192704-175192704 on Build GRCh38
rs867878301 Chr.1: 175192846-175192846 on Build GRCh38
rs1048787703 Chr.1: 175192904-175192904 on Build GRCh38
rs10912903 Chr.1: 175192794-175192794 on Build GRCh38
rs907729828 Chr.1: 175192641-175192641 on Build GRCh38
rs1020566617 Chr.1: 175192796-175192796 on Build GRCh38
rs12087240 Chr.1: 175192608-175192608 on Build GRCh38
rs778900471 Chr.1: 175192756-175192756 on Build GRCh38
rs1032973537 Chr.1: 175192870-175192870 on Build GRCh38
rs929871881 Chr.1: 175192896-175192896 on Build GRCh38
rs2272784 Chr.1: 175192713-175192713 on Build GRCh38
rs952138160 Chr.1: 175192603-175192604 on Build GRCh38
rs985060512 Chr.1: 175192821-175192821 on Build GRCh38
rs922961725 Chr.1: 175192675-175192675 on Build GRCh38
rs896014418 Chr.1: 175192786-175192786 on Build GRCh38
rs1014442365 Chr.1: 175192929-175192929 on Build GRCh38
rs929658230 Chr.1: 175192734-175192734 on Build GRCh38
rs911959816 Chr.1: 175192879-175192879 on Build GRCh38

View Full Product Details