Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175192770-175193264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0040
Entrez Gene ID
9674
Gene Name
KIAA0040
Gene Aliases
-
UniGene
Hs.518138
Gene Chromosome Location
Chr.1: 175156986-175193300 on Build GRCh38
Transcripts
NM_001162893.1 NM_001162894.1 NM_001162895.1 NM_001319230.1 NM_001319231.1 NM_014656.2 XM_017002909.1 XM_017002910.1 XM_017002911.1

SNP Detail

SNP ID Chromosome Location
rs928816804 Chr.1: 175192843-175192843 on Build GRCh38
rs1014999575 Chr.1: 175193042-175193042 on Build GRCh38
rs907192304 Chr.1: 175192788-175192788 on Build GRCh38
rs1019891206 Chr.1: 175193093-175193093 on Build GRCh38
rs576181792 Chr.1: 175192981-175192981 on Build GRCh38
rs370416362 Chr.1: 175193159-175193159 on Build GRCh38
rs918512140 Chr.1: 175192895-175192895 on Build GRCh38
rs986178959 Chr.1: 175192905-175192905 on Build GRCh38
rs979176316 Chr.1: 175192865-175192865 on Build GRCh38
rs961874105 Chr.1: 175192887-175192887 on Build GRCh38
rs949166610 Chr.1: 175193137-175193137 on Build GRCh38
rs953499215 Chr.1: 175192873-175192873 on Build GRCh38
rs542498779 Chr.1: 175193156-175193156 on Build GRCh38
rs1055576071 Chr.1: 175192913-175192913 on Build GRCh38
rs965195999 Chr.1: 175193097-175193097 on Build GRCh38
rs61359813 Chr.1: 175192842-175192843 on Build GRCh38
rs566911634 Chr.1: 175192909-175192909 on Build GRCh38
rs891967684 Chr.1: 175192941-175192941 on Build GRCh38
rs3753558 Chr.1: 175193171-175193171 on Build GRCh38
rs976019330 Chr.1: 175192889-175192889 on Build GRCh38
rs953430573 Chr.1: 175192976-175192976 on Build GRCh38
rs931922629 Chr.1: 175192899-175192900 on Build GRCh38
rs997615412 Chr.1: 175193106-175193106 on Build GRCh38
rs976527993 Chr.1: 175192894-175192894 on Build GRCh38
rs957730917 Chr.1: 175193123-175193123 on Build GRCh38
rs115368522 Chr.1: 175192938-175192938 on Build GRCh38
rs953389015 Chr.1: 175193070-175193070 on Build GRCh38
rs895544310 Chr.1: 175192916-175192916 on Build GRCh38
rs918459848 Chr.1: 175193022-175193022 on Build GRCh38
rs949741157 Chr.1: 175193107-175193107 on Build GRCh38
rs1042978867 Chr.1: 175193023-175193023 on Build GRCh38
rs1000754884 Chr.1: 175193145-175193145 on Build GRCh38
rs950699922 Chr.1: 175193124-175193124 on Build GRCh38
rs936989162 Chr.1: 175192910-175192910 on Build GRCh38
rs1019001555 Chr.1: 175192985-175192985 on Build GRCh38
rs952104346 Chr.1: 175192807-175192807 on Build GRCh38
rs1041484208 Chr.1: 175193109-175193109 on Build GRCh38
rs903436988 Chr.1: 175192940-175192940 on Build GRCh38
rs984083494 Chr.1: 175193039-175193039 on Build GRCh38
rs889665289 Chr.1: 175193036-175193036 on Build GRCh38
rs1007568422 Chr.1: 175193181-175193181 on Build GRCh38
rs866896840 Chr.1: 175193207-175193207 on Build GRCh38
rs1001906685 Chr.1: 175192790-175192790 on Build GRCh38
rs375987394 Chr.1: 175193221-175193221 on Build GRCh38
rs1011171603 Chr.1: 175193018-175193018 on Build GRCh38
rs865951281 Chr.1: 175192826-175192826 on Build GRCh38
rs1030184838 Chr.1: 175193065-175193065 on Build GRCh38
rs1000490125 Chr.1: 175192971-175192971 on Build GRCh38
rs1052789365 Chr.1: 175192858-175192858 on Build GRCh38
rs529790811 Chr.1: 175193125-175193125 on Build GRCh38
rs937040432 Chr.1: 175193087-175193087 on Build GRCh38
rs966067283 Chr.1: 175193003-175193003 on Build GRCh38
rs1026379791 Chr.1: 175193119-175193119 on Build GRCh38
rs967591472 Chr.1: 175192864-175192864 on Build GRCh38
rs1044992534 Chr.1: 175192836-175192836 on Build GRCh38
rs190560821 Chr.1: 175193099-175193099 on Build GRCh38
rs976557588 Chr.1: 175193011-175193011 on Build GRCh38
rs371658604 Chr.1: 175192874-175192874 on Build GRCh38
rs976283451 Chr.1: 175193064-175193064 on Build GRCh38
rs948923270 Chr.1: 175192831-175192831 on Build GRCh38
rs1006145474 Chr.1: 175193234-175193234 on Build GRCh38
rs984851497 Chr.1: 175192847-175192847 on Build GRCh38
rs3753559 Chr.1: 175193132-175193132 on Build GRCh38
rs1018862494 Chr.1: 175193193-175193193 on Build GRCh38
rs533617046 Chr.1: 175192798-175192798 on Build GRCh38
rs943894709 Chr.1: 175192883-175192883 on Build GRCh38
rs946230818 Chr.1: 175192944-175192944 on Build GRCh38
rs867878301 Chr.1: 175192846-175192846 on Build GRCh38
rs1048787703 Chr.1: 175192904-175192904 on Build GRCh38
rs10912903 Chr.1: 175192794-175192794 on Build GRCh38
rs987880251 Chr.1: 175193132-175193132 on Build GRCh38
rs1020566617 Chr.1: 175192796-175192796 on Build GRCh38
rs1054733741 Chr.1: 175193158-175193158 on Build GRCh38
rs951283541 Chr.1: 175193037-175193037 on Build GRCh38
rs1032973537 Chr.1: 175192870-175192870 on Build GRCh38
rs3753560 Chr.1: 175193060-175193060 on Build GRCh38
rs556378309 Chr.1: 175192974-175192974 on Build GRCh38
rs929871881 Chr.1: 175192896-175192896 on Build GRCh38
rs903383414 Chr.1: 175193129-175193129 on Build GRCh38
rs1056073113 Chr.1: 175193194-175193194 on Build GRCh38
rs867700202 Chr.1: 175193079-175193079 on Build GRCh38
rs887775291 Chr.1: 175193205-175193205 on Build GRCh38
rs985060512 Chr.1: 175192821-175192821 on Build GRCh38
rs1014442365 Chr.1: 175192929-175192929 on Build GRCh38
rs917008663 Chr.1: 175193100-175193100 on Build GRCh38
rs911959816 Chr.1: 175192879-175192879 on Build GRCh38

View Full Product Details