Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94374140-94374648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD14B
Entrez Gene ID
84641
Gene Name
major facilitator superfamily domain containing 14B
Gene Aliases
HIATL1
UniGene
Hs.555996
Gene Chromosome Location
Chr.9: 94374327-94460920 on Build GRCh38
Transcripts
NM_032558.2 XM_006717305.3 XM_011519114.2 XM_017015218.1 XM_017015220.1 XM_017015221.1

SNP Detail

SNP ID Chromosome Location
rs1049739889 Chr.9: 94374326-94374326 on Build GRCh38
rs567496060 Chr.9: 94374291-94374291 on Build GRCh38
rs956208851 Chr.9: 94374258-94374258 on Build GRCh38
rs980795557 Chr.9: 94374372-94374372 on Build GRCh38
rs1046233954 Chr.9: 94374600-94374600 on Build GRCh38
rs3118796 Chr.9: 94374602-94374602 on Build GRCh38
rs935940660 Chr.9: 94374582-94374582 on Build GRCh38
rs1036848313 Chr.9: 94374462-94374462 on Build GRCh38
rs1028803815 Chr.9: 94374210-94374210 on Build GRCh38
rs562596000 Chr.9: 94374606-94374606 on Build GRCh38
rs537981314 Chr.9: 94374296-94374296 on Build GRCh38
rs971659032 Chr.9: 94374366-94374366 on Build GRCh38
rs933918049 Chr.9: 94374491-94374491 on Build GRCh38
rs867344762 Chr.9: 94374219-94374219 on Build GRCh38
rs1016634219 Chr.9: 94374553-94374553 on Build GRCh38
rs926272671 Chr.9: 94374402-94374403 on Build GRCh38
rs757682372 Chr.9: 94374354-94374354 on Build GRCh38
rs907403314 Chr.9: 94374543-94374543 on Build GRCh38
rs891860648 Chr.9: 94374532-94374532 on Build GRCh38
rs1036954887 Chr.9: 94374190-94374190 on Build GRCh38
rs533842268 Chr.9: 94374554-94374554 on Build GRCh38
rs867917043 Chr.9: 94374270-94374270 on Build GRCh38
rs1010419980 Chr.9: 94374358-94374358 on Build GRCh38
rs1034609494 Chr.9: 94374545-94374545 on Build GRCh38
rs1000236029 Chr.9: 94374341-94374341 on Build GRCh38
rs998537162 Chr.9: 94374325-94374325 on Build GRCh38
rs573865851 Chr.9: 94374585-94374585 on Build GRCh38
rs925475185 Chr.9: 94374525-94374525 on Build GRCh38
rs1054443752 Chr.9: 94374527-94374527 on Build GRCh38
rs544388960 Chr.9: 94374595-94374595 on Build GRCh38
rs1049673776 Chr.9: 94374496-94374496 on Build GRCh38
rs555628760 Chr.9: 94374568-94374568 on Build GRCh38
rs1033063700 Chr.9: 94374349-94374349 on Build GRCh38
rs901108113 Chr.9: 94374323-94374323 on Build GRCh38
rs539026069 Chr.9: 94374415-94374415 on Build GRCh38
rs1006507636 Chr.9: 94374622-94374622 on Build GRCh38
rs1048210718 Chr.9: 94374290-94374290 on Build GRCh38
rs1037328018 Chr.9: 94374321-94374321 on Build GRCh38
rs937734888 Chr.9: 94374285-94374286 on Build GRCh38
rs139259210 Chr.9: 94374239-94374239 on Build GRCh38
rs190024467 Chr.9: 94374268-94374268 on Build GRCh38
rs1000267274 Chr.9: 94374225-94374225 on Build GRCh38
rs891823501 Chr.9: 94374594-94374594 on Build GRCh38
rs958999425 Chr.9: 94374552-94374552 on Build GRCh38
rs939826637 Chr.9: 94374453-94374453 on Build GRCh38
rs12375475 Chr.9: 94374451-94374451 on Build GRCh38
rs889820035 Chr.9: 94374330-94374330 on Build GRCh38
rs889749502 Chr.9: 94374511-94374511 on Build GRCh38
rs1004129235 Chr.9: 94374180-94374180 on Build GRCh38
rs888010757 Chr.9: 94374614-94374614 on Build GRCh38
rs1001753657 Chr.9: 94374544-94374544 on Build GRCh38
rs1024469670 Chr.9: 94374539-94374539 on Build GRCh38
rs981782319 Chr.9: 94374278-94374278 on Build GRCh38
rs926305234 Chr.9: 94374280-94374280 on Build GRCh38
rs970773531 Chr.9: 94374272-94374272 on Build GRCh38
rs370397234 Chr.9: 94374282-94374282 on Build GRCh38
rs936015678 Chr.9: 94374513-94374513 on Build GRCh38
rs142413498 Chr.9: 94374538-94374538 on Build GRCh38
rs373101299 Chr.9: 94374360-94374360 on Build GRCh38
rs553605377 Chr.9: 94374212-94374212 on Build GRCh38
rs549876392 Chr.9: 94374299-94374299 on Build GRCh38
rs571378322 Chr.9: 94374413-94374413 on Build GRCh38
rs901136205 Chr.9: 94374201-94374201 on Build GRCh38
rs985313042 Chr.9: 94374567-94374567 on Build GRCh38
rs781520697 Chr.9: 94374397-94374397 on Build GRCh38
rs922458661 Chr.9: 94374485-94374485 on Build GRCh38
rs959028906 Chr.9: 94374403-94374403 on Build GRCh38
rs961143741 Chr.9: 94374555-94374555 on Build GRCh38
rs933823775 Chr.9: 94374565-94374565 on Build GRCh38
rs998210761 Chr.9: 94374205-94374205 on Build GRCh38
rs972930639 Chr.9: 94374558-94374558 on Build GRCh38
rs911154076 Chr.9: 94374574-94374574 on Build GRCh38

View Full Product Details