Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94459790-94460308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD14B
Entrez Gene ID
84641
Gene Name
major facilitator superfamily domain containing 14B
Gene Aliases
HIATL1
UniGene
Hs.555996
Gene Chromosome Location
Chr.9: 94374327-94460920 on Build GRCh38
Transcripts
NM_032558.2 XM_006717305.3 XM_011519114.2 XM_017015217.1 XM_017015218.1 XM_017015219.1 XM_017015220.1 XM_017015221.1

SNP Detail

SNP ID Chromosome Location
rs566004419 Chr.9: 94460064-94460064 on Build GRCh38
rs937823355 Chr.9: 94460258-94460258 on Build GRCh38
rs904454531 Chr.9: 94460254-94460254 on Build GRCh38
rs1004183562 Chr.9: 94459883-94459885 on Build GRCh38
rs200229509 Chr.9: 94460132-94460132 on Build GRCh38
rs918782062 Chr.9: 94460196-94460196 on Build GRCh38
rs574340218 Chr.9: 94460190-94460190 on Build GRCh38
rs1029413136 Chr.9: 94459957-94459957 on Build GRCh38
rs551496741 Chr.9: 94460138-94460138 on Build GRCh38
rs987744274 Chr.9: 94460190-94460191 on Build GRCh38
rs1033226329 Chr.9: 94460153-94460153 on Build GRCh38
rs901233539 Chr.9: 94459936-94459936 on Build GRCh38
rs937920407 Chr.9: 94459830-94459830 on Build GRCh38
rs916662248 Chr.9: 94460221-94460221 on Build GRCh38
rs748708467 Chr.9: 94459835-94459837 on Build GRCh38
rs962908686 Chr.9: 94460097-94460097 on Build GRCh38
rs926506865 Chr.9: 94459818-94459818 on Build GRCh38
rs972887432 Chr.9: 94460118-94460118 on Build GRCh38
rs879052824 Chr.9: 94459887-94459887 on Build GRCh38
rs953880640 Chr.9: 94459986-94459986 on Build GRCh38
rs998367148 Chr.9: 94459930-94459930 on Build GRCh38
rs888365604 Chr.9: 94459849-94459849 on Build GRCh38
rs954270937 Chr.9: 94460128-94460128 on Build GRCh38
rs562549469 Chr.9: 94459908-94459908 on Build GRCh38
rs1000049978 Chr.9: 94459854-94459854 on Build GRCh38
rs1000142906 Chr.9: 94460277-94460277 on Build GRCh38
rs551241345 Chr.9: 94460109-94460109 on Build GRCh38
rs188102492 Chr.9: 94460089-94460089 on Build GRCh38
rs1056989706 Chr.9: 94460216-94460216 on Build GRCh38
rs1053006711 Chr.9: 94460282-94460282 on Build GRCh38
rs190794725 Chr.9: 94460198-94460198 on Build GRCh38
rs374527212 Chr.9: 94460225-94460225 on Build GRCh38
rs904405939 Chr.9: 94459840-94459840 on Build GRCh38
rs111867004 Chr.9: 94459904-94459904 on Build GRCh38
rs926448317 Chr.9: 94460253-94460256 on Build GRCh38
rs1031481301 Chr.9: 94459879-94459879 on Build GRCh38
rs1046197049 Chr.9: 94459813-94459813 on Build GRCh38
rs544783874 Chr.9: 94459911-94459914 on Build GRCh38
rs41281150 Chr.9: 94460265-94460265 on Build GRCh38
rs1043723539 Chr.9: 94460249-94460249 on Build GRCh38
rs554837404 Chr.9: 94460158-94460158 on Build GRCh38

View Full Product Details