Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98845898-98846394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT12
Entrez Gene ID
79695
Gene Name
polypeptide N-acetylgalactosaminyltransferase 12
Gene Aliases
CRCS1, GalNAc-T12
UniGene
Hs.47099
Gene Chromosome Location
Chr.9: 98807699-98850081 on Build GRCh38
Transcripts
NM_024642.4 XM_006717287.1 XM_011519018.2 XM_011519020.1 XM_017015133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4631338 c.1540G>A p.D514N Chr.9: 98846058-98846058 on Build GRCh38
1103160 c.1564G>A p.E522K Chr.9: 98846082-98846082 on Build GRCh38
3902516 c.1489C>T p.R497C Chr.9: 98846007-98846007 on Build GRCh38
4626281 c.1501C>A p.H501N Chr.9: 98846019-98846019 on Build GRCh38
4929476 c.1580A>G p.E527G Chr.9: 98846098-98846098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371029073 Chr.9: 98846289-98846289 on Build GRCh38
rs148647485 Chr.9: 98846092-98846092 on Build GRCh38
rs903491812 Chr.9: 98846191-98846191 on Build GRCh38
rs751724626 Chr.9: 98846019-98846019 on Build GRCh38
rs572700544 Chr.9: 98846117-98846117 on Build GRCh38
rs142096902 Chr.9: 98846101-98846101 on Build GRCh38
rs763159492 Chr.9: 98846105-98846105 on Build GRCh38
rs962673173 Chr.9: 98846356-98846356 on Build GRCh38
rs781780984 Chr.9: 98845962-98845962 on Build GRCh38
rs369089798 Chr.9: 98846033-98846033 on Build GRCh38
rs112993082 Chr.9: 98846016-98846016 on Build GRCh38
rs267606840 Chr.9: 98845990-98845990 on Build GRCh38
rs955510774 Chr.9: 98846011-98846011 on Build GRCh38
rs769128117 Chr.9: 98845955-98845955 on Build GRCh38
rs748688612 Chr.9: 98846045-98846045 on Build GRCh38
rs955640466 Chr.9: 98846270-98846270 on Build GRCh38
rs79574929 Chr.9: 98846127-98846127 on Build GRCh38
rs765327993 Chr.9: 98846009-98846009 on Build GRCh38
rs771570119 Chr.9: 98846293-98846293 on Build GRCh38
rs747340539 Chr.9: 98845953-98845953 on Build GRCh38
rs545000049 Chr.9: 98846346-98846346 on Build GRCh38
rs770554889 Chr.9: 98845985-98845985 on Build GRCh38
rs143860974 Chr.9: 98846081-98846081 on Build GRCh38
rs765816109 Chr.9: 98846024-98846025 on Build GRCh38
rs753813631 Chr.9: 98846163-98846163 on Build GRCh38
rs200112438 Chr.9: 98846008-98846008 on Build GRCh38
rs564986735 Chr.9: 98846358-98846358 on Build GRCh38
rs1000496980 Chr.9: 98846197-98846197 on Build GRCh38
rs35632007 Chr.9: 98846015-98846015 on Build GRCh38
rs770337578 Chr.9: 98846070-98846070 on Build GRCh38
rs536735094 Chr.9: 98845967-98845967 on Build GRCh38
rs878855094 Chr.9: 98846069-98846069 on Build GRCh38
rs760180024 Chr.9: 98845960-98845961 on Build GRCh38
rs137917376 Chr.9: 98845983-98845983 on Build GRCh38
rs930272397 Chr.9: 98846365-98846365 on Build GRCh38
rs574723319 Chr.9: 98846192-98846192 on Build GRCh38
rs552902108 Chr.9: 98845924-98845936 on Build GRCh38
rs200892718 Chr.9: 98846170-98846170 on Build GRCh38
rs774123474 Chr.9: 98846072-98846072 on Build GRCh38
rs756649181 Chr.9: 98846050-98846050 on Build GRCh38
rs201736296 Chr.9: 98846168-98846168 on Build GRCh38
rs575599270 Chr.9: 98846315-98846315 on Build GRCh38
rs892404582 Chr.9: 98846200-98846200 on Build GRCh38
rs777152516 Chr.9: 98846058-98846058 on Build GRCh38
rs780285965 Chr.9: 98845946-98845946 on Build GRCh38
rs950715714 Chr.9: 98846169-98846169 on Build GRCh38
rs985503031 Chr.9: 98846292-98846292 on Build GRCh38
rs753044857 Chr.9: 98846171-98846171 on Build GRCh38
rs778940636 Chr.9: 98845928-98845928 on Build GRCh38
rs1054734444 Chr.9: 98846318-98846318 on Build GRCh38
rs757397437 Chr.9: 98845927-98845927 on Build GRCh38
rs377188258 Chr.9: 98846018-98846018 on Build GRCh38
rs999070067 Chr.9: 98846043-98846043 on Build GRCh38
rs748778853 Chr.9: 98846064-98846064 on Build GRCh38
rs369843697 Chr.9: 98845933-98845933 on Build GRCh38
rs1030196202 Chr.9: 98846239-98846239 on Build GRCh38
rs372665132 Chr.9: 98846042-98846042 on Build GRCh38
rs553783609 Chr.9: 98846198-98846198 on Build GRCh38
rs542295626 Chr.9: 98846193-98846193 on Build GRCh38
rs774034115 Chr.9: 98845988-98845988 on Build GRCh38
rs113674846 Chr.9: 98846244-98846244 on Build GRCh38
rs757588259 Chr.9: 98846102-98846102 on Build GRCh38
rs759388241 Chr.9: 98845989-98845989 on Build GRCh38
rs373044668 Chr.9: 98845991-98845991 on Build GRCh38
rs1044710747 Chr.9: 98846184-98846184 on Build GRCh38
rs756706295 Chr.9: 98846172-98846172 on Build GRCh38
rs372728638 Chr.9: 98845932-98845932 on Build GRCh38
rs775453262 Chr.9: 98846007-98846007 on Build GRCh38
rs997743648 Chr.9: 98846214-98846214 on Build GRCh38
rs7858352 Chr.9: 98846361-98846361 on Build GRCh38
rs927896334 Chr.9: 98845931-98845931 on Build GRCh38
rs998507291 Chr.9: 98846316-98846316 on Build GRCh38
rs919219222 Chr.9: 98846135-98846135 on Build GRCh38
rs893362100 Chr.9: 98846326-98846326 on Build GRCh38
rs200303625 Chr.9: 98846082-98846082 on Build GRCh38
rs1050753632 Chr.9: 98846297-98846297 on Build GRCh38
rs772731021 Chr.9: 98845982-98845982 on Build GRCh38
rs751877685 Chr.9: 98846151-98846151 on Build GRCh38

View Full Product Details