Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101360832-101361369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAAT
Entrez Gene ID
570
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases
BACAT, BAT
UniGene
Hs.284712
Gene Chromosome Location
Chr.9: 101360417-101385005 on Build GRCh38
Transcripts
NM_001127610.1 NM_001701.3

SNP Detail

SNP ID Chromosome Location
rs377765138 Chr.9: 101361343-101361343 on Build GRCh38
rs1039571361 Chr.9: 101360985-101360985 on Build GRCh38
rs565046115 Chr.9: 101361155-101361155 on Build GRCh38
rs890340160 Chr.9: 101361130-101361130 on Build GRCh38
rs929652235 Chr.9: 101360919-101360924 on Build GRCh38
rs917545373 Chr.9: 101360980-101360980 on Build GRCh38
rs934070622 Chr.9: 101361017-101361017 on Build GRCh38
rs975289378 Chr.9: 101361273-101361273 on Build GRCh38
rs567316584 Chr.9: 101361193-101361193 on Build GRCh38
rs886063282 Chr.9: 101361292-101361292 on Build GRCh38
rs200796812 Chr.9: 101361305-101361305 on Build GRCh38
rs570341484 Chr.9: 101361264-101361264 on Build GRCh38
rs35538457 Chr.9: 101361252-101361253 on Build GRCh38
rs1004761067 Chr.9: 101361131-101361131 on Build GRCh38
rs1013893119 Chr.9: 101361258-101361258 on Build GRCh38
rs371076104 Chr.9: 101361035-101361035 on Build GRCh38
rs529692710 Chr.9: 101361200-101361200 on Build GRCh38
rs946286759 Chr.9: 101360982-101360982 on Build GRCh38
rs59311341 Chr.9: 101360970-101360970 on Build GRCh38
rs1041801133 Chr.9: 101361032-101361032 on Build GRCh38
rs977682350 Chr.9: 101361285-101361285 on Build GRCh38
rs926127084 Chr.9: 101361074-101361074 on Build GRCh38
rs111954077 Chr.9: 101361121-101361121 on Build GRCh38
rs572228256 Chr.9: 101360900-101360900 on Build GRCh38
rs760757882 Chr.9: 101361223-101361223 on Build GRCh38
rs542605798 Chr.9: 101360962-101360962 on Build GRCh38
rs958398145 Chr.9: 101361246-101361246 on Build GRCh38
rs973581382 Chr.9: 101360942-101360942 on Build GRCh38
rs574486400 Chr.9: 101360960-101360960 on Build GRCh38
rs924701513 Chr.9: 101361309-101361309 on Build GRCh38
rs547506053 Chr.9: 101361168-101361168 on Build GRCh38
rs1014719804 Chr.9: 101360872-101360872 on Build GRCh38
rs536464923 Chr.9: 101361298-101361298 on Build GRCh38
rs984069873 Chr.9: 101360947-101360947 on Build GRCh38
rs781579686 Chr.9: 101361224-101361224 on Build GRCh38
rs971588366 Chr.9: 101361185-101361185 on Build GRCh38
rs545205256 Chr.9: 101361087-101361087 on Build GRCh38
rs936071039 Chr.9: 101361082-101361082 on Build GRCh38
rs922465345 Chr.9: 101361280-101361280 on Build GRCh38
rs76498584 Chr.9: 101361338-101361338 on Build GRCh38
rs982303668 Chr.9: 101361191-101361191 on Build GRCh38
rs188189145 Chr.9: 101361289-101361289 on Build GRCh38
rs10120345 Chr.9: 101360953-101360953 on Build GRCh38
rs778507457 Chr.9: 101360993-101360993 on Build GRCh38
rs970894631 Chr.9: 101360879-101360879 on Build GRCh38
rs1051625702 Chr.9: 101361122-101361122 on Build GRCh38
rs903215104 Chr.9: 101361033-101361033 on Build GRCh38
rs531442189 Chr.9: 101361054-101361054 on Build GRCh38
rs918289127 Chr.9: 101360905-101360905 on Build GRCh38
rs751970647 Chr.9: 101361011-101361011 on Build GRCh38
rs1000137492 Chr.9: 101361034-101361034 on Build GRCh38
rs534833120 Chr.9: 101361345-101361345 on Build GRCh38
rs894748787 Chr.9: 101361083-101361083 on Build GRCh38
rs541077339 Chr.9: 101360949-101360949 on Build GRCh38
rs967599371 Chr.9: 101361266-101361266 on Build GRCh38
rs527386727 Chr.9: 101361164-101361164 on Build GRCh38
rs755320451 Chr.9: 101361112-101361112 on Build GRCh38
rs550241219 Chr.9: 101361210-101361210 on Build GRCh38
rs531601325 Chr.9: 101361253-101361253 on Build GRCh38

View Full Product Details