Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101361258-101361758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAAT
Entrez Gene ID
570
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases
BACAT, BAT
UniGene
Hs.284712
Gene Chromosome Location
Chr.9: 101360417-101385005 on Build GRCh38
Transcripts
NM_001127610.1 NM_001701.3

SNP Detail

SNP ID Chromosome Location
rs994129398 Chr.9: 101361652-101361652 on Build GRCh38
rs7043029 Chr.9: 101361480-101361480 on Build GRCh38
rs377765138 Chr.9: 101361343-101361343 on Build GRCh38
rs907470131 Chr.9: 101361626-101361626 on Build GRCh38
rs907393727 Chr.9: 101361391-101361392 on Build GRCh38
rs573463713 Chr.9: 101361357-101361357 on Build GRCh38
rs536464923 Chr.9: 101361298-101361298 on Build GRCh38
rs16936057 Chr.9: 101361533-101361533 on Build GRCh38
rs894175515 Chr.9: 101361492-101361492 on Build GRCh38
rs41301517 Chr.9: 101361473-101361473 on Build GRCh38
rs577168710 Chr.9: 101361548-101361548 on Build GRCh38
rs373585277 Chr.9: 101361532-101361532 on Build GRCh38
rs1033718581 Chr.9: 101361542-101361542 on Build GRCh38
rs1045974598 Chr.9: 101361391-101361391 on Build GRCh38
rs887907692 Chr.9: 101361685-101361685 on Build GRCh38
rs921446266 Chr.9: 101361632-101361632 on Build GRCh38
rs886063282 Chr.9: 101361292-101361292 on Build GRCh38
rs76498584 Chr.9: 101361338-101361338 on Build GRCh38
rs200796812 Chr.9: 101361305-101361305 on Build GRCh38
rs1047901656 Chr.9: 101361672-101361672 on Build GRCh38
rs986859859 Chr.9: 101361701-101361701 on Build GRCh38
rs554642026 Chr.9: 101361378-101361378 on Build GRCh38
rs576308259 Chr.9: 101361426-101361426 on Build GRCh38
rs188189145 Chr.9: 101361289-101361289 on Build GRCh38
rs955404917 Chr.9: 101361515-101361515 on Build GRCh38
rs1014029967 Chr.9: 101361512-101361512 on Build GRCh38
rs1016614502 Chr.9: 101361440-101361440 on Build GRCh38
rs977682350 Chr.9: 101361285-101361285 on Build GRCh38
rs181417867 Chr.9: 101361641-101361641 on Build GRCh38
rs74606977 Chr.9: 101361399-101361399 on Build GRCh38
rs1036268224 Chr.9: 101361417-101361417 on Build GRCh38
rs948710263 Chr.9: 101361581-101361581 on Build GRCh38
rs866621942 Chr.9: 101361491-101361491 on Build GRCh38
rs1054429085 Chr.9: 101361537-101361537 on Build GRCh38
rs534833120 Chr.9: 101361345-101361345 on Build GRCh38
rs977299473 Chr.9: 101361523-101361523 on Build GRCh38
rs551905862 Chr.9: 101361398-101361399 on Build GRCh38
rs975953562 Chr.9: 101361560-101361560 on Build GRCh38
rs1046427884 Chr.9: 101361604-101361604 on Build GRCh38
rs41316534 Chr.9: 101361387-101361387 on Build GRCh38
rs924701513 Chr.9: 101361309-101361309 on Build GRCh38
rs1023946264 Chr.9: 101361526-101361526 on Build GRCh38
rs543786788 Chr.9: 101361398-101361398 on Build GRCh38

View Full Product Details