Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108936219-108936721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM206A
Entrez Gene ID
54942
Gene Name
family with sequence similarity 206 member A
Gene Aliases
C9orf6, CG-8, Simiate
UniGene
Hs.29276
Gene Chromosome Location
Chr.9: 108934393-108950745 on Build GRCh38
Transcripts
NM_017832.3 XM_011518816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682870 c.243C>A p.V81V Chr.9: 108936419-108936419 on Build GRCh38
6481374 c.150G>T p.L50F Chr.9: 108936326-108936326 on Build GRCh38
3902873 c.261+2T>C p.? Chr.9: 108936439-108936439 on Build GRCh38
6536819 c.196A>G p.I66V Chr.9: 108936372-108936372 on Build GRCh38
4405835 c.163C>T p.P55S Chr.9: 108936339-108936339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763426640 Chr.9: 108936503-108936503 on Build GRCh38
rs780697700 Chr.9: 108936245-108936245 on Build GRCh38
rs563994567 Chr.9: 108936663-108936663 on Build GRCh38
rs979692563 Chr.9: 108936552-108936552 on Build GRCh38
rs184918618 Chr.9: 108936658-108936658 on Build GRCh38
rs747079742 Chr.9: 108936471-108936471 on Build GRCh38
rs751039447 Chr.9: 108936428-108936428 on Build GRCh38
rs780122982 Chr.9: 108936466-108936466 on Build GRCh38
rs1039102691 Chr.9: 108936475-108936475 on Build GRCh38
rs201770790 Chr.9: 108936281-108936281 on Build GRCh38
rs560717022 Chr.9: 108936579-108936579 on Build GRCh38
rs762359423 Chr.9: 108936418-108936418 on Build GRCh38
rs979388169 Chr.9: 108936268-108936268 on Build GRCh38
rs1024108048 Chr.9: 108936244-108936244 on Build GRCh38
rs150725760 Chr.9: 108936435-108936435 on Build GRCh38
rs929515690 Chr.9: 108936359-108936359 on Build GRCh38
rs1040891660 Chr.9: 108936282-108936282 on Build GRCh38
rs372499776 Chr.9: 108936441-108936441 on Build GRCh38
rs775728990 Chr.9: 108936371-108936371 on Build GRCh38
rs745918490 Chr.9: 108936352-108936352 on Build GRCh38
rs760934146 Chr.9: 108936260-108936260 on Build GRCh38
rs368424181 Chr.9: 108936297-108936297 on Build GRCh38
rs117239473 Chr.9: 108936337-108936337 on Build GRCh38
rs375430911 Chr.9: 108936325-108936325 on Build GRCh38
rs369983774 Chr.9: 108936419-108936419 on Build GRCh38
rs972417040 Chr.9: 108936302-108936302 on Build GRCh38
rs367971271 Chr.9: 108936284-108936284 on Build GRCh38
rs1053018334 Chr.9: 108936550-108936550 on Build GRCh38
rs370302809 Chr.9: 108936271-108936271 on Build GRCh38
rs761495479 Chr.9: 108936283-108936283 on Build GRCh38
rs1050266328 Chr.9: 108936322-108936322 on Build GRCh38
rs976111097 Chr.9: 108936626-108936626 on Build GRCh38
rs767709014 Chr.9: 108936362-108936365 on Build GRCh38
rs758552237 Chr.9: 108936465-108936465 on Build GRCh38
rs751576724 Chr.9: 108936304-108936304 on Build GRCh38
rs891805316 Chr.9: 108936570-108936570 on Build GRCh38
rs773045726 Chr.9: 108936408-108936408 on Build GRCh38
rs369393874 Chr.9: 108936477-108936477 on Build GRCh38
rs771095283 Chr.9: 108936345-108936345 on Build GRCh38
rs148367154 Chr.9: 108936319-108936319 on Build GRCh38
rs767557342 Chr.9: 108936365-108936365 on Build GRCh38
rs962359332 Chr.9: 108936291-108936291 on Build GRCh38
rs201136708 Chr.9: 108936341-108936341 on Build GRCh38
rs542450367 Chr.9: 108936531-108936531 on Build GRCh38
rs779086062 Chr.9: 108936452-108936452 on Build GRCh38
rs374811797 Chr.9: 108936474-108936474 on Build GRCh38
rs541205016 Chr.9: 108936601-108936601 on Build GRCh38
rs746154891 Chr.9: 108936461-108936461 on Build GRCh38
rs770316896 Chr.9: 108936369-108936369 on Build GRCh38
rs527867742 Chr.9: 108936593-108936593 on Build GRCh38
rs556836038 Chr.9: 108936439-108936439 on Build GRCh38
rs10979615 Chr.9: 108936509-108936509 on Build GRCh38
rs372660944 Chr.9: 108936312-108936312 on Build GRCh38
rs772251201 Chr.9: 108936354-108936354 on Build GRCh38
rs779137585 Chr.9: 108936350-108936350 on Build GRCh38
rs754889343 Chr.9: 108936430-108936430 on Build GRCh38
rs191909577 Chr.9: 108936286-108936286 on Build GRCh38
rs200258029 Chr.9: 108936386-108936386 on Build GRCh38
rs775854895 Chr.9: 108936360-108936360 on Build GRCh38
rs764949272 Chr.9: 108936396-108936396 on Build GRCh38
rs762296322 Chr.9: 108936392-108936392 on Build GRCh38

View Full Product Details