Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108952355-108952856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNAL1
Entrez Gene ID
8727
Gene Name
catenin alpha like 1
Gene Aliases
ACRP, CLLP, alpha-CATU
UniGene
Hs.58488
Gene Chromosome Location
Chr.9: 108942569-109013594 on Build GRCh38
Transcripts
NM_001286974.1 NM_003798.3 XM_005252291.3 XM_017015250.1 XM_017015251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6572812 c.1672G>C p.D558H Chr.9: 108952452-108952452 on Build GRCh38
6572813 c.1672G>C p.D558H Chr.9: 108952452-108952452 on Build GRCh38
1103692 c.1672G>A p.D558N Chr.9: 108952452-108952452 on Build GRCh38
5345679 c.1689C>A p.A563A Chr.9: 108952355-108952355 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs910653256 Chr.9: 108952794-108952795 on Build GRCh38
rs776678644 Chr.9: 108952411-108952411 on Build GRCh38
rs138264904 Chr.9: 108952798-108952798 on Build GRCh38
rs766593973 Chr.9: 108952544-108952545 on Build GRCh38
rs201955013 Chr.9: 108952428-108952428 on Build GRCh38
rs374055269 Chr.9: 108952537-108952537 on Build GRCh38
rs377052710 Chr.9: 108952478-108952478 on Build GRCh38
rs1025547428 Chr.9: 108952693-108952693 on Build GRCh38
rs926604202 Chr.9: 108952416-108952416 on Build GRCh38
rs774185002 Chr.9: 108952508-108952513 on Build GRCh38
rs538528837 Chr.9: 108952531-108952531 on Build GRCh38
rs372333801 Chr.9: 108952536-108952536 on Build GRCh38
rs374673138 Chr.9: 108952710-108952710 on Build GRCh38
rs754431959 Chr.9: 108952521-108952521 on Build GRCh38
rs34922868 Chr.9: 108952459-108952459 on Build GRCh38
rs747940760 Chr.9: 108952422-108952422 on Build GRCh38
rs925577363 Chr.9: 108952399-108952399 on Build GRCh38
rs765524419 Chr.9: 108952523-108952523 on Build GRCh38
rs370369097 Chr.9: 108952511-108952511 on Build GRCh38
rs993555763 Chr.9: 108952615-108952615 on Build GRCh38
rs759404385 Chr.9: 108952646-108952646 on Build GRCh38
rs575064364 Chr.9: 108952550-108952550 on Build GRCh38
rs973345002 Chr.9: 108952760-108952760 on Build GRCh38
rs765171468 Chr.9: 108952769-108952769 on Build GRCh38
rs28361164 Chr.9: 108952645-108952645 on Build GRCh38
rs767697207 Chr.9: 108952636-108952636 on Build GRCh38
rs555922094 Chr.9: 108952766-108952766 on Build GRCh38
rs1051766596 Chr.9: 108952764-108952764 on Build GRCh38
rs779796675 Chr.9: 108952533-108952533 on Build GRCh38
rs28361165 Chr.9: 108952633-108952633 on Build GRCh38
rs759774317 Chr.9: 108952447-108952447 on Build GRCh38
rs988544742 Chr.9: 108952741-108952742 on Build GRCh38
rs868303230 Chr.9: 108952446-108952446 on Build GRCh38
rs563730264 Chr.9: 108952759-108952759 on Build GRCh38
rs373436178 Chr.9: 108952545-108952546 on Build GRCh38
rs184054575 Chr.9: 108952784-108952784 on Build GRCh38
rs768597170 Chr.9: 108952505-108952506 on Build GRCh38
rs1017380919 Chr.9: 108952574-108952574 on Build GRCh38
rs115211704 Chr.9: 108952752-108952752 on Build GRCh38
rs191491997 Chr.9: 108952534-108952534 on Build GRCh38
rs780988926 Chr.9: 108952546-108952546 on Build GRCh38
rs956907437 Chr.9: 108952654-108952654 on Build GRCh38
rs980960141 Chr.9: 108952679-108952679 on Build GRCh38
rs750804937 Chr.9: 108952529-108952529 on Build GRCh38
rs988425031 Chr.9: 108952672-108952672 on Build GRCh38
rs749160663 Chr.9: 108952490-108952492 on Build GRCh38
rs543293131 Chr.9: 108952790-108952790 on Build GRCh38
rs754572519 Chr.9: 108952545-108952545 on Build GRCh38
rs1051540934 Chr.9: 108952502-108952502 on Build GRCh38
rs970949454 Chr.9: 108952736-108952736 on Build GRCh38
rs548514291 Chr.9: 108952390-108952390 on Build GRCh38
rs375698582 Chr.9: 108952815-108952815 on Build GRCh38
rs28361166 Chr.9: 108952616-108952616 on Build GRCh38
rs1051799126 Chr.9: 108952517-108952517 on Build GRCh38
rs545744773 Chr.9: 108952758-108952758 on Build GRCh38
rs777441339 Chr.9: 108952408-108952408 on Build GRCh38
rs754077027 Chr.9: 108952434-108952434 on Build GRCh38
rs773147940 Chr.9: 108952437-108952437 on Build GRCh38
rs936649855 Chr.9: 108952401-108952401 on Build GRCh38
rs761580883 Chr.9: 108952511-108952512 on Build GRCh38
rs942012630 Chr.9: 108952424-108952424 on Build GRCh38
rs925513217 Chr.9: 108952731-108952731 on Build GRCh38
rs534166580 Chr.9: 108952410-108952410 on Build GRCh38

View Full Product Details