Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69324471-69324964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
UniGene
Hs.118003
Gene Chromosome Location
Chr.9: 69324572-69392461 on Build GRCh38
Transcripts
NM_001127608.1 XM_005252307.4 XM_017015325.1

SNP Detail

SNP ID Chromosome Location
rs542274416 Chr.9: 69324834-69324834 on Build GRCh38
rs1033060089 Chr.9: 69324679-69324679 on Build GRCh38
rs971280635 Chr.9: 69324581-69324581 on Build GRCh38
rs1040587368 Chr.9: 69324642-69324642 on Build GRCh38
rs139986906 Chr.9: 69324621-69324621 on Build GRCh38
rs1048502893 Chr.9: 69324894-69324894 on Build GRCh38
rs919594922 Chr.9: 69324613-69324613 on Build GRCh38
rs539252912 Chr.9: 69324658-69324658 on Build GRCh38
rs959994011 Chr.9: 69324598-69324598 on Build GRCh38
rs956616205 Chr.9: 69324861-69324861 on Build GRCh38
rs866798526 Chr.9: 69324890-69324890 on Build GRCh38
rs1039654217 Chr.9: 69324827-69324827 on Build GRCh38
rs900648165 Chr.9: 69324667-69324667 on Build GRCh38
rs568025774 Chr.9: 69324603-69324603 on Build GRCh38
rs574794195 Chr.9: 69324813-69324813 on Build GRCh38
rs537057602 Chr.9: 69324554-69324554 on Build GRCh38
rs552760454 Chr.9: 69324523-69324523 on Build GRCh38
rs978535947 Chr.9: 69324636-69324636 on Build GRCh38
rs929705027 Chr.9: 69324884-69324884 on Build GRCh38
rs375234820 Chr.9: 69324904-69324904 on Build GRCh38
rs944958125 Chr.9: 69324905-69324905 on Build GRCh38
rs570569087 Chr.9: 69324526-69324526 on Build GRCh38
rs1035978935 Chr.9: 69324586-69324586 on Build GRCh38
rs575167703 Chr.9: 69324639-69324639 on Build GRCh38
rs534695921 Chr.9: 69324561-69324561 on Build GRCh38
rs1004420962 Chr.9: 69324916-69324916 on Build GRCh38
rs899865712 Chr.9: 69324858-69324858 on Build GRCh38
rs937112620 Chr.9: 69324592-69324592 on Build GRCh38
rs1040618932 Chr.9: 69324918-69324918 on Build GRCh38
rs576971989 Chr.9: 69324718-69324718 on Build GRCh38
rs927114505 Chr.9: 69324587-69324587 on Build GRCh38
rs893208965 Chr.9: 69324705-69324705 on Build GRCh38
rs991125315 Chr.9: 69324601-69324601 on Build GRCh38
rs919724476 Chr.9: 69324831-69324831 on Build GRCh38
rs10867357 Chr.9: 69324515-69324515 on Build GRCh38
rs1010272809 Chr.9: 69324711-69324711 on Build GRCh38
rs1052618476 Chr.9: 69324748-69324748 on Build GRCh38
rs1035542338 Chr.9: 69324927-69324927 on Build GRCh38
rs929617765 Chr.9: 69324614-69324614 on Build GRCh38
rs887203326 Chr.9: 69324907-69324907 on Build GRCh38
rs1008612548 Chr.9: 69324506-69324506 on Build GRCh38
rs563796644 Chr.9: 69324513-69324513 on Build GRCh38
rs912212965 Chr.9: 69324903-69324903 on Build GRCh38
rs935536194 Chr.9: 69324666-69324666 on Build GRCh38
rs995564323 Chr.9: 69324518-69324518 on Build GRCh38
rs1020691410 Chr.9: 69324714-69324714 on Build GRCh38
rs6559468 Chr.9: 69324930-69324930 on Build GRCh38
rs886722835 Chr.9: 69324632-69324632 on Build GRCh38
rs925433744 Chr.9: 69324657-69324657 on Build GRCh38
rs73649651 Chr.9: 69324584-69324584 on Build GRCh38
rs989138428 Chr.9: 69324539-69324539 on Build GRCh38
rs1012863145 Chr.9: 69324939-69324939 on Build GRCh38
rs145935254 Chr.9: 69324620-69324620 on Build GRCh38
rs745739607 Chr.9: 69324624-69324624 on Build GRCh38
rs996273880 Chr.9: 69324677-69324677 on Build GRCh38
rs78986996 Chr.9: 69324770-69324770 on Build GRCh38
rs958499300 Chr.9: 69324758-69324758 on Build GRCh38
rs944313712 Chr.9: 69324793-69324793 on Build GRCh38
rs540882984 Chr.9: 69324708-69324708 on Build GRCh38
rs981303065 Chr.9: 69324741-69324741 on Build GRCh38
rs1026755100 Chr.9: 69324538-69324538 on Build GRCh38
rs1020240476 Chr.9: 69324544-69324544 on Build GRCh38
rs548850584 Chr.9: 69324510-69324510 on Build GRCh38
rs751279126 Chr.9: 69324642-69324643 on Build GRCh38
rs541494781 Chr.9: 69324744-69324744 on Build GRCh38
rs753974487 Chr.9: 69324653-69324653 on Build GRCh38

View Full Product Details