Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69456157-69456656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA1
Entrez Gene ID
320
Gene Name
amyloid beta precursor protein binding family A member 1
Gene Aliases
D9S411E, LIN10, MINT1, X11, X11A, X11ALPHA
UniGene
Hs.171939
Gene Chromosome Location
Chr.9: 69427532-69673012 on Build GRCh38
Transcripts
NM_001163.3 XM_005251968.3 XM_011518617.2 XM_017014670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109723 c.1662C>T p.I554I Chr.9: 69456373-69456373 on Build GRCh38
6856537 c.1747delA p.R583fs*5 Chr.9: 69456288-69456288 on Build GRCh38
5346471 c.1771G>A p.V591I Chr.9: 69456264-69456264 on Build GRCh38
278897 c.1776C>T p.F592F Chr.9: 69456259-69456259 on Build GRCh38
6115716 c.1701C>A p.N567K Chr.9: 69456334-69456334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771955316 Chr.9: 69456341-69456341 on Build GRCh38
rs747776055 Chr.9: 69456215-69456215 on Build GRCh38
rs369001002 Chr.9: 69456216-69456216 on Build GRCh38
rs760579040 Chr.9: 69456456-69456457 on Build GRCh38
rs911621596 Chr.9: 69456218-69456218 on Build GRCh38
rs769295451 Chr.9: 69456258-69456258 on Build GRCh38
rs143979575 Chr.9: 69456426-69456426 on Build GRCh38
rs955469535 Chr.9: 69456329-69456329 on Build GRCh38
rs745770993 Chr.9: 69456404-69456404 on Build GRCh38
rs181071682 Chr.9: 69456292-69456292 on Build GRCh38
rs904112830 Chr.9: 69456585-69456585 on Build GRCh38
rs759341933 Chr.9: 69456257-69456257 on Build GRCh38
rs1055575156 Chr.9: 69456524-69456524 on Build GRCh38
rs754506281 Chr.9: 69456307-69456307 on Build GRCh38
rs199844166 Chr.9: 69456354-69456354 on Build GRCh38
rs752115782 Chr.9: 69456454-69456454 on Build GRCh38
rs371943178 Chr.9: 69456392-69456392 on Build GRCh38
rs755783658 Chr.9: 69456369-69456369 on Build GRCh38
rs183303717 Chr.9: 69456578-69456578 on Build GRCh38
rs368799633 Chr.9: 69456352-69456352 on Build GRCh38
rs935614187 Chr.9: 69456491-69456491 on Build GRCh38
rs780763273 Chr.9: 69456201-69456201 on Build GRCh38
rs112803189 Chr.9: 69456259-69456259 on Build GRCh38
rs775939545 Chr.9: 69456504-69456504 on Build GRCh38
rs753546502 Chr.9: 69456382-69456382 on Build GRCh38
rs777319156 Chr.9: 69456314-69456314 on Build GRCh38
rs770573595 Chr.9: 69456322-69456322 on Build GRCh38
rs950986014 Chr.9: 69456346-69456371 on Build GRCh38
rs146869320 Chr.9: 69456373-69456373 on Build GRCh38
rs572878528 Chr.9: 69456342-69456342 on Build GRCh38
rs906648417 Chr.9: 69456559-69456559 on Build GRCh38
rs201589566 Chr.9: 69456241-69456241 on Build GRCh38
rs768508523 Chr.9: 69456413-69456413 on Build GRCh38
rs987350128 Chr.9: 69456594-69456596 on Build GRCh38
rs557817337 Chr.9: 69456338-69456338 on Build GRCh38
rs750215572 Chr.9: 69456240-69456240 on Build GRCh38
rs1035203109 Chr.9: 69456263-69456263 on Build GRCh38
rs746177560 Chr.9: 69456381-69456381 on Build GRCh38
rs374176608 Chr.9: 69456356-69456356 on Build GRCh38
rs186990223 Chr.9: 69456313-69456313 on Build GRCh38
rs767949732 Chr.9: 69456463-69456463 on Build GRCh38
rs564013390 Chr.9: 69456515-69456515 on Build GRCh38
rs766858258 Chr.9: 69456453-69456453 on Build GRCh38
rs780425686 Chr.9: 69456305-69456305 on Build GRCh38
rs374314289 Chr.9: 69456219-69456219 on Build GRCh38
rs955398638 Chr.9: 69456619-69456619 on Build GRCh38
rs374525521 Chr.9: 69456476-69456476 on Build GRCh38
rs938307112 Chr.9: 69456523-69456523 on Build GRCh38
rs756781899 Chr.9: 69456388-69456388 on Build GRCh38
rs188750369 Chr.9: 69456590-69456590 on Build GRCh38
rs758711760 Chr.9: 69456298-69456298 on Build GRCh38
rs759765670 Chr.9: 69456458-69456458 on Build GRCh38
rs564578727 Chr.9: 69456563-69456563 on Build GRCh38
rs778545929 Chr.9: 69456481-69456481 on Build GRCh38
rs760930916 Chr.9: 69456344-69456344 on Build GRCh38
rs772186318 Chr.9: 69456408-69456408 on Build GRCh38
rs906841167 Chr.9: 69456505-69456505 on Build GRCh38
rs765665436 Chr.9: 69456283-69456283 on Build GRCh38
rs529100051 Chr.9: 69456587-69456587 on Build GRCh38
rs757011030 Chr.9: 69456477-69456477 on Build GRCh38
rs545907084 Chr.9: 69456555-69456555 on Build GRCh38
rs763433932 Chr.9: 69456446-69456446 on Build GRCh38
rs375286686 Chr.9: 69456393-69456393 on Build GRCh38
rs766705353 Chr.9: 69456357-69456357 on Build GRCh38
rs149106686 Chr.9: 69456577-69456577 on Build GRCh38
rs535189113 Chr.9: 69456350-69456350 on Build GRCh38
rs746715453 Chr.9: 69456412-69456412 on Build GRCh38
rs757777831 Chr.9: 69456281-69456281 on Build GRCh38
rs1571851 Chr.9: 69456466-69456466 on Build GRCh38
rs943098373 Chr.9: 69456432-69456432 on Build GRCh38
rs749223214 Chr.9: 69456232-69456232 on Build GRCh38
rs202066084 Chr.9: 69456414-69456414 on Build GRCh38
rs368416881 Chr.9: 69456371-69456371 on Build GRCh38
rs550114743 Chr.9: 69456268-69456268 on Build GRCh38
rs1019208530 Chr.9: 69456609-69456609 on Build GRCh38
rs201117945 Chr.9: 69456200-69456200 on Build GRCh38
rs987706331 Chr.9: 69456423-69456423 on Build GRCh38
rs201954883 Chr.9: 69456321-69456321 on Build GRCh38
rs764203670 Chr.9: 69456265-69456265 on Build GRCh38
rs751164226 Chr.9: 69456264-69456264 on Build GRCh38
rs894206697 Chr.9: 69456569-69456569 on Build GRCh38
rs1039090608 Chr.9: 69456282-69456282 on Build GRCh38
rs774287316 Chr.9: 69456256-69456256 on Build GRCh38
rs762051788 Chr.9: 69456418-69456418 on Build GRCh38
rs773326051 Chr.9: 69456436-69456436 on Build GRCh38
rs1001131971 Chr.9: 69456293-69456293 on Build GRCh38
rs140724658 Chr.9: 69456409-69456409 on Build GRCh38
rs575581393 Chr.9: 69456517-69456517 on Build GRCh38
rs774376859 Chr.9: 69456334-69456334 on Build GRCh38
rs370688256 Chr.9: 69456353-69456353 on Build GRCh38
rs993883739 Chr.9: 69456195-69456195 on Build GRCh38
rs1046982942 Chr.9: 69456526-69456526 on Build GRCh38
rs919895073 Chr.9: 69456456-69456456 on Build GRCh38

View Full Product Details