Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76503692-76504208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT1
Entrez Gene ID
2650
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2
UniGene
Hs.521568
Gene Chromosome Location
Chr.9: 76441666-76507416 on Build GRCh38
Transcripts
NM_001097633.1 NM_001097634.1 NM_001097635.1 NM_001097636.1 NM_001490.4 XM_017014620.1 XM_017014621.1

SNP Detail

SNP ID Chromosome Location
rs557684688 Chr.9: 76503914-76503914 on Build GRCh38
rs899969039 Chr.9: 76503938-76503938 on Build GRCh38
rs373430820 Chr.9: 76504124-76504128 on Build GRCh38
rs913304435 Chr.9: 76503821-76503821 on Build GRCh38
rs947396688 Chr.9: 76503840-76503840 on Build GRCh38
rs928790227 Chr.9: 76503785-76503785 on Build GRCh38
rs142834264 Chr.9: 76503943-76503943 on Build GRCh38
rs991198757 Chr.9: 76504027-76504027 on Build GRCh38
rs1051097862 Chr.9: 76504148-76504148 on Build GRCh38
rs904086580 Chr.9: 76503978-76503978 on Build GRCh38
rs991187335 Chr.9: 76503780-76503780 on Build GRCh38
rs772694784 Chr.9: 76503964-76503964 on Build GRCh38
rs561713871 Chr.9: 76504107-76504107 on Build GRCh38
rs932755020 Chr.9: 76504144-76504147 on Build GRCh38
rs191384358 Chr.9: 76503962-76503962 on Build GRCh38
rs779195733 Chr.9: 76504161-76504161 on Build GRCh38
rs560994326 Chr.9: 76503955-76503955 on Build GRCh38
rs921879624 Chr.9: 76503894-76503894 on Build GRCh38
rs970334784 Chr.9: 76503776-76503776 on Build GRCh38
rs756453881 Chr.9: 76504018-76504018 on Build GRCh38
rs941221969 Chr.9: 76503898-76503898 on Build GRCh38
rs971443215 Chr.9: 76504054-76504054 on Build GRCh38
rs983082949 Chr.9: 76503896-76503896 on Build GRCh38
rs988904959 Chr.9: 76503807-76503807 on Build GRCh38
rs199537430 Chr.9: 76503718-76503718 on Build GRCh38
rs960221221 Chr.9: 76504098-76504098 on Build GRCh38
rs898901554 Chr.9: 76504004-76504004 on Build GRCh38
rs764493703 Chr.9: 76504102-76504102 on Build GRCh38
rs113122064 Chr.9: 76504138-76504138 on Build GRCh38
rs982457675 Chr.9: 76504056-76504056 on Build GRCh38
rs199566462 Chr.9: 76504163-76504163 on Build GRCh38
rs1035915862 Chr.9: 76504072-76504072 on Build GRCh38
rs537323044 Chr.9: 76503769-76503769 on Build GRCh38
rs979222080 Chr.9: 76504150-76504150 on Build GRCh38
rs945391844 Chr.9: 76504157-76504157 on Build GRCh38
rs762389698 Chr.9: 76503965-76503965 on Build GRCh38
rs978780990 Chr.9: 76503865-76503865 on Build GRCh38
rs1057419 Chr.9: 76503745-76503745 on Build GRCh38
rs867876364 Chr.9: 76504066-76504066 on Build GRCh38
rs761718662 Chr.9: 76503832-76503832 on Build GRCh38
rs540768389 Chr.9: 76503748-76503748 on Build GRCh38
rs1001812314 Chr.9: 76504070-76504070 on Build GRCh38
rs1038151009 Chr.9: 76503911-76503912 on Build GRCh38
rs960171935 Chr.9: 76503799-76503799 on Build GRCh38
rs757831986 Chr.9: 76504155-76504155 on Build GRCh38
rs979611357 Chr.9: 76503757-76503757 on Build GRCh38
rs570385631 Chr.9: 76504172-76504172 on Build GRCh38
rs908480191 Chr.9: 76504105-76504105 on Build GRCh38
rs754434672 Chr.9: 76504124-76504124 on Build GRCh38
rs577302091 Chr.9: 76503922-76503922 on Build GRCh38
rs908385692 Chr.9: 76503897-76503897 on Build GRCh38
rs904817106 Chr.9: 76504043-76504043 on Build GRCh38
rs1032784730 Chr.9: 76503759-76503759 on Build GRCh38
rs980726755 Chr.9: 76503777-76503777 on Build GRCh38
rs41288761 Chr.9: 76504012-76504012 on Build GRCh38
rs921055596 Chr.9: 76504141-76504141 on Build GRCh38
rs707739 Chr.9: 76503987-76503987 on Build GRCh38

View Full Product Details