Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83837183-83837683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
UniGene
Hs.697514
Gene Chromosome Location
Chr.9: 83832795-83921539 on Build GRCh38
Transcripts
NM_001271927.1 NM_001271928.1 NM_017576.2 XM_011518848.2 XM_011518849.2 XM_011518850.2 XM_011518854.2 XM_011518857.2 XM_011518858.2 XM_017014900.1 XM_017014901.1 XM_017014902.1 XM_017014903.1 XM_017014904.1 XM_017014905.1 XM_017014906.1 XM_017014907.1 XM_017014908.1 XM_017014909.1 XM_017014910.1 XM_017014911.1 XM_017014912.1 XM_017014913.1 XM_017014915.1 XM_017014916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6039561 c.3936delC p.S1313fs*5 Chr.9: 83837271-83837271 on Build GRCh38
223343 c.3874C>T p.P1292S Chr.9: 83837333-83837333 on Build GRCh38
4164069 c.3875C>T p.P1292L Chr.9: 83837332-83837332 on Build GRCh38
6013519 c.3800C>T p.P1267L Chr.9: 83837407-83837407 on Build GRCh38
261664 c.3923C>A p.S1308Y Chr.9: 83837284-83837284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775815260 Chr.9: 83837213-83837213 on Build GRCh38
rs768932345 Chr.9: 83837443-83837443 on Build GRCh38
rs866103281 Chr.9: 83837605-83837605 on Build GRCh38
rs2542780 Chr.9: 83837480-83837480 on Build GRCh38
rs781620659 Chr.9: 83837332-83837332 on Build GRCh38
rs374990519 Chr.9: 83837250-83837250 on Build GRCh38
rs1009452017 Chr.9: 83837217-83837217 on Build GRCh38
rs775622981 Chr.9: 83837271-83837271 on Build GRCh38
rs761903243 Chr.9: 83837245-83837245 on Build GRCh38
rs528796605 Chr.9: 83837380-83837380 on Build GRCh38
rs2780139 Chr.9: 83837597-83837597 on Build GRCh38
rs769982330 Chr.9: 83837351-83837351 on Build GRCh38
rs140117578 Chr.9: 83837311-83837311 on Build GRCh38
rs201708301 Chr.9: 83837371-83837371 on Build GRCh38
rs1036405268 Chr.9: 83837450-83837452 on Build GRCh38
rs770791845 Chr.9: 83837212-83837223 on Build GRCh38
rs769116359 Chr.9: 83837233-83837233 on Build GRCh38
rs148632153 Chr.9: 83837579-83837579 on Build GRCh38
rs769173452 Chr.9: 83837318-83837318 on Build GRCh38
rs112299613 Chr.9: 83837655-83837655 on Build GRCh38
rs756162275 Chr.9: 83837272-83837272 on Build GRCh38
rs747561292 Chr.9: 83837215-83837215 on Build GRCh38
rs779629692 Chr.9: 83837525-83837525 on Build GRCh38
rs561598508 Chr.9: 83837309-83837309 on Build GRCh38
rs147814329 Chr.9: 83837641-83837641 on Build GRCh38
rs764848149 Chr.9: 83837513-83837513 on Build GRCh38
rs778438479 Chr.9: 83837287-83837287 on Build GRCh38
rs764144016 Chr.9: 83837422-83837422 on Build GRCh38
rs149899595 Chr.9: 83837428-83837428 on Build GRCh38
rs1009232940 Chr.9: 83837587-83837587 on Build GRCh38
rs771248074 Chr.9: 83837401-83837401 on Build GRCh38
rs765389677 Chr.9: 83837247-83837247 on Build GRCh38
rs767480512 Chr.9: 83837263-83837263 on Build GRCh38
rs953882593 Chr.9: 83837647-83837647 on Build GRCh38
rs756736442 Chr.9: 83837284-83837284 on Build GRCh38
rs753745000 Chr.9: 83837425-83837425 on Build GRCh38
rs905380950 Chr.9: 83837524-83837524 on Build GRCh38
rs775457038 Chr.9: 83837404-83837404 on Build GRCh38
rs566727478 Chr.9: 83837648-83837648 on Build GRCh38
rs749527975 Chr.9: 83837427-83837429 on Build GRCh38
rs776719927 Chr.9: 83837238-83837238 on Build GRCh38
rs1055101 Chr.9: 83837539-83837539 on Build GRCh38
rs751933982 Chr.9: 83837535-83837535 on Build GRCh38
rs919485448 Chr.9: 83837468-83837468 on Build GRCh38
rs763083436 Chr.9: 83837252-83837252 on Build GRCh38
rs965659221 Chr.9: 83837623-83837623 on Build GRCh38
rs764086639 Chr.9: 83837276-83837276 on Build GRCh38
rs540787693 Chr.9: 83837405-83837405 on Build GRCh38
rs752519365 Chr.9: 83837266-83837266 on Build GRCh38
rs1053542774 Chr.9: 83837575-83837575 on Build GRCh38
rs532802658 Chr.9: 83837519-83837519 on Build GRCh38
rs369903546 Chr.9: 83837515-83837515 on Build GRCh38
rs748179128 Chr.9: 83837344-83837344 on Build GRCh38

View Full Product Details