Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83853428-83853930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
UniGene
Hs.697514
Gene Chromosome Location
Chr.9: 83832795-83921539 on Build GRCh38
Transcripts
NM_001271927.1 NM_001271928.1 NM_017576.2 XM_011518848.2 XM_011518849.2 XM_011518850.2 XM_011518854.2 XM_011518856.2 XM_011518857.2 XM_011518858.2 XM_017014900.1 XM_017014901.1 XM_017014902.1 XM_017014903.1 XM_017014904.1 XM_017014905.1 XM_017014906.1 XM_017014907.1 XM_017014908.1 XM_017014909.1 XM_017014910.1 XM_017014911.1 XM_017014912.1 XM_017014913.1 XM_017014915.1 XM_017014916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764031 c.3186T>A p.I1062I Chr.9: 83853800-83853800 on Build GRCh38
754010 c.3327T>C p.I1109I Chr.9: 83853659-83853659 on Build GRCh38
1463303 c.3156A>G p.E1052E Chr.9: 83853830-83853830 on Build GRCh38
6889273 c.3151-1G>A p.? Chr.9: 83853836-83853836 on Build GRCh38
6307452 c.3337C>A p.L1113I Chr.9: 83853649-83853649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028371114 Chr.9: 83853566-83853566 on Build GRCh38
rs761550435 Chr.9: 83853592-83853592 on Build GRCh38
rs2542785 Chr.9: 83853469-83853469 on Build GRCh38
rs879094423 Chr.9: 83853739-83853739 on Build GRCh38
rs780834995 Chr.9: 83853702-83853702 on Build GRCh38
rs764019170 Chr.9: 83853895-83853895 on Build GRCh38
rs779980486 Chr.9: 83853688-83853688 on Build GRCh38
rs746503082 Chr.9: 83853694-83853694 on Build GRCh38
rs182257916 Chr.9: 83853846-83853846 on Build GRCh38
rs371974528 Chr.9: 83853780-83853780 on Build GRCh38
rs565946178 Chr.9: 83853700-83853700 on Build GRCh38
rs531876952 Chr.9: 83853644-83853644 on Build GRCh38
rs771355639 Chr.9: 83853761-83853761 on Build GRCh38
rs143915976 Chr.9: 83853828-83853828 on Build GRCh38
rs954290127 Chr.9: 83853574-83853574 on Build GRCh38
rs557876823 Chr.9: 83853881-83853881 on Build GRCh38
rs759556164 Chr.9: 83853652-83853652 on Build GRCh38
rs776353624 Chr.9: 83853585-83853585 on Build GRCh38
rs749616984 Chr.9: 83853759-83853759 on Build GRCh38
rs369251410 Chr.9: 83853805-83853805 on Build GRCh38
rs295274 Chr.9: 83853800-83853800 on Build GRCh38
rs2780147 Chr.9: 83853464-83853464 on Build GRCh38
rs765024852 Chr.9: 83853886-83853887 on Build GRCh38
rs756317575 Chr.9: 83853673-83853673 on Build GRCh38
rs370735815 Chr.9: 83853811-83853811 on Build GRCh38
rs150848943 Chr.9: 83853650-83853650 on Build GRCh38
rs758141623 Chr.9: 83853814-83853814 on Build GRCh38
rs768431842 Chr.9: 83853579-83853579 on Build GRCh38
rs780888167 Chr.9: 83853843-83853843 on Build GRCh38
rs774856531 Chr.9: 83853767-83853767 on Build GRCh38
rs1054546396 Chr.9: 83853465-83853465 on Build GRCh38
rs891679730 Chr.9: 83853483-83853483 on Build GRCh38
rs767645576 Chr.9: 83853781-83853781 on Build GRCh38
rs771394909 Chr.9: 83853883-83853883 on Build GRCh38
rs755852835 Chr.9: 83853858-83853858 on Build GRCh38
rs747850098 Chr.9: 83853711-83853711 on Build GRCh38
rs773480141 Chr.9: 83853751-83853751 on Build GRCh38
rs773601599 Chr.9: 83853617-83853617 on Build GRCh38
rs1010141855 Chr.9: 83853513-83853513 on Build GRCh38
rs1029256833 Chr.9: 83853904-83853904 on Build GRCh38
rs775528711 Chr.9: 83853784-83853784 on Build GRCh38
rs777427669 Chr.9: 83853865-83853865 on Build GRCh38
rs758286417 Chr.9: 83853680-83853680 on Build GRCh38
rs146522709 Chr.9: 83853725-83853725 on Build GRCh38
rs754309991 Chr.9: 83853678-83853679 on Build GRCh38
rs980240572 Chr.9: 83853772-83853772 on Build GRCh38
rs999824287 Chr.9: 83853455-83853455 on Build GRCh38
rs774568802 Chr.9: 83853884-83853884 on Build GRCh38
rs965863210 Chr.9: 83853551-83853551 on Build GRCh38
rs144794163 Chr.9: 83853560-83853560 on Build GRCh38
rs143627829 Chr.9: 83853677-83853677 on Build GRCh38
rs909861058 Chr.9: 83853655-83853655 on Build GRCh38
rs529481603 Chr.9: 83853712-83853712 on Build GRCh38
rs770338804 Chr.9: 83853601-83853601 on Build GRCh38
rs766080244 Chr.9: 83853815-83853815 on Build GRCh38
rs763409241 Chr.9: 83853633-83853633 on Build GRCh38
rs995988464 Chr.9: 83853903-83853903 on Build GRCh38
rs950185206 Chr.9: 83853750-83853750 on Build GRCh38
rs1018914523 Chr.9: 83853536-83853536 on Build GRCh38
rs767731776 Chr.9: 83853653-83853653 on Build GRCh38
rs2542786 Chr.9: 83853503-83853503 on Build GRCh38
rs917532006 Chr.9: 83853723-83853723 on Build GRCh38
rs771359304 Chr.9: 83853830-83853832 on Build GRCh38
rs111290584 Chr.9: 83853826-83853826 on Build GRCh38
rs752987077 Chr.9: 83853672-83853672 on Build GRCh38
rs765070190 Chr.9: 83853674-83853674 on Build GRCh38

View Full Product Details