Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135809553-135810066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP1
Entrez Gene ID
157922
Gene Name
calmodulin regulated spectrin associated protein 1
Gene Aliases
-
UniGene
Hs.522493
Gene Chromosome Location
Chr.9: 135808487-135907551 on Build GRCh38
Transcripts
NM_015447.3 XM_005263396.3 XM_005263397.1 XM_011518272.1 XM_011518273.2 XM_011518274.1 XM_017014301.1 XM_017014302.1

SNP Detail

SNP ID Chromosome Location
rs1034235825 Chr.9: 135809862-135809862 on Build GRCh38
rs751134561 Chr.9: 135809776-135809776 on Build GRCh38
rs866531446 Chr.9: 135809922-135809925 on Build GRCh38
rs11553781 Chr.9: 135809997-135809997 on Build GRCh38
rs756059376 Chr.9: 135809941-135809941 on Build GRCh38
rs1046721146 Chr.9: 135809799-135809799 on Build GRCh38
rs900905421 Chr.9: 135809774-135809774 on Build GRCh38
rs941020055 Chr.9: 135809944-135809944 on Build GRCh38
rs541725007 Chr.9: 135810018-135810018 on Build GRCh38
rs942956380 Chr.9: 135809987-135809987 on Build GRCh38
rs567078197 Chr.9: 135809700-135809700 on Build GRCh38
rs193109960 Chr.9: 135809658-135809658 on Build GRCh38
rs1030997152 Chr.9: 135809933-135809933 on Build GRCh38
rs955376431 Chr.9: 135809731-135809731 on Build GRCh38
rs779247727 Chr.9: 135809804-135809804 on Build GRCh38
rs7838 Chr.9: 135809677-135809677 on Build GRCh38
rs575402273 Chr.9: 135809948-135809948 on Build GRCh38
rs745807467 Chr.9: 135809930-135809930 on Build GRCh38
rs559653861 Chr.9: 135809791-135809791 on Build GRCh38
rs115943853 Chr.9: 135810021-135810021 on Build GRCh38
rs911088707 Chr.9: 135809756-135809756 on Build GRCh38
rs1024582953 Chr.9: 135809915-135809915 on Build GRCh38
rs1038072441 Chr.9: 135809980-135809980 on Build GRCh38
rs1055779672 Chr.9: 135809617-135809617 on Build GRCh38
rs1055385658 Chr.9: 135809900-135809900 on Build GRCh38
rs62584792 Chr.9: 135809687-135809687 on Build GRCh38
rs546546450 Chr.9: 135809733-135809733 on Build GRCh38
rs557054241 Chr.9: 135809919-135809919 on Build GRCh38
rs369751836 Chr.9: 135809960-135809961 on Build GRCh38
rs890963802 Chr.9: 135810022-135810022 on Build GRCh38
rs571244473 Chr.9: 135809739-135809739 on Build GRCh38
rs1007112569 Chr.9: 135810037-135810037 on Build GRCh38
rs765596133 Chr.9: 135809579-135809579 on Build GRCh38
rs1042864028 Chr.9: 135809785-135809785 on Build GRCh38
rs755377144 Chr.9: 135809742-135809742 on Build GRCh38
rs56381246 Chr.9: 135809806-135809806 on Build GRCh38
rs3201247 Chr.9: 135809879-135809879 on Build GRCh38
rs945488883 Chr.9: 135809784-135809784 on Build GRCh38
rs574244407 Chr.9: 135810007-135810007 on Build GRCh38
rs772470612 Chr.9: 135809940-135809941 on Build GRCh38
rs185152787 Chr.9: 135809981-135809981 on Build GRCh38
rs958033218 Chr.9: 135809953-135809954 on Build GRCh38
rs1014058327 Chr.9: 135809883-135809883 on Build GRCh38
rs141287658 Chr.9: 135809926-135809929 on Build GRCh38
rs780053854 Chr.9: 135810009-135810009 on Build GRCh38
rs1005384599 Chr.9: 135809844-135809844 on Build GRCh38
rs894282478 Chr.9: 135809864-135809864 on Build GRCh38
rs989410110 Chr.9: 135809760-135809760 on Build GRCh38
rs534615370 Chr.9: 135809726-135809726 on Build GRCh38
rs971113121 Chr.9: 135809674-135809674 on Build GRCh38
rs964698122 Chr.9: 135809757-135809757 on Build GRCh38
rs1029535798 Chr.9: 135809727-135809727 on Build GRCh38
rs866197328 Chr.9: 135809872-135809872 on Build GRCh38
rs1005184062 Chr.9: 135809673-135809673 on Build GRCh38
rs370116535 Chr.9: 135809713-135809714 on Build GRCh38
rs1003840298 Chr.9: 135809591-135809591 on Build GRCh38
rs989702839 Chr.9: 135809957-135809958 on Build GRCh38

View Full Product Details