Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136665328-136665826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL7
Entrez Gene ID
51162
Gene Name
EGF like domain multiple 7
Gene Aliases
NEU1, VE-STATIN, ZNEU1
UniGene
Hs.91481
Gene Chromosome Location
Chr.9: 136654753-136672678 on Build GRCh38
Transcripts
NM_016215.4 NM_201446.2 NR_045110.1 NR_045111.1 NR_046367.1 XM_006717141.3 XM_011518764.1 XM_011518765.1 XM_011518766.1 XM_011518767.1 XM_011518768.1 XM_017014795.1 XM_017014796.1 XM_017014797.1 XM_017014798.1 XM_017014799.1

SNP Detail

SNP ID Chromosome Location
rs951390029 Chr.9: 136665487-136665487 on Build GRCh38
rs1045894655 Chr.9: 136665648-136665648 on Build GRCh38
rs954700141 Chr.9: 136665489-136665489 on Build GRCh38
rs552305608 Chr.9: 136665591-136665591 on Build GRCh38
rs563325166 Chr.9: 136665550-136665550 on Build GRCh38
rs894089024 Chr.9: 136665696-136665696 on Build GRCh38
rs1029705914 Chr.9: 136665474-136665474 on Build GRCh38
rs147182274 Chr.9: 136665453-136665453 on Build GRCh38
rs145298862 Chr.9: 136665400-136665400 on Build GRCh38
rs934351374 Chr.9: 136665652-136665652 on Build GRCh38
rs1016877078 Chr.9: 136665778-136665778 on Build GRCh38
rs1011151677 Chr.9: 136665708-136665708 on Build GRCh38
rs1052697348 Chr.9: 136665419-136665419 on Build GRCh38
rs781484013 Chr.9: 136665423-136665423 on Build GRCh38
rs891602682 Chr.9: 136665730-136665730 on Build GRCh38
rs899999301 Chr.9: 136665743-136665743 on Build GRCh38
rs559246130 Chr.9: 136665505-136665505 on Build GRCh38
rs1004192361 Chr.9: 136665771-136665771 on Build GRCh38
rs1010497741 Chr.9: 136665736-136665736 on Build GRCh38
rs998330012 Chr.9: 136665750-136665750 on Build GRCh38
rs1030192516 Chr.9: 136665757-136665757 on Build GRCh38
rs939026198 Chr.9: 136665361-136665361 on Build GRCh38
rs536403423 Chr.9: 136665375-136665375 on Build GRCh38
rs1039930522 Chr.9: 136665740-136665740 on Build GRCh38
rs370806059 Chr.9: 136665382-136665382 on Build GRCh38
rs577701171 Chr.9: 136665525-136665525 on Build GRCh38
rs949825324 Chr.9: 136665374-136665374 on Build GRCh38
rs991833273 Chr.9: 136665792-136665792 on Build GRCh38
rs559266949 Chr.9: 136665645-136665645 on Build GRCh38
rs924351853 Chr.9: 136665649-136665649 on Build GRCh38
rs1008915606 Chr.9: 136665495-136665495 on Build GRCh38
rs991365527 Chr.9: 136665604-136665604 on Build GRCh38
rs185916526 Chr.9: 136665629-136665629 on Build GRCh38
rs989302004 Chr.9: 136665520-136665520 on Build GRCh38
rs543449191 Chr.9: 136665549-136665549 on Build GRCh38
rs962657111 Chr.9: 136665779-136665779 on Build GRCh38
rs368256737 Chr.9: 136665349-136665349 on Build GRCh38
rs1026820400 Chr.9: 136665467-136665467 on Build GRCh38
rs1017509474 Chr.9: 136665500-136665500 on Build GRCh38
rs1013019877 Chr.9: 136665401-136665401 on Build GRCh38
rs899237242 Chr.9: 136665436-136665436 on Build GRCh38
rs964650075 Chr.9: 136665509-136665509 on Build GRCh38
rs949891554 Chr.9: 136665647-136665647 on Build GRCh38
rs993634900 Chr.9: 136665447-136665447 on Build GRCh38
rs1022164392 Chr.9: 136665521-136665521 on Build GRCh38
rs951463182 Chr.9: 136665769-136665769 on Build GRCh38
rs962584655 Chr.9: 136665544-136665544 on Build GRCh38
rs7847142 Chr.9: 136665688-136665688 on Build GRCh38
rs998692698 Chr.9: 136665469-136665469 on Build GRCh38

View Full Product Details