Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137047967-137048463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

SNP Detail

SNP ID Chromosome Location
rs888381980 Chr.9: 137048023-137048023 on Build GRCh38
rs950677620 Chr.9: 137048268-137048269 on Build GRCh38
rs998149638 Chr.9: 137048275-137048275 on Build GRCh38
rs1029984026 Chr.9: 137048408-137048408 on Build GRCh38
rs916165763 Chr.9: 137048350-137048350 on Build GRCh38
rs762463629 Chr.9: 137048074-137048074 on Build GRCh38
rs867612830 Chr.9: 137048017-137048017 on Build GRCh38
rs567331978 Chr.9: 137048193-137048193 on Build GRCh38
rs1029590432 Chr.9: 137048283-137048283 on Build GRCh38
rs929619860 Chr.9: 137048368-137048368 on Build GRCh38
rs983801440 Chr.9: 137048269-137048271 on Build GRCh38
rs35674457 Chr.9: 137048416-137048416 on Build GRCh38
rs62584466 Chr.9: 137048360-137048360 on Build GRCh38
rs991783849 Chr.9: 137048154-137048154 on Build GRCh38
rs1005793084 Chr.9: 137048226-137048226 on Build GRCh38
rs1033497816 Chr.9: 137048293-137048293 on Build GRCh38
rs927704251 Chr.9: 137048198-137048198 on Build GRCh38
rs888316293 Chr.9: 137048224-137048224 on Build GRCh38
rs998140214 Chr.9: 137048083-137048083 on Build GRCh38
rs367550311 Chr.9: 137047992-137047992 on Build GRCh38
rs909622276 Chr.9: 137048389-137048389 on Build GRCh38
rs77892574 Chr.9: 137048124-137048124 on Build GRCh38
rs371788098 Chr.9: 137048274-137048274 on Build GRCh38
rs923158549 Chr.9: 137048267-137048267 on Build GRCh38
rs756522474 Chr.9: 137048353-137048353 on Build GRCh38
rs552414762 Chr.9: 137048027-137048027 on Build GRCh38
rs967403175 Chr.9: 137048106-137048106 on Build GRCh38
rs964511684 Chr.9: 137048384-137048384 on Build GRCh38
rs957594850 Chr.9: 137048300-137048300 on Build GRCh38
rs1029705007 Chr.9: 137048101-137048101 on Build GRCh38
rs964313599 Chr.9: 137048183-137048183 on Build GRCh38
rs941207426 Chr.9: 137048434-137048434 on Build GRCh38
rs978007019 Chr.9: 137048112-137048112 on Build GRCh38
rs971604053 Chr.9: 137048351-137048351 on Build GRCh38
rs1025611347 Chr.9: 137048081-137048081 on Build GRCh38
rs957751397 Chr.9: 137048145-137048145 on Build GRCh38
rs1046846290 Chr.9: 137048386-137048386 on Build GRCh38
rs1055207298 Chr.9: 137048290-137048290 on Build GRCh38
rs1047235936 Chr.9: 137048202-137048202 on Build GRCh38
rs116945158 Chr.9: 137048292-137048292 on Build GRCh38
rs992062324 Chr.9: 137048342-137048342 on Build GRCh38
rs570914688 Chr.9: 137048042-137048042 on Build GRCh38
rs929711042 Chr.9: 137048199-137048199 on Build GRCh38
rs868686008 Chr.9: 137048329-137048329 on Build GRCh38
rs529942630 Chr.9: 137048238-137048238 on Build GRCh38
rs958225309 Chr.9: 137048142-137048142 on Build GRCh38
rs1005491964 Chr.9: 137048028-137048028 on Build GRCh38
rs947739254 Chr.9: 137048168-137048168 on Build GRCh38
rs189977498 Chr.9: 137048290-137048290 on Build GRCh38
rs376843719 Chr.9: 137048031-137048031 on Build GRCh38
rs1046056010 Chr.9: 137048183-137048183 on Build GRCh38
rs373027087 Chr.9: 137048167-137048167 on Build GRCh38

View Full Product Details