Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137048339-137048834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654285 c.1464C>T p.S488S Chr.9: 137048681-137048681 on Build GRCh38
6654286 c.1401C>G p.L467L Chr.9: 137048744-137048744 on Build GRCh38
3763808 c.1485T>C p.I495I Chr.9: 137048660-137048660 on Build GRCh38
4958369 c.1416C>T p.A472A Chr.9: 137048729-137048729 on Build GRCh38
6654287 c.1460A>G p.H487R Chr.9: 137048685-137048685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17853460 Chr.9: 137048660-137048660 on Build GRCh38
rs984644896 Chr.9: 137048594-137048594 on Build GRCh38
rs751443222 Chr.9: 137048797-137048797 on Build GRCh38
rs1023109047 Chr.9: 137048647-137048647 on Build GRCh38
rs774712962 Chr.9: 137048685-137048686 on Build GRCh38
rs975235613 Chr.9: 137048716-137048716 on Build GRCh38
rs951121034 Chr.9: 137048673-137048673 on Build GRCh38
rs933812175 Chr.9: 137048737-137048737 on Build GRCh38
rs775094455 Chr.9: 137048787-137048787 on Build GRCh38
rs377553411 Chr.9: 137048635-137048635 on Build GRCh38
rs1029984026 Chr.9: 137048408-137048408 on Build GRCh38
rs544581062 Chr.9: 137048661-137048661 on Build GRCh38
rs752368424 Chr.9: 137048708-137048708 on Build GRCh38
rs768928494 Chr.9: 137048671-137048671 on Build GRCh38
rs759104050 Chr.9: 137048706-137048706 on Build GRCh38
rs772827838 Chr.9: 137048787-137048805 on Build GRCh38
rs929619860 Chr.9: 137048368-137048368 on Build GRCh38
rs759159007 Chr.9: 137048773-137048773 on Build GRCh38
rs750327305 Chr.9: 137048813-137048813 on Build GRCh38
rs1033136887 Chr.9: 137048518-137048518 on Build GRCh38
rs35674457 Chr.9: 137048416-137048416 on Build GRCh38
rs747269810 Chr.9: 137048750-137048750 on Build GRCh38
rs749836204 Chr.9: 137048607-137048608 on Build GRCh38
rs903096853 Chr.9: 137048798-137048798 on Build GRCh38
rs971656444 Chr.9: 137048649-137048649 on Build GRCh38
rs139168222 Chr.9: 137048707-137048707 on Build GRCh38
rs777256965 Chr.9: 137048769-137048769 on Build GRCh38
rs772134556 Chr.9: 137048748-137048748 on Build GRCh38
rs948518481 Chr.9: 137048616-137048616 on Build GRCh38
rs934680226 Chr.9: 137048807-137048807 on Build GRCh38
rs999253756 Chr.9: 137048516-137048516 on Build GRCh38
rs990652437 Chr.9: 137048531-137048532 on Build GRCh38
rs763590098 Chr.9: 137048795-137048795 on Build GRCh38
rs771527565 Chr.9: 137048762-137048762 on Build GRCh38
rs971871756 Chr.9: 137048564-137048564 on Build GRCh38
rs1039862528 Chr.9: 137048738-137048738 on Build GRCh38
rs899774753 Chr.9: 137048449-137048449 on Build GRCh38
rs62584466 Chr.9: 137048360-137048360 on Build GRCh38
rs141498031 Chr.9: 137048711-137048711 on Build GRCh38
rs573033077 Chr.9: 137048609-137048609 on Build GRCh38
rs893169666 Chr.9: 137048523-137048523 on Build GRCh38
rs761496670 Chr.9: 137048808-137048808 on Build GRCh38
rs930349431 Chr.9: 137048574-137048574 on Build GRCh38
rs909622276 Chr.9: 137048389-137048389 on Build GRCh38
rs998036008 Chr.9: 137048459-137048459 on Build GRCh38
rs574901335 Chr.9: 137048704-137048704 on Build GRCh38
rs532522458 Chr.9: 137048730-137048730 on Build GRCh38
rs761265539 Chr.9: 137048701-137048701 on Build GRCh38
rs373902676 Chr.9: 137048626-137048626 on Build GRCh38
rs915847287 Chr.9: 137048607-137048607 on Build GRCh38
rs200372025 Chr.9: 137048682-137048682 on Build GRCh38
rs146486854 Chr.9: 137048699-137048699 on Build GRCh38
rs916642318 Chr.9: 137048535-137048535 on Build GRCh38
rs762099217 Chr.9: 137048746-137048747 on Build GRCh38
rs964511684 Chr.9: 137048384-137048384 on Build GRCh38
rs941207426 Chr.9: 137048434-137048434 on Build GRCh38
rs768996770 Chr.9: 137048629-137048629 on Build GRCh38
rs903220385 Chr.9: 137048485-137048485 on Build GRCh38
rs773406935 Chr.9: 137048700-137048700 on Build GRCh38
rs954163767 Chr.9: 137048541-137048541 on Build GRCh38
rs866701012 Chr.9: 137048553-137048553 on Build GRCh38
rs751258047 Chr.9: 137048715-137048715 on Build GRCh38
rs1046846290 Chr.9: 137048386-137048386 on Build GRCh38
rs757227399 Chr.9: 137048725-137048725 on Build GRCh38
rs1045549843 Chr.9: 137048622-137048622 on Build GRCh38
rs1050950119 Chr.9: 137048467-137048467 on Build GRCh38
rs762711133 Chr.9: 137048789-137048789 on Build GRCh38
rs748558678 Chr.9: 137048694-137048694 on Build GRCh38
rs966193661 Chr.9: 137048562-137048562 on Build GRCh38
rs986981863 Chr.9: 137048545-137048545 on Build GRCh38
rs918915396 Chr.9: 137048567-137048567 on Build GRCh38
rs552499259 Chr.9: 137048749-137048749 on Build GRCh38
rs112381056 Chr.9: 137048492-137048492 on Build GRCh38
rs747001240 Chr.9: 137048654-137048654 on Build GRCh38
rs753972936 Chr.9: 137048731-137048731 on Build GRCh38
rs779143753 Chr.9: 137048739-137048739 on Build GRCh38
rs763975453 Chr.9: 137048714-137048714 on Build GRCh38
rs777613806 Chr.9: 137048651-137048651 on Build GRCh38
rs921119451 Chr.9: 137048726-137048726 on Build GRCh38
rs755025538 Chr.9: 137048732-137048732 on Build GRCh38
rs983364605 Chr.9: 137048472-137048472 on Build GRCh38
rs748399268 Chr.9: 137048746-137048746 on Build GRCh38
rs769461523 Chr.9: 137048779-137048779 on Build GRCh38
rs1039479087 Chr.9: 137048445-137048445 on Build GRCh38
rs774539211 Chr.9: 137048691-137048691 on Build GRCh38
rs767224684 Chr.9: 137048767-137048773 on Build GRCh38

View Full Product Details