Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137048805-137049318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5345752 c.1298C>G p.A433G Chr.9: 137048847-137048847 on Build GRCh38
3763809 c.1248C>T p.D416D Chr.9: 137048977-137048977 on Build GRCh38
5658579 c.1268T>C p.V423A Chr.9: 137048957-137048957 on Build GRCh38
1461303 c.1298C>T p.A433V Chr.9: 137048847-137048847 on Build GRCh38
5977163 c.1179C>T p.A393A Chr.9: 137049046-137049046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778301758 Chr.9: 137049092-137049092 on Build GRCh38
rs762361439 Chr.9: 137048856-137048856 on Build GRCh38
rs755447073 Chr.9: 137049232-137049232 on Build GRCh38
rs750143962 Chr.9: 137049133-137049133 on Build GRCh38
rs554966421 Chr.9: 137049146-137049146 on Build GRCh38
rs764416173 Chr.9: 137048944-137048944 on Build GRCh38
rs898859191 Chr.9: 137048957-137048957 on Build GRCh38
rs180778128 Chr.9: 137048968-137048968 on Build GRCh38
rs550570851 Chr.9: 137048890-137048890 on Build GRCh38
rs1003349742 Chr.9: 137049241-137049241 on Build GRCh38
rs948706107 Chr.9: 137049163-137049163 on Build GRCh38
rs1008415710 Chr.9: 137049256-137049256 on Build GRCh38
rs777952982 Chr.9: 137049185-137049185 on Build GRCh38
rs756528638 Chr.9: 137049087-137049087 on Build GRCh38
rs746397360 Chr.9: 137048831-137048831 on Build GRCh38
rs768023893 Chr.9: 137048979-137048979 on Build GRCh38
rs1048332609 Chr.9: 137049041-137049041 on Build GRCh38
rs115672624 Chr.9: 137049253-137049253 on Build GRCh38
rs373768869 Chr.9: 137048848-137048848 on Build GRCh38
rs45457700 Chr.9: 137049085-137049085 on Build GRCh38
rs377260878 Chr.9: 137048965-137048965 on Build GRCh38
rs751597294 Chr.9: 137049065-137049065 on Build GRCh38
rs914494484 Chr.9: 137049162-137049162 on Build GRCh38
rs777391003 Chr.9: 137049107-137049107 on Build GRCh38
rs7873815 Chr.9: 137048977-137048977 on Build GRCh38
rs761967508 Chr.9: 137049191-137049191 on Build GRCh38
rs773875166 Chr.9: 137048980-137048980 on Build GRCh38
rs567115924 Chr.9: 137048935-137048935 on Build GRCh38
rs200961887 Chr.9: 137048861-137048861 on Build GRCh38
rs1038398966 Chr.9: 137049233-137049233 on Build GRCh38
rs975822382 Chr.9: 137049291-137049291 on Build GRCh38
rs956107235 Chr.9: 137049292-137049292 on Build GRCh38
rs754953637 Chr.9: 137048914-137048923 on Build GRCh38
rs971008885 Chr.9: 137048863-137048863 on Build GRCh38
rs1010944194 Chr.9: 137049280-137049280 on Build GRCh38
rs996475885 Chr.9: 137049239-137049239 on Build GRCh38
rs775141465 Chr.9: 137048841-137048841 on Build GRCh38
rs747780229 Chr.9: 137048991-137048991 on Build GRCh38
rs1039956992 Chr.9: 137048914-137048915 on Build GRCh38
rs778810351 Chr.9: 137049043-137049043 on Build GRCh38
rs963978860 Chr.9: 137049290-137049290 on Build GRCh38
rs1026572540 Chr.9: 137048896-137048896 on Build GRCh38
rs771780022 Chr.9: 137049106-137049106 on Build GRCh38
rs779471409 Chr.9: 137049234-137049234 on Build GRCh38
rs886188261 Chr.9: 137048940-137048940 on Build GRCh38
rs371094769 Chr.9: 137049050-137049050 on Build GRCh38
rs989553593 Chr.9: 137049039-137049039 on Build GRCh38
rs569953168 Chr.9: 137049204-137049204 on Build GRCh38
rs1031060996 Chr.9: 137049018-137049018 on Build GRCh38
rs752603291 Chr.9: 137049048-137049048 on Build GRCh38
rs1003052514 Chr.9: 137049029-137049029 on Build GRCh38
rs373169384 Chr.9: 137048936-137048936 on Build GRCh38
rs767250469 Chr.9: 137048868-137048868 on Build GRCh38
rs924557476 Chr.9: 137048960-137048960 on Build GRCh38
rs7390981 Chr.9: 137049097-137049097 on Build GRCh38
rs774359077 Chr.9: 137049171-137049171 on Build GRCh38
rs555837268 Chr.9: 137049252-137049252 on Build GRCh38
rs765943048 Chr.9: 137048895-137048918 on Build GRCh38
rs536251205 Chr.9: 137049211-137049211 on Build GRCh38
rs553845490 Chr.9: 137049214-137049214 on Build GRCh38
rs771657346 Chr.9: 137049007-137049007 on Build GRCh38
rs766679372 Chr.9: 137049215-137049215 on Build GRCh38
rs755466572 Chr.9: 137049090-137049090 on Build GRCh38
rs772945073 Chr.9: 137048876-137048876 on Build GRCh38
rs1003041237 Chr.9: 137048870-137048870 on Build GRCh38
rs560861771 Chr.9: 137048920-137048930 on Build GRCh38
rs368497640 Chr.9: 137049132-137049132 on Build GRCh38
rs753237383 Chr.9: 137048914-137048918 on Build GRCh38
rs749794737 Chr.9: 137049089-137049089 on Build GRCh38
rs989961742 Chr.9: 137049296-137049296 on Build GRCh38
rs907248514 Chr.9: 137049028-137049028 on Build GRCh38
rs756748898 Chr.9: 137048976-137048976 on Build GRCh38
rs540334692 Chr.9: 137049209-137049209 on Build GRCh38
rs374463317 Chr.9: 137049046-137049046 on Build GRCh38
rs901337184 Chr.9: 137048924-137048924 on Build GRCh38
rs1016988756 Chr.9: 137048908-137048908 on Build GRCh38
rs1051489198 Chr.9: 137048937-137048937 on Build GRCh38
rs372478178 Chr.9: 137049283-137049283 on Build GRCh38
rs759404134 Chr.9: 137049166-137049166 on Build GRCh38
rs900723236 Chr.9: 137049236-137049236 on Build GRCh38
rs950999696 Chr.9: 137048900-137048900 on Build GRCh38
rs955088455 Chr.9: 137049122-137049122 on Build GRCh38
rs868390289 Chr.9: 137049131-137049131 on Build GRCh38
rs751759312 Chr.9: 137048953-137048953 on Build GRCh38
rs770743751 Chr.9: 137049145-137049145 on Build GRCh38
rs1028032104 Chr.9: 137049115-137049115 on Build GRCh38
rs746846153 Chr.9: 137049108-137049108 on Build GRCh38
rs564511306 Chr.9: 137048894-137048912 on Build GRCh38
rs962388903 Chr.9: 137048927-137048927 on Build GRCh38
rs1044951420 Chr.9: 137049221-137049221 on Build GRCh38
rs189421012 Chr.9: 137049271-137049271 on Build GRCh38
rs758258946 Chr.9: 137049125-137049125 on Build GRCh38
rs766353234 Chr.9: 137049036-137049036 on Build GRCh38
rs758385044 Chr.9: 137048943-137048943 on Build GRCh38
rs746866556 Chr.9: 137049240-137049240 on Build GRCh38
rs938725160 Chr.9: 137049184-137049184 on Build GRCh38
rs924415207 Chr.9: 137049143-137049143 on Build GRCh38
rs558929657 Chr.9: 137049017-137049017 on Build GRCh38
rs768417706 Chr.9: 137048858-137048858 on Build GRCh38
rs746165675 Chr.9: 137048973-137048973 on Build GRCh38
rs777223526 Chr.9: 137049284-137049284 on Build GRCh38
rs990508783 Chr.9: 137049152-137049152 on Build GRCh38
rs1037477877 Chr.9: 137049201-137049201 on Build GRCh38
rs117388172 Chr.9: 137049222-137049222 on Build GRCh38
rs757422958 Chr.9: 137049084-137049084 on Build GRCh38
rs975682620 Chr.9: 137049095-137049095 on Build GRCh38
rs982482349 Chr.9: 137048905-137048905 on Build GRCh38
rs760459410 Chr.9: 137049033-137049033 on Build GRCh38
rs776632994 Chr.9: 137049263-137049263 on Build GRCh38
rs765013535 Chr.9: 137049027-137049049 on Build GRCh38
rs1004525038 Chr.9: 137048941-137048943 on Build GRCh38
rs1044540301 Chr.9: 137048999-137048999 on Build GRCh38
rs781618182 Chr.9: 137049149-137049150 on Build GRCh38
rs780751115 Chr.9: 137049088-137049088 on Build GRCh38
rs760620897 Chr.9: 137048907-137048907 on Build GRCh38
rs781654598 Chr.9: 137048830-137048830 on Build GRCh38
rs746684639 Chr.9: 137049286-137049286 on Build GRCh38
rs566715987 Chr.9: 137048970-137048970 on Build GRCh38

View Full Product Details