Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127506021-127506515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129B
Entrez Gene ID
64855
Gene Name
family with sequence similarity 129 member B
Gene Aliases
C9orf88, MEG-3, MINERVA, OC58, bA356B19.6
UniGene
Hs.522401
Gene Chromosome Location
Chr.9: 127505338-127579007 on Build GRCh38
Transcripts
NM_001035534.2 NM_022833.3 XM_005252135.2 XM_011518925.1

SNP Detail

SNP ID Chromosome Location
rs999116954 Chr.9: 127506289-127506289 on Build GRCh38
rs771010784 Chr.9: 127506370-127506370 on Build GRCh38
rs949776843 Chr.9: 127506106-127506106 on Build GRCh38
rs915972621 Chr.9: 127506249-127506249 on Build GRCh38
rs959208758 Chr.9: 127506221-127506221 on Build GRCh38
rs771931501 Chr.9: 127506288-127506288 on Build GRCh38
rs200329460 Chr.9: 127506152-127506152 on Build GRCh38
rs1017326346 Chr.9: 127506366-127506366 on Build GRCh38
rs1033140197 Chr.9: 127506103-127506103 on Build GRCh38
rs760587895 Chr.9: 127506360-127506360 on Build GRCh38
rs768244811 Chr.9: 127506046-127506046 on Build GRCh38
rs550276230 Chr.9: 127506060-127506060 on Build GRCh38
rs528641367 Chr.9: 127506322-127506322 on Build GRCh38
rs574426303 Chr.9: 127506200-127506200 on Build GRCh38
rs1055868397 Chr.9: 127506241-127506241 on Build GRCh38
rs543238980 Chr.9: 127506217-127506217 on Build GRCh38
rs899713352 Chr.9: 127506144-127506144 on Build GRCh38
rs992026136 Chr.9: 127506058-127506058 on Build GRCh38
rs903439180 Chr.9: 127506285-127506285 on Build GRCh38
rs199659552 Chr.9: 127506132-127506132 on Build GRCh38
rs11545940 Chr.9: 127506077-127506077 on Build GRCh38
rs908191193 Chr.9: 127506128-127506128 on Build GRCh38
rs772090950 Chr.9: 127506279-127506279 on Build GRCh38
rs560932301 Chr.9: 127506067-127506067 on Build GRCh38
rs975696873 Chr.9: 127506201-127506201 on Build GRCh38
rs199894907 Chr.9: 127506492-127506494 on Build GRCh38
rs11545936 Chr.9: 127506383-127506383 on Build GRCh38
rs1025933878 Chr.9: 127506406-127506406 on Build GRCh38
rs985069245 Chr.9: 127506210-127506210 on Build GRCh38
rs1046794470 Chr.9: 127506116-127506116 on Build GRCh38
rs148927499 Chr.9: 127506137-127506138 on Build GRCh38
rs996694997 Chr.9: 127506162-127506162 on Build GRCh38
rs941045749 Chr.9: 127506417-127506417 on Build GRCh38
rs1007300571 Chr.9: 127506159-127506159 on Build GRCh38
rs950726252 Chr.9: 127506424-127506424 on Build GRCh38
rs1014540511 Chr.9: 127506054-127506054 on Build GRCh38
rs375950327 Chr.9: 127506458-127506458 on Build GRCh38
rs967362511 Chr.9: 127506214-127506214 on Build GRCh38
rs190077457 Chr.9: 127506262-127506262 on Build GRCh38
rs990621540 Chr.9: 127506490-127506490 on Build GRCh38
rs1055068692 Chr.9: 127506301-127506301 on Build GRCh38
rs894468653 Chr.9: 127506050-127506050 on Build GRCh38
rs957386078 Chr.9: 127506124-127506124 on Build GRCh38
rs991708769 Chr.9: 127506260-127506260 on Build GRCh38
rs56147938 Chr.9: 127506151-127506152 on Build GRCh38
rs71495646 Chr.9: 127506136-127506137 on Build GRCh38
rs950042760 Chr.9: 127506264-127506264 on Build GRCh38
rs773147717 Chr.9: 127506331-127506331 on Build GRCh38
rs563264293 Chr.9: 127506204-127506204 on Build GRCh38
rs548295484 Chr.9: 127506425-127506425 on Build GRCh38
rs553629852 Chr.9: 127506384-127506384 on Build GRCh38
rs909061371 Chr.9: 127506449-127506449 on Build GRCh38
rs917411324 Chr.9: 127506296-127506296 on Build GRCh38
rs940983690 Chr.9: 127506137-127506140 on Build GRCh38
rs532886579 Chr.9: 127506205-127506205 on Build GRCh38
rs1000569084 Chr.9: 127506216-127506216 on Build GRCh38
rs566293978 Chr.9: 127506269-127506269 on Build GRCh38
rs941470458 Chr.9: 127506223-127506223 on Build GRCh38
rs58280857 Chr.9: 127506076-127506076 on Build GRCh38
rs921231857 Chr.9: 127506441-127506441 on Build GRCh38
rs557646586 Chr.9: 127506198-127506198 on Build GRCh38
rs965739147 Chr.9: 127506371-127506371 on Build GRCh38
rs1017251524 Chr.9: 127506180-127506180 on Build GRCh38
rs932547796 Chr.9: 127506482-127506482 on Build GRCh38
rs3831357 Chr.9: 127506148-127506149 on Build GRCh38

View Full Product Details