Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127509897-127510390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129B
Entrez Gene ID
64855
Gene Name
family with sequence similarity 129 member B
Gene Aliases
C9orf88, MEG-3, MINERVA, OC58, bA356B19.6
UniGene
Hs.522401
Gene Chromosome Location
Chr.9: 127505338-127579007 on Build GRCh38
Transcripts
NM_001035534.2 NM_022833.3 XM_005252135.2 XM_011518925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530709 c.940A>G p.I314V Chr.9: 127510328-127510328 on Build GRCh38
3904226 c.1140C>T p.G380G Chr.9: 127510167-127510167 on Build GRCh38
150720 c.1161+8C>T p.? Chr.9: 127510138-127510138 on Build GRCh38
3904228 c.1021C>T p.P341S Chr.9: 127510286-127510286 on Build GRCh38
5951385 c.1119C>T p.G373G Chr.9: 127510149-127510149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754799436 Chr.9: 127510064-127510064 on Build GRCh38
rs937003636 Chr.9: 127510028-127510028 on Build GRCh38
rs779884675 Chr.9: 127510364-127510364 on Build GRCh38
rs749677034 Chr.9: 127510097-127510097 on Build GRCh38
rs542373191 Chr.9: 127510370-127510370 on Build GRCh38
rs145531660 Chr.9: 127510196-127510196 on Build GRCh38
rs937998338 Chr.9: 127509927-127509927 on Build GRCh38
rs771826687 Chr.9: 127510340-127510343 on Build GRCh38
rs762112827 Chr.9: 127510224-127510224 on Build GRCh38
rs935810225 Chr.9: 127510056-127510056 on Build GRCh38
rs765730690 Chr.9: 127510344-127510344 on Build GRCh38
rs370582135 Chr.9: 127510293-127510293 on Build GRCh38
rs901458469 Chr.9: 127510078-127510078 on Build GRCh38
rs750008275 Chr.9: 127510359-127510359 on Build GRCh38
rs992158481 Chr.9: 127509937-127509937 on Build GRCh38
rs1032327996 Chr.9: 127509943-127509943 on Build GRCh38
rs1053809602 Chr.9: 127510059-127510059 on Build GRCh38
rs1021547146 Chr.9: 127510271-127510271 on Build GRCh38
rs772398032 Chr.9: 127510322-127510322 on Build GRCh38
rs769781257 Chr.9: 127510118-127510118 on Build GRCh38
rs768713976 Chr.9: 127510199-127510199 on Build GRCh38
rs773677918 Chr.9: 127510230-127510230 on Build GRCh38
rs761615690 Chr.9: 127510337-127510337 on Build GRCh38
rs997171243 Chr.9: 127510023-127510023 on Build GRCh38
rs374128661 Chr.9: 127510300-127510300 on Build GRCh38
rs889831626 Chr.9: 127510065-127510065 on Build GRCh38
rs1019610251 Chr.9: 127510076-127510076 on Build GRCh38
rs1007304050 Chr.9: 127510072-127510072 on Build GRCh38
rs866287118 Chr.9: 127509923-127509923 on Build GRCh38
rs748275528 Chr.9: 127510319-127510319 on Build GRCh38
rs761011053 Chr.9: 127510328-127510328 on Build GRCh38
rs762129044 Chr.9: 127510137-127510137 on Build GRCh38
rs146148622 Chr.9: 127510034-127510034 on Build GRCh38
rs796472128 Chr.9: 127510355-127510356 on Build GRCh38
rs1005695759 Chr.9: 127510164-127510164 on Build GRCh38
rs992473434 Chr.9: 127510029-127510029 on Build GRCh38
rs759375297 Chr.9: 127510366-127510367 on Build GRCh38
rs774772769 Chr.9: 127510323-127510324 on Build GRCh38
rs199641739 Chr.9: 127510133-127510133 on Build GRCh38
rs745730530 Chr.9: 127510181-127510181 on Build GRCh38
rs375863565 Chr.9: 127510119-127510119 on Build GRCh38
rs759415108 Chr.9: 127510117-127510117 on Build GRCh38
rs768532882 Chr.9: 127510311-127510311 on Build GRCh38
rs1025811740 Chr.9: 127509998-127509998 on Build GRCh38
rs764202103 Chr.9: 127510126-127510126 on Build GRCh38
rs952471813 Chr.9: 127510015-127510015 on Build GRCh38
rs760219590 Chr.9: 127510355-127510356 on Build GRCh38
rs372647854 Chr.9: 127510167-127510167 on Build GRCh38
rs752123683 Chr.9: 127510151-127510151 on Build GRCh38
rs370990792 Chr.9: 127510343-127510343 on Build GRCh38
rs776876414 Chr.9: 127510330-127510330 on Build GRCh38
rs755447369 Chr.9: 127510258-127510258 on Build GRCh38
rs141849503 Chr.9: 127509954-127509954 on Build GRCh38
rs377686230 Chr.9: 127510305-127510305 on Build GRCh38
rs750871615 Chr.9: 127510148-127510148 on Build GRCh38
rs752518593 Chr.9: 127510370-127510371 on Build GRCh38
rs751132229 Chr.9: 127510260-127510260 on Build GRCh38
rs377128721 Chr.9: 127510281-127510281 on Build GRCh38
rs780863400 Chr.9: 127510279-127510279 on Build GRCh38
rs540149997 Chr.9: 127510161-127510161 on Build GRCh38
rs780953007 Chr.9: 127510178-127510178 on Build GRCh38
rs763604934 Chr.9: 127510259-127510259 on Build GRCh38
rs11788951 Chr.9: 127510138-127510138 on Build GRCh38
rs774307933 Chr.9: 127510200-127510200 on Build GRCh38
rs761168162 Chr.9: 127510246-127510246 on Build GRCh38
rs760079593 Chr.9: 127510335-127510335 on Build GRCh38
rs770676266 Chr.9: 127510113-127510113 on Build GRCh38
rs766673423 Chr.9: 127510253-127510253 on Build GRCh38
rs1010285462 Chr.9: 127510222-127510222 on Build GRCh38
rs973352196 Chr.9: 127509972-127509972 on Build GRCh38
rs181682964 Chr.9: 127510046-127510046 on Build GRCh38
rs768835547 Chr.9: 127510320-127510321 on Build GRCh38
rs755733421 Chr.9: 127510360-127510360 on Build GRCh38
rs138815572 Chr.9: 127510197-127510197 on Build GRCh38
rs927006556 Chr.9: 127510024-127510024 on Build GRCh38
rs538750234 Chr.9: 127510120-127510120 on Build GRCh38
rs983856069 Chr.9: 127510021-127510021 on Build GRCh38
rs375950908 Chr.9: 127510351-127510351 on Build GRCh38
rs889800330 Chr.9: 127509948-127509948 on Build GRCh38
rs1000892421 Chr.9: 127509941-127509941 on Build GRCh38
rs765003445 Chr.9: 127510367-127510368 on Build GRCh38
rs201066039 Chr.9: 127510352-127510352 on Build GRCh38
rs779715730 Chr.9: 127510301-127510301 on Build GRCh38
rs72767960 Chr.9: 127510040-127510040 on Build GRCh38
rs201600255 Chr.9: 127510365-127510365 on Build GRCh38
rs369376143 Chr.9: 127510323-127510323 on Build GRCh38
rs531214740 Chr.9: 127509928-127509928 on Build GRCh38
rs979894156 Chr.9: 127510047-127510047 on Build GRCh38
rs769689758 Chr.9: 127510191-127510191 on Build GRCh38
rs945975931 Chr.9: 127510038-127510038 on Build GRCh38
rs746666359 Chr.9: 127510098-127510098 on Build GRCh38
rs556385146 Chr.9: 127510079-127510079 on Build GRCh38
rs967723354 Chr.9: 127510235-127510235 on Build GRCh38
rs571150868 Chr.9: 127509968-127509968 on Build GRCh38
rs929267046 Chr.9: 127510090-127510090 on Build GRCh38
rs140423692 Chr.9: 127510166-127510166 on Build GRCh38
rs749213184 Chr.9: 127510198-127510198 on Build GRCh38
rs553619776 Chr.9: 127510276-127510276 on Build GRCh38
rs751039998 Chr.9: 127510347-127510347 on Build GRCh38
rs772886226 Chr.9: 127510348-127510348 on Build GRCh38
rs770588471 Chr.9: 127510173-127510173 on Build GRCh38
rs574506801 Chr.9: 127510304-127510304 on Build GRCh38
rs753656236 Chr.9: 127510365-127510366 on Build GRCh38
rs754333552 Chr.9: 127510257-127510257 on Build GRCh38
rs767914409 Chr.9: 127510226-127510226 on Build GRCh38
rs528022223 Chr.9: 127509969-127509969 on Build GRCh38
rs977327490 Chr.9: 127510278-127510278 on Build GRCh38
rs756681587 Chr.9: 127510149-127510149 on Build GRCh38
rs369810394 Chr.9: 127510116-127510116 on Build GRCh38
rs774480906 Chr.9: 127510318-127510318 on Build GRCh38
rs751762963 Chr.9: 127510128-127510128 on Build GRCh38

View Full Product Details