Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128835401-128835906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KYAT1
Entrez Gene ID
883
Gene Name
kynurenine aminotransferase 1
Gene Aliases
CCBL1, GTK, KAT1, KATI
UniGene
Hs.495250
Gene Chromosome Location
Chr.9: 128832942-128882551 on Build GRCh38
Transcripts
NM_001122671.1 NM_001122672.1 NM_001287390.1 NM_004059.4 NR_109829.1 XM_006717311.3 XM_011519165.2 XM_011519167.2 XM_011519168.1 XM_011519169.1 XM_011519170.2 XM_017015260.1 XM_017015261.1 XM_017015262.1 XM_017015263.1 XM_017015264.1 XM_017015265.1 XM_017015266.1 XM_017015267.1 XM_017015268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4784404 c.1266C>T p.G422G Chr.9: 128835539-128835539 on Build GRCh38
3664132 c.1120T>C p.C374R Chr.9: 128835796-128835796 on Build GRCh38
3847791 c.1003C>G p.Q335E Chr.9: 128835520-128835520 on Build GRCh38
4862768 c.1226G>A p.R409H Chr.9: 128835579-128835579 on Build GRCh38
138735 c.881G>A p.R294Q Chr.9: 128835642-128835642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752518739 Chr.9: 128835622-128835622 on Build GRCh38
rs757568453 Chr.9: 128835634-128835634 on Build GRCh38
rs753014857 Chr.9: 128835509-128835509 on Build GRCh38
rs760349031 Chr.9: 128835593-128835593 on Build GRCh38
rs200677798 Chr.9: 128835558-128835558 on Build GRCh38
rs889431792 Chr.9: 128835626-128835626 on Build GRCh38
rs755666803 Chr.9: 128835529-128835529 on Build GRCh38
rs377350447 Chr.9: 128835656-128835656 on Build GRCh38
rs767811954 Chr.9: 128835794-128835794 on Build GRCh38
rs375618315 Chr.9: 128835625-128835625 on Build GRCh38
rs1014794196 Chr.9: 128835791-128835791 on Build GRCh38
rs371772792 Chr.9: 128835838-128835838 on Build GRCh38
rs767142268 Chr.9: 128835518-128835518 on Build GRCh38
rs751383208 Chr.9: 128835789-128835789 on Build GRCh38
rs1057342951 Chr.9: 128835580-128835580 on Build GRCh38
rs762606831 Chr.9: 128835854-128835854 on Build GRCh38
rs913480048 Chr.9: 128835583-128835583 on Build GRCh38
rs946290214 Chr.9: 128835803-128835803 on Build GRCh38
rs768859816 Chr.9: 128835686-128835686 on Build GRCh38
rs747845627 Chr.9: 128835867-128835867 on Build GRCh38
rs765190007 Chr.9: 128835714-128835714 on Build GRCh38
rs779938346 Chr.9: 128835820-128835820 on Build GRCh38
rs752589649 Chr.9: 128835742-128835743 on Build GRCh38
rs771542618 Chr.9: 128835466-128835466 on Build GRCh38
rs754565435 Chr.9: 128835564-128835565 on Build GRCh38
rs1031856039 Chr.9: 128835520-128835520 on Build GRCh38
rs370680048 Chr.9: 128835624-128835624 on Build GRCh38
rs776109397 Chr.9: 128835702-128835702 on Build GRCh38
rs781352323 Chr.9: 128835642-128835642 on Build GRCh38
rs370304994 Chr.9: 128835757-128835757 on Build GRCh38
rs546763740 Chr.9: 128835432-128835432 on Build GRCh38
rs772390765 Chr.9: 128835841-128835841 on Build GRCh38
rs925525623 Chr.9: 128835856-128835856 on Build GRCh38
rs368898642 Chr.9: 128835748-128835748 on Build GRCh38
rs770551888 Chr.9: 128835764-128835764 on Build GRCh38
rs748347564 Chr.9: 128835450-128835450 on Build GRCh38
rs148851411 Chr.9: 128835881-128835881 on Build GRCh38
rs778226332 Chr.9: 128835842-128835842 on Build GRCh38
rs761873399 Chr.9: 128835885-128835885 on Build GRCh38
rs768476222 Chr.9: 128835444-128835444 on Build GRCh38
rs767172486 Chr.9: 128835586-128835586 on Build GRCh38
rs765590390 Chr.9: 128835508-128835508 on Build GRCh38
rs992207048 Chr.9: 128835810-128835810 on Build GRCh38
rs1000604085 Chr.9: 128835617-128835617 on Build GRCh38
rs760153693 Chr.9: 128835506-128835506 on Build GRCh38
rs948268293 Chr.9: 128835693-128835693 on Build GRCh38
rs374406437 Chr.9: 128835594-128835594 on Build GRCh38
rs904036599 Chr.9: 128835469-128835469 on Build GRCh38
rs376312507 Chr.9: 128835554-128835554 on Build GRCh38
rs373497046 Chr.9: 128835573-128835573 on Build GRCh38
rs764789925 Chr.9: 128835742-128835742 on Build GRCh38
rs201434056 Chr.9: 128835524-128835524 on Build GRCh38
rs778706625 Chr.9: 128835548-128835548 on Build GRCh38
rs780333135 Chr.9: 128835805-128835805 on Build GRCh38
rs370088270 Chr.9: 128835590-128835590 on Build GRCh38
rs369761548 Chr.9: 128835643-128835643 on Build GRCh38
rs761461352 Chr.9: 128835517-128835517 on Build GRCh38
rs374767996 Chr.9: 128835762-128835762 on Build GRCh38
rs369251259 Chr.9: 128835445-128835445 on Build GRCh38
rs764604643 Chr.9: 128835779-128835779 on Build GRCh38
rs769794749 Chr.9: 128835745-128835745 on Build GRCh38
rs144446586 Chr.9: 128835715-128835715 on Build GRCh38
rs759021492 Chr.9: 128835713-128835713 on Build GRCh38
rs751403330 Chr.9: 128835741-128835741 on Build GRCh38
rs557793663 Chr.9: 128835615-128835615 on Build GRCh38
rs374471786 Chr.9: 128835716-128835716 on Build GRCh38
rs749938698 Chr.9: 128835422-128835422 on Build GRCh38
rs564161998 Chr.9: 128835851-128835851 on Build GRCh38
rs754887935 Chr.9: 128835547-128835547 on Build GRCh38
rs745768091 Chr.9: 128835700-128835700 on Build GRCh38
rs775081171 Chr.9: 128835787-128835787 on Build GRCh38
rs771096858 Chr.9: 128835559-128835559 on Build GRCh38
rs774764176 Chr.9: 128835781-128835781 on Build GRCh38
rs368009186 Chr.9: 128835793-128835793 on Build GRCh38
rs754018813 Chr.9: 128835821-128835821 on Build GRCh38
rs146595012 Chr.9: 128835728-128835728 on Build GRCh38
rs750569862 Chr.9: 128835806-128835806 on Build GRCh38
rs375126495 Chr.9: 128835847-128835847 on Build GRCh38
rs375187999 Chr.9: 128835431-128835431 on Build GRCh38
rs745938044 Chr.9: 128835747-128835747 on Build GRCh38
rs202219883 Chr.9: 128835788-128835788 on Build GRCh38
rs183808885 Chr.9: 128835655-128835655 on Build GRCh38
rs774003247 Chr.9: 128835539-128835539 on Build GRCh38
rs529498855 Chr.9: 128835465-128835465 on Build GRCh38
rs770205049 Chr.9: 128835574-128835574 on Build GRCh38
rs777318537 Chr.9: 128835472-128835472 on Build GRCh38
rs755466817 Chr.9: 128835825-128835825 on Build GRCh38
rs891529483 Chr.9: 128835718-128835718 on Build GRCh38
rs774696475 Chr.9: 128835761-128835761 on Build GRCh38
rs946351344 Chr.9: 128835584-128835584 on Build GRCh38
rs758288939 Chr.9: 128835806-128835813 on Build GRCh38
rs747964382 Chr.9: 128835552-128835552 on Build GRCh38
rs776875030 Chr.9: 128835565-128835565 on Build GRCh38
rs371258028 Chr.9: 128835753-128835753 on Build GRCh38
rs754941481 Chr.9: 128835601-128835601 on Build GRCh38
rs760256993 Chr.9: 128835696-128835696 on Build GRCh38
rs753733096 Chr.9: 128835542-128835542 on Build GRCh38
rs759264765 Chr.9: 128835778-128835778 on Build GRCh38
rs779445893 Chr.9: 128835826-128835826 on Build GRCh38
rs774717134 Chr.9: 128835688-128835688 on Build GRCh38
rs372740953 Chr.9: 128835660-128835660 on Build GRCh38
rs769932177 Chr.9: 128835756-128835756 on Build GRCh38
rs999919100 Chr.9: 128835477-128835477 on Build GRCh38
rs774466852 Chr.9: 128835882-128835882 on Build GRCh38
rs377480189 Chr.9: 128835599-128835599 on Build GRCh38
rs766089431 Chr.9: 128835701-128835701 on Build GRCh38
rs757547653 Chr.9: 128835744-128835744 on Build GRCh38
rs773340033 Chr.9: 128835451-128835451 on Build GRCh38
rs369630666 Chr.9: 128835848-128835848 on Build GRCh38
rs575651890 Chr.9: 128835817-128835817 on Build GRCh38
rs139758426 Chr.9: 128835579-128835579 on Build GRCh38
rs768852737 Chr.9: 128835770-128835770 on Build GRCh38
rs199968196 Chr.9: 128835758-128835758 on Build GRCh38

View Full Product Details