Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129322147-129322652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf106
Entrez Gene ID
414318
Gene Name
chromosome 9 open reading frame 106
Gene Aliases
bA65J3.5
UniGene
Hs.449434
Gene Chromosome Location
Chr.9: 129321016-129322603 on Build GRCh38
Transcripts
NM_001012715.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3375117 c.530G>T p.G177V Chr.9: 129322343-129322343 on Build GRCh38
5512618 c.525C>T p.P175P Chr.9: 129322338-129322338 on Build GRCh38
5527830 c.473G>A p.R158Q Chr.9: 129322286-129322286 on Build GRCh38
3367580 c.393A>T p.V131V Chr.9: 129322206-129322206 on Build GRCh38
5711157 c.613C>T p.P205S Chr.9: 129322426-129322426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761978634 Chr.9: 129322376-129322376 on Build GRCh38
rs779938855 Chr.9: 129322442-129322442 on Build GRCh38
rs954177327 Chr.9: 129322583-129322583 on Build GRCh38
rs967745089 Chr.9: 129322580-129322580 on Build GRCh38
rs368910305 Chr.9: 129322283-129322283 on Build GRCh38
rs771655090 Chr.9: 129322176-129322176 on Build GRCh38
rs914072984 Chr.9: 129322286-129322286 on Build GRCh38
rs919080221 Chr.9: 129322620-129322620 on Build GRCh38
rs571594617 Chr.9: 129322270-129322270 on Build GRCh38
rs374960650 Chr.9: 129322516-129322516 on Build GRCh38
rs767434652 Chr.9: 129322382-129322382 on Build GRCh38
rs968234838 Chr.9: 129322517-129322517 on Build GRCh38
rs746486645 Chr.9: 129322282-129322282 on Build GRCh38
rs773293795 Chr.9: 129322191-129322191 on Build GRCh38
rs777600349 Chr.9: 129322406-129322406 on Build GRCh38
rs367946854 Chr.9: 129322531-129322531 on Build GRCh38
rs764095894 Chr.9: 129322239-129322239 on Build GRCh38
rs569682251 Chr.9: 129322515-129322515 on Build GRCh38
rs371637019 Chr.9: 129322508-129322508 on Build GRCh38
rs375407261 Chr.9: 129322450-129322450 on Build GRCh38
rs767745940 Chr.9: 129322355-129322358 on Build GRCh38
rs775694379 Chr.9: 129322292-129322292 on Build GRCh38
rs538614402 Chr.9: 129322593-129322593 on Build GRCh38
rs373771554 Chr.9: 129322234-129322234 on Build GRCh38
rs752868624 Chr.9: 129322227-129322227 on Build GRCh38
rs746774164 Chr.9: 129322588-129322588 on Build GRCh38
rs769720061 Chr.9: 129322360-129322360 on Build GRCh38
rs528507021 Chr.9: 129322338-129322338 on Build GRCh38
rs762717555 Chr.9: 129322366-129322366 on Build GRCh38
rs912557092 Chr.9: 129322592-129322592 on Build GRCh38
rs768909681 Chr.9: 129322369-129322369 on Build GRCh38
rs372659457 Chr.9: 129322285-129322285 on Build GRCh38
rs756797593 Chr.9: 129322424-129322424 on Build GRCh38
rs780633535 Chr.9: 129322427-129322427 on Build GRCh38
rs751635672 Chr.9: 129322242-129322242 on Build GRCh38
rs536357968 Chr.9: 129322488-129322488 on Build GRCh38
rs1051261601 Chr.9: 129322337-129322337 on Build GRCh38
rs774650127 Chr.9: 129322479-129322479 on Build GRCh38
rs763230458 Chr.9: 129322307-129322307 on Build GRCh38
rs375903553 Chr.9: 129322308-129322308 on Build GRCh38
rs370868810 Chr.9: 129322328-129322328 on Build GRCh38
rs912681927 Chr.9: 129322334-129322334 on Build GRCh38
rs757256315 Chr.9: 129322314-129322314 on Build GRCh38
rs368643086 Chr.9: 129322507-129322507 on Build GRCh38
rs913770 Chr.9: 129322342-129322342 on Build GRCh38
rs372178696 Chr.9: 129322537-129322537 on Build GRCh38
rs765535336 Chr.9: 129322225-129322225 on Build GRCh38
rs750850560 Chr.9: 129322325-129322325 on Build GRCh38
rs749115687 Chr.9: 129322447-129322447 on Build GRCh38
rs750940133 Chr.9: 129322386-129322386 on Build GRCh38
rs376448159 Chr.9: 129322278-129322278 on Build GRCh38
rs1034834948 Chr.9: 129322461-129322461 on Build GRCh38
rs747741624 Chr.9: 129322269-129322269 on Build GRCh38
rs759230317 Chr.9: 129322221-129322221 on Build GRCh38
rs527946656 Chr.9: 129322327-129322327 on Build GRCh38
rs745372337 Chr.9: 129322428-129322428 on Build GRCh38
rs769773152 Chr.9: 129322433-129322433 on Build GRCh38
rs572383294 Chr.9: 129322624-129322624 on Build GRCh38
rs913771 Chr.9: 129322606-129322606 on Build GRCh38
rs759405838 Chr.9: 129322291-129322291 on Build GRCh38
rs767176860 Chr.9: 129322552-129322552 on Build GRCh38
rs754302075 Chr.9: 129322394-129322394 on Build GRCh38
rs74931254 Chr.9: 129322171-129322171 on Build GRCh38
rs756919429 Chr.9: 129322551-129322551 on Build GRCh38
rs969095002 Chr.9: 129322229-129322229 on Build GRCh38
rs371997288 Chr.9: 129322548-129322548 on Build GRCh38
rs370810952 Chr.9: 129322371-129322371 on Build GRCh38
rs781204386 Chr.9: 129322351-129322351 on Build GRCh38
rs752531443 Chr.9: 129322178-129322182 on Build GRCh38
rs770306814 Chr.9: 129322348-129322348 on Build GRCh38
rs751690680 Chr.9: 129322311-129322311 on Build GRCh38
rs778727573 Chr.9: 129322262-129322262 on Build GRCh38
rs1001918784 Chr.9: 129322414-129322414 on Build GRCh38
rs761149540 Chr.9: 129322387-129322387 on Build GRCh38
rs758168947 Chr.9: 129322348-129322348 on Build GRCh38
rs756398406 Chr.9: 129322259-129322259 on Build GRCh38
rs756218424 Chr.9: 129322536-129322539 on Build GRCh38
rs774119284 Chr.9: 129322174-129322174 on Build GRCh38
rs758486276 Chr.9: 129322236-129322236 on Build GRCh38
rs766977584 Chr.9: 129322391-129322391 on Build GRCh38
rs547992621 Chr.9: 129322339-129322339 on Build GRCh38
rs113276848 Chr.9: 129322192-129322192 on Build GRCh38
rs897681848 Chr.9: 129322345-129322345 on Build GRCh38
rs541990348 Chr.9: 129322603-129322603 on Build GRCh38
rs775324292 Chr.9: 129322364-129322364 on Build GRCh38
rs750600809 Chr.9: 129322389-129322389 on Build GRCh38
rs551703959 Chr.9: 129322254-129322254 on Build GRCh38
rs760814090 Chr.9: 129322510-129322510 on Build GRCh38
rs987393930 Chr.9: 129322605-129322605 on Build GRCh38
rs531586302 Chr.9: 129322251-129322251 on Build GRCh38
rs781531554 Chr.9: 129322253-129322253 on Build GRCh38
rs374033998 Chr.9: 129322330-129322330 on Build GRCh38
rs369653324 Chr.9: 129322178-129322178 on Build GRCh38
rs1028611889 Chr.9: 129322554-129322554 on Build GRCh38
rs768104983 Chr.9: 129322316-129322316 on Build GRCh38
rs930503948 Chr.9: 129322629-129322629 on Build GRCh38
rs776545344 Chr.9: 129322214-129322214 on Build GRCh38
rs766221128 Chr.9: 129322209-129322209 on Build GRCh38
rs755361737 Chr.9: 129322404-129322404 on Build GRCh38
rs751123087 Chr.9: 129322411-129322411 on Build GRCh38
rs747694945 Chr.9: 129322175-129322175 on Build GRCh38
rs866222620 Chr.9: 129322463-129322463 on Build GRCh38
rs745726705 Chr.9: 129322352-129322352 on Build GRCh38

View Full Product Details