Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54071041-54071552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120C
Entrez Gene ID
54954
Gene Name
family with sequence similarity 120C
Gene Aliases
CXorf17, ORF34
UniGene
Hs.86045
Gene Chromosome Location
Chr.X: 54068324-54183281 on Build GRCh38
Transcripts
NM_001300788.1 NM_017848.5

SNP Detail

SNP ID Chromosome Location
rs781825513 Chr.X: 54071290-54071290 on Build GRCh38
rs964951325 Chr.X: 54071239-54071239 on Build GRCh38
rs782114740 Chr.X: 54071168-54071168 on Build GRCh38
rs782236325 Chr.X: 54071275-54071275 on Build GRCh38
rs920628077 Chr.X: 54071339-54071339 on Build GRCh38
rs1032254294 Chr.X: 54071092-54071092 on Build GRCh38
rs938500337 Chr.X: 54071470-54071470 on Build GRCh38
rs186126946 Chr.X: 54071201-54071201 on Build GRCh38
rs983576378 Chr.X: 54071435-54071435 on Build GRCh38
rs1017785727 Chr.X: 54071198-54071198 on Build GRCh38
rs189038554 Chr.X: 54071380-54071380 on Build GRCh38
rs45552536 Chr.X: 54071487-54071487 on Build GRCh38
rs371017503 Chr.X: 54071187-54071187 on Build GRCh38
rs781944626 Chr.X: 54071434-54071434 on Build GRCh38
rs782489727 Chr.X: 54071427-54071427 on Build GRCh38
rs782778131 Chr.X: 54071169-54071169 on Build GRCh38
rs782312394 Chr.X: 54071448-54071448 on Build GRCh38
rs782231626 Chr.X: 54071445-54071445 on Build GRCh38
rs782724529 Chr.X: 54071420-54071420 on Build GRCh38

View Full Product Details