Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54085620-54086156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120C
Entrez Gene ID
54954
Gene Name
family with sequence similarity 120C
Gene Aliases
CXorf17, ORF34
UniGene
Hs.86045
Gene Chromosome Location
Chr.X: 54068324-54183281 on Build GRCh38
Transcripts
NM_001300788.1 NM_017848.5 XM_006724589.3

SNP Detail

SNP ID Chromosome Location
rs781872460 Chr.X: 54085957-54085957 on Build GRCh38
rs782794402 Chr.X: 54085949-54085949 on Build GRCh38
rs149799597 Chr.X: 54085797-54085797 on Build GRCh38
rs782693570 Chr.X: 54085678-54085678 on Build GRCh38
rs926348446 Chr.X: 54085845-54085845 on Build GRCh38
rs782315833 Chr.X: 54085903-54085903 on Build GRCh38
rs782597475 Chr.X: 54085768-54085768 on Build GRCh38
rs368833346 Chr.X: 54085827-54085827 on Build GRCh38
rs782178348 Chr.X: 54085782-54085782 on Build GRCh38
rs41304786 Chr.X: 54085752-54085752 on Build GRCh38
rs782272855 Chr.X: 54085807-54085807 on Build GRCh38
rs781833238 Chr.X: 54085971-54085971 on Build GRCh38
rs1005674791 Chr.X: 54085668-54085668 on Build GRCh38
rs140976163 Chr.X: 54085763-54085763 on Build GRCh38
rs781910813 Chr.X: 54085931-54085931 on Build GRCh38
rs782773638 Chr.X: 54085759-54085759 on Build GRCh38
rs781906749 Chr.X: 54085679-54085679 on Build GRCh38
rs374652022 Chr.X: 54086066-54086066 on Build GRCh38
rs181955061 Chr.X: 54085995-54085995 on Build GRCh38
rs889759867 Chr.X: 54086098-54086098 on Build GRCh38
rs934282504 Chr.X: 54086093-54086093 on Build GRCh38
rs782114436 Chr.X: 54085958-54085958 on Build GRCh38
rs782674279 Chr.X: 54085800-54085800 on Build GRCh38
rs1026620820 Chr.X: 54085793-54085793 on Build GRCh38
rs993349162 Chr.X: 54085766-54085766 on Build GRCh38
rs782498893 Chr.X: 54086034-54086034 on Build GRCh38
rs373496740 Chr.X: 54085940-54085940 on Build GRCh38
rs782076605 Chr.X: 54085861-54085861 on Build GRCh38
rs1050185968 Chr.X: 54086096-54086096 on Build GRCh38
rs959106269 Chr.X: 54085846-54085846 on Build GRCh38
rs782132241 Chr.X: 54085695-54085695 on Build GRCh38
rs1042572782 Chr.X: 54086062-54086062 on Build GRCh38
rs188806284 Chr.X: 54085847-54085847 on Build GRCh38
rs782451806 Chr.X: 54085966-54085966 on Build GRCh38
rs782534877 Chr.X: 54085727-54085727 on Build GRCh38
rs781808727 Chr.X: 54085964-54085964 on Build GRCh38
rs782726240 Chr.X: 54085938-54085938 on Build GRCh38
rs368191687 Chr.X: 54085754-54085754 on Build GRCh38

View Full Product Details