Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70042102-70042599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT2
Entrez Gene ID
158835
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases
ARAT, DC4, DGAT2L4, MFAT, WS
UniGene
Hs.661507
Gene Chromosome Location
Chr.X: 70040542-70049970 on Build GRCh38
Transcripts
NM_001002254.1 XM_011530876.2

SNP Detail

SNP ID Chromosome Location
rs1001786052 Chr.X: 70042237-70042237 on Build GRCh38
rs762841384 Chr.X: 70042349-70042349 on Build GRCh38
rs756822595 Chr.X: 70042160-70042160 on Build GRCh38
rs1026829095 Chr.X: 70042320-70042320 on Build GRCh38
rs745380837 Chr.X: 70042188-70042188 on Build GRCh38
rs776710370 Chr.X: 70042437-70042437 on Build GRCh38
rs773213242 Chr.X: 70042270-70042270 on Build GRCh38
rs192973444 Chr.X: 70042480-70042480 on Build GRCh38
rs373537022 Chr.X: 70042176-70042176 on Build GRCh38
rs760843361 Chr.X: 70042281-70042281 on Build GRCh38
rs748984613 Chr.X: 70042217-70042217 on Build GRCh38
rs759677065 Chr.X: 70042456-70042456 on Build GRCh38
rs777658368 Chr.X: 70042531-70042531 on Build GRCh38
rs369168084 Chr.X: 70042391-70042391 on Build GRCh38
rs772011125 Chr.X: 70042252-70042252 on Build GRCh38
rs748142011 Chr.X: 70042251-70042251 on Build GRCh38
rs754716533 Chr.X: 70042379-70042379 on Build GRCh38
rs1015381749 Chr.X: 70042454-70042454 on Build GRCh38
rs755671268 Chr.X: 70042358-70042358 on Build GRCh38
rs2296767 Chr.X: 70042573-70042573 on Build GRCh38
rs747031983 Chr.X: 70042552-70042552 on Build GRCh38
rs763442483 Chr.X: 70042449-70042449 on Build GRCh38
rs188078036 Chr.X: 70042457-70042457 on Build GRCh38
rs759785986 Chr.X: 70042304-70042304 on Build GRCh38
rs778087249 Chr.X: 70042516-70042516 on Build GRCh38
rs747040613 Chr.X: 70042422-70042422 on Build GRCh38
rs776963283 Chr.X: 70042303-70042303 on Build GRCh38
rs772838454 Chr.X: 70042317-70042317 on Build GRCh38
rs768736379 Chr.X: 70042502-70042502 on Build GRCh38
rs780287028 Chr.X: 70042481-70042481 on Build GRCh38
rs953980455 Chr.X: 70042477-70042477 on Build GRCh38
rs751557127 Chr.X: 70042431-70042431 on Build GRCh38
rs766163803 Chr.X: 70042350-70042350 on Build GRCh38
rs753296093 Chr.X: 70042137-70042137 on Build GRCh38
rs755651655 Chr.X: 70042204-70042204 on Build GRCh38
rs779784501 Chr.X: 70042205-70042205 on Build GRCh38
rs988756884 Chr.X: 70042542-70042542 on Build GRCh38
rs759857064 Chr.X: 70042438-70042438 on Build GRCh38
rs375637323 Chr.X: 70042289-70042289 on Build GRCh38
rs765697172 Chr.X: 70042306-70042306 on Build GRCh38
rs779543494 Chr.X: 70042368-70042368 on Build GRCh38
rs778622024 Chr.X: 70042382-70042382 on Build GRCh38
rs778820383 Chr.X: 70042247-70042247 on Build GRCh38
rs1029491648 Chr.X: 70042460-70042460 on Build GRCh38
rs753594861 Chr.X: 70042371-70042371 on Build GRCh38
rs55925441 Chr.X: 70042495-70042495 on Build GRCh38
rs771961912 Chr.X: 70042398-70042398 on Build GRCh38
rs768538489 Chr.X: 70042242-70042242 on Build GRCh38
rs372751672 Chr.X: 70042401-70042401 on Build GRCh38
rs1028286707 Chr.X: 70042246-70042246 on Build GRCh38
rs976412058 Chr.X: 70042459-70042459 on Build GRCh38
rs775930854 Chr.X: 70042441-70042441 on Build GRCh38
rs778571114 Chr.X: 70042484-70042484 on Build GRCh38
rs764646463 Chr.X: 70042450-70042450 on Build GRCh38

View Full Product Details