Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70445087-70445576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG3
Entrez Gene ID
1741
Gene Name
discs large MAGUK scaffold protein 3
Gene Aliases
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
UniGene
Hs.721586
Gene Chromosome Location
Chr.X: 70444850-70505490 on Build GRCh38
Transcripts
NM_021120.3 XM_006724625.2 XM_006724626.2 XM_011530883.1

SNP Detail

SNP ID Chromosome Location
rs759180031 Chr.X: 70445113-70445114 on Build GRCh38
rs757417647 Chr.X: 70445193-70445193 on Build GRCh38
rs747060140 Chr.X: 70445282-70445282 on Build GRCh38
rs775915913 Chr.X: 70445345-70445345 on Build GRCh38
rs765001408 Chr.X: 70445403-70445403 on Build GRCh38
rs904017241 Chr.X: 70445174-70445174 on Build GRCh38
rs764051171 Chr.X: 70445132-70445132 on Build GRCh38
rs763609801 Chr.X: 70445353-70445353 on Build GRCh38
rs758372897 Chr.X: 70445420-70445420 on Build GRCh38
rs1001042267 Chr.X: 70445182-70445182 on Build GRCh38
rs950741512 Chr.X: 70445318-70445318 on Build GRCh38
rs749634953 Chr.X: 70445491-70445491 on Build GRCh38
rs770041632 Chr.X: 70445488-70445488 on Build GRCh38
rs370577131 Chr.X: 70445177-70445177 on Build GRCh38
rs778855350 Chr.X: 70445329-70445329 on Build GRCh38
rs756067925 Chr.X: 70445129-70445129 on Build GRCh38
rs954877440 Chr.X: 70445117-70445117 on Build GRCh38
rs766072313 Chr.X: 70445397-70445397 on Build GRCh38
rs759162614 Chr.X: 70445542-70445542 on Build GRCh38
rs749988770 Chr.X: 70445390-70445390 on Build GRCh38
rs752431046 Chr.X: 70445114-70445125 on Build GRCh38
rs781479423 Chr.X: 70445474-70445474 on Build GRCh38
rs866776696 Chr.X: 70445553-70445553 on Build GRCh38
rs776193643 Chr.X: 70445530-70445530 on Build GRCh38
rs11797456 Chr.X: 70445505-70445505 on Build GRCh38
rs959740155 Chr.X: 70445257-70445257 on Build GRCh38
rs1042702642 Chr.X: 70445144-70445144 on Build GRCh38
rs990103562 Chr.X: 70445395-70445395 on Build GRCh38
rs754657913 Chr.X: 70445402-70445402 on Build GRCh38
rs760217541 Chr.X: 70445394-70445394 on Build GRCh38
rs776667412 Chr.X: 70445294-70445294 on Build GRCh38
rs866252799 Chr.X: 70445541-70445541 on Build GRCh38
rs774945210 Chr.X: 70445493-70445493 on Build GRCh38
rs777648158 Chr.X: 70445435-70445435 on Build GRCh38
rs986270625 Chr.X: 70445173-70445173 on Build GRCh38
rs753368389 Chr.X: 70445398-70445398 on Build GRCh38
rs767300320 Chr.X: 70445114-70445116 on Build GRCh38
rs752468221 Chr.X: 70445414-70445414 on Build GRCh38
rs914271007 Chr.X: 70445312-70445312 on Build GRCh38
rs771173179 Chr.X: 70445292-70445292 on Build GRCh38
rs771984786 Chr.X: 70445198-70445198 on Build GRCh38
rs765875488 Chr.X: 70445523-70445523 on Build GRCh38
rs1041633529 Chr.X: 70445417-70445417 on Build GRCh38
rs910976467 Chr.X: 70445213-70445213 on Build GRCh38
rs746141766 Chr.X: 70445485-70445485 on Build GRCh38
rs764673027 Chr.X: 70445374-70445374 on Build GRCh38
rs764913253 Chr.X: 70445549-70445549 on Build GRCh38
rs757269383 Chr.X: 70445453-70445453 on Build GRCh38
rs374006597 Chr.X: 70445452-70445452 on Build GRCh38
rs759957195 Chr.X: 70445307-70445307 on Build GRCh38
rs982660834 Chr.X: 70445327-70445327 on Build GRCh38
rs777880846 Chr.X: 70445201-70445201 on Build GRCh38
rs376766177 Chr.X: 70445163-70445163 on Build GRCh38
rs775764782 Chr.X: 70445489-70445489 on Build GRCh38
rs907098191 Chr.X: 70445431-70445431 on Build GRCh38

View Full Product Details