Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230837425-230837918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf198
Entrez Gene ID
84886
Gene Name
chromosome 1 open reading frame 198
Gene Aliases
-
UniGene
Hs.520494
Gene Chromosome Location
Chr.1: 230837119-230869589 on Build GRCh38
Transcripts
NM_001136494.1 NM_001136495.1 NM_032800.2 XM_017002599.1

SNP Detail

SNP ID Chromosome Location
rs181309766 Chr.1: 230837820-230837820 on Build GRCh38
rs1038781135 Chr.1: 230837777-230837777 on Build GRCh38
rs987385890 Chr.1: 230837709-230837709 on Build GRCh38
rs12699 Chr.1: 230837661-230837661 on Build GRCh38
rs556875425 Chr.1: 230837487-230837487 on Build GRCh38
rs990102862 Chr.1: 230837794-230837794 on Build GRCh38
rs1041812745 Chr.1: 230837858-230837858 on Build GRCh38
rs958327073 Chr.1: 230837532-230837532 on Build GRCh38
rs563072505 Chr.1: 230837752-230837752 on Build GRCh38
rs1042203456 Chr.1: 230837574-230837574 on Build GRCh38
rs939175765 Chr.1: 230837878-230837878 on Build GRCh38
rs542957270 Chr.1: 230837748-230837748 on Build GRCh38
rs911453500 Chr.1: 230837716-230837716 on Build GRCh38
rs964654671 Chr.1: 230837751-230837751 on Build GRCh38
rs933484187 Chr.1: 230837821-230837821 on Build GRCh38
rs1016617663 Chr.1: 230837636-230837636 on Build GRCh38
rs1034916749 Chr.1: 230837610-230837610 on Build GRCh38
rs576580432 Chr.1: 230837761-230837761 on Build GRCh38
rs189808025 Chr.1: 230837635-230837635 on Build GRCh38
rs533818391 Chr.1: 230837642-230837642 on Build GRCh38
rs776336492 Chr.1: 230837779-230837779 on Build GRCh38
rs946119976 Chr.1: 230837837-230837837 on Build GRCh38
rs545080511 Chr.1: 230837797-230837797 on Build GRCh38
rs574235389 Chr.1: 230837662-230837662 on Build GRCh38
rs989999289 Chr.1: 230837609-230837609 on Build GRCh38
rs962445115 Chr.1: 230837641-230837641 on Build GRCh38
rs1056704494 Chr.1: 230837891-230837891 on Build GRCh38
rs928724301 Chr.1: 230837863-230837863 on Build GRCh38
rs568960451 Chr.1: 230837446-230837446 on Build GRCh38
rs1054389835 Chr.1: 230837457-230837457 on Build GRCh38
rs928995231 Chr.1: 230837682-230837682 on Build GRCh38
rs547426091 Chr.1: 230837896-230837896 on Build GRCh38
rs185671158 Chr.1: 230837893-230837893 on Build GRCh38
rs972516480 Chr.1: 230837659-230837659 on Build GRCh38
rs946172265 Chr.1: 230837564-230837564 on Build GRCh38
rs773878541 Chr.1: 230837555-230837555 on Build GRCh38
rs999532602 Chr.1: 230837892-230837892 on Build GRCh38
rs537917441 Chr.1: 230837547-230837547 on Build GRCh38
rs1003857549 Chr.1: 230837597-230837597 on Build GRCh38
rs769335981 Chr.1: 230837876-230837876 on Build GRCh38
rs1032439762 Chr.1: 230837464-230837464 on Build GRCh38
rs941736685 Chr.1: 230837750-230837750 on Build GRCh38
rs983053020 Chr.1: 230837712-230837712 on Build GRCh38
rs557922338 Chr.1: 230837590-230837590 on Build GRCh38
rs895074227 Chr.1: 230837632-230837632 on Build GRCh38

View Full Product Details