Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231334827-231335328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC8
Entrez Gene ID
149371
Gene Name
exocyst complex component 8
Gene Aliases
EXO84, Exo84p, SEC84
UniGene
Hs.356198
Gene Chromosome Location
Chr.1: 231332731-231337872 on Build GRCh38
Transcripts
NM_175876.4

SNP Detail

SNP ID Chromosome Location
rs139418927 Chr.1: 231335146-231335146 on Build GRCh38
rs918341568 Chr.1: 231334900-231334900 on Build GRCh38
rs1026022174 Chr.1: 231335147-231335147 on Build GRCh38
rs79933732 Chr.1: 231335239-231335239 on Build GRCh38
rs985036080 Chr.1: 231334964-231334964 on Build GRCh38
rs149976318 Chr.1: 231335211-231335211 on Build GRCh38
rs549890719 Chr.1: 231335255-231335259 on Build GRCh38
rs973054590 Chr.1: 231335105-231335105 on Build GRCh38
rs965226472 Chr.1: 231334984-231334984 on Build GRCh38
rs918468586 Chr.1: 231335150-231335150 on Build GRCh38
rs111249567 Chr.1: 231335285-231335285 on Build GRCh38
rs909785547 Chr.1: 231334869-231334869 on Build GRCh38
rs372061729 Chr.1: 231335060-231335060 on Build GRCh38
rs551844805 Chr.1: 231335125-231335125 on Build GRCh38
rs964955587 Chr.1: 231335188-231335188 on Build GRCh38
rs192371240 Chr.1: 231335005-231335005 on Build GRCh38
rs925309104 Chr.1: 231335249-231335249 on Build GRCh38
rs868389455 Chr.1: 231334968-231334968 on Build GRCh38
rs951681514 Chr.1: 231335149-231335149 on Build GRCh38
rs910753912 Chr.1: 231334978-231334979 on Build GRCh38
rs10489609 Chr.1: 231334904-231334904 on Build GRCh38
rs935948743 Chr.1: 231335029-231335032 on Build GRCh38
rs945438896 Chr.1: 231335016-231335017 on Build GRCh38
rs972554788 Chr.1: 231334899-231334899 on Build GRCh38
rs910942828 Chr.1: 231335221-231335221 on Build GRCh38
rs978134138 Chr.1: 231335240-231335243 on Build GRCh38
rs955368352 Chr.1: 231335192-231335192 on Build GRCh38
rs958721708 Chr.1: 231335051-231335051 on Build GRCh38
rs537820933 Chr.1: 231335017-231335017 on Build GRCh38
rs112748180 Chr.1: 231335218-231335218 on Build GRCh38
rs201313695 Chr.1: 231335025-231335026 on Build GRCh38
rs893365344 Chr.1: 231335017-231335017 on Build GRCh38
rs1050800942 Chr.1: 231334929-231334929 on Build GRCh38
rs1054649729 Chr.1: 231334977-231334977 on Build GRCh38
rs993244325 Chr.1: 231335137-231335137 on Build GRCh38
rs201460962 Chr.1: 231335025-231335025 on Build GRCh38
rs1045535614 Chr.1: 231335110-231335110 on Build GRCh38
rs990753020 Chr.1: 231335290-231335290 on Build GRCh38
rs867800961 Chr.1: 231335253-231335253 on Build GRCh38
rs936692706 Chr.1: 231335257-231335259 on Build GRCh38
rs753638896 Chr.1: 231335133-231335133 on Build GRCh38
rs941202405 Chr.1: 231334889-231334889 on Build GRCh38
rs971143574 Chr.1: 231334860-231334860 on Build GRCh38
rs1025947406 Chr.1: 231334906-231334906 on Build GRCh38
rs1002997436 Chr.1: 231335028-231335028 on Build GRCh38
rs185609384 Chr.1: 231335081-231335081 on Build GRCh38
rs1018284553 Chr.1: 231335176-231335176 on Build GRCh38
rs75696360 Chr.1: 231334938-231334938 on Build GRCh38
rs915823381 Chr.1: 231335299-231335299 on Build GRCh38
rs11122272 Chr.1: 231335026-231335026 on Build GRCh38
rs962625508 Chr.1: 231335101-231335101 on Build GRCh38
rs1034413537 Chr.1: 231335039-231335039 on Build GRCh38
rs1010823339 Chr.1: 231335021-231335021 on Build GRCh38
rs1006502991 Chr.1: 231335164-231335164 on Build GRCh38
rs548677216 Chr.1: 231335174-231335174 on Build GRCh38
rs923356161 Chr.1: 231335012-231335013 on Build GRCh38
rs894368122 Chr.1: 231335061-231335061 on Build GRCh38
rs976246912 Chr.1: 231334990-231334990 on Build GRCh38
rs967229540 Chr.1: 231335234-231335234 on Build GRCh38
rs942357963 Chr.1: 231334975-231334975 on Build GRCh38
rs1006196367 Chr.1: 231335054-231335054 on Build GRCh38
rs74852749 Chr.1: 231335240-231335240 on Build GRCh38

View Full Product Details