Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50133366-50133908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM21A
Entrez Gene ID
387680
Gene Name
family with sequence similarity 21 member A
Gene Aliases
FAM21B, bA56A21.1, bA98I6.1
UniGene
Hs.365286
Gene Chromosome Location
Chr.10: 50067888-50133509 on Build GRCh38
Transcripts
NM_001005751.2 NM_001291398.1 XM_005269806.3 XM_005269807.3 XM_005269808.3 XM_005269810.3 XM_006717831.3 XM_006717832.3 XM_011539786.2 XM_017016227.1 XM_017016228.1 XM_017016229.1 XM_017016230.1 XM_017016231.1 XM_017016232.1 XM_017016233.1 XM_017016234.1 XM_017016235.1

SNP Detail

SNP ID Chromosome Location
rs917527218 Chr.10: 50133425-50133425 on Build GRCh38
rs550959149 Chr.10: 50133807-50133807 on Build GRCh38
rs761132207 Chr.10: 50133408-50133408 on Build GRCh38
rs980327367 Chr.10: 50133458-50133458 on Build GRCh38
rs567971144 Chr.10: 50133869-50133869 on Build GRCh38
rs528847282 Chr.10: 50133877-50133877 on Build GRCh38
rs531158069 Chr.10: 50133635-50133635 on Build GRCh38
rs2813284 Chr.10: 50133532-50133532 on Build GRCh38
rs1007227317 Chr.10: 50133815-50133815 on Build GRCh38
rs1051044705 Chr.10: 50133813-50133813 on Build GRCh38
rs1037540603 Chr.10: 50133597-50133597 on Build GRCh38
rs948945156 Chr.10: 50133434-50133434 on Build GRCh38
rs941498208 Chr.10: 50133589-50133589 on Build GRCh38
rs897216449 Chr.10: 50133778-50133778 on Build GRCh38
rs910048462 Chr.10: 50133468-50133468 on Build GRCh38
rs753845039 Chr.10: 50133514-50133514 on Build GRCh38
rs889734998 Chr.10: 50133814-50133814 on Build GRCh38
rs2805252 Chr.10: 50133776-50133776 on Build GRCh38
rs377270279 Chr.10: 50133822-50133822 on Build GRCh38

View Full Product Details