Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50210680-50211205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
UniGene
Hs.512645
Gene Chromosome Location
Chr.10: 50184861-50251564 on Build GRCh38
Transcripts
NM_001143974.1 NM_019893.2 XM_011539970.2 XM_011539971.2 XM_011539972.2 XM_011539973.2 XM_017016435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321617 c.1271C>A p.A424D Chr.10: 50211091-50211091 on Build GRCh38
6202624 c.1333-8T>G p.? Chr.10: 50210912-50210912 on Build GRCh38
6321618 c.1214C>A p.A405D Chr.10: 50211091-50211091 on Build GRCh38
6202625 c.1276-8T>G p.? Chr.10: 50210912-50210912 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747068281 Chr.10: 50210835-50210835 on Build GRCh38
rs891289857 Chr.10: 50210754-50210754 on Build GRCh38
rs770698150 Chr.10: 50210903-50210903 on Build GRCh38
rs760223490 Chr.10: 50211128-50211128 on Build GRCh38
rs111329151 Chr.10: 50211166-50211166 on Build GRCh38
rs981244650 Chr.10: 50210995-50210995 on Build GRCh38
rs992270 Chr.10: 50210866-50210866 on Build GRCh38
rs772561919 Chr.10: 50210819-50210819 on Build GRCh38
rs780147044 Chr.10: 50210834-50210834 on Build GRCh38
rs1020441883 Chr.10: 50210743-50210743 on Build GRCh38
rs746431049 Chr.10: 50210788-50210788 on Build GRCh38
rs527279808 Chr.10: 50211135-50211135 on Build GRCh38
rs907585652 Chr.10: 50210867-50210867 on Build GRCh38
rs1010265420 Chr.10: 50210716-50210716 on Build GRCh38
rs56209738 Chr.10: 50211182-50211182 on Build GRCh38
rs370757287 Chr.10: 50210927-50210927 on Build GRCh38
rs1054491675 Chr.10: 50211092-50211092 on Build GRCh38
rs878921426 Chr.10: 50211182-50211183 on Build GRCh38
rs1018535469 Chr.10: 50210759-50210759 on Build GRCh38
rs767544067 Chr.10: 50211047-50211047 on Build GRCh38
rs547402222 Chr.10: 50211156-50211156 on Build GRCh38
rs893122130 Chr.10: 50211148-50211148 on Build GRCh38
rs1008309759 Chr.10: 50210757-50210757 on Build GRCh38
rs201529348 Chr.10: 50210844-50210844 on Build GRCh38
rs762657520 Chr.10: 50210895-50210895 on Build GRCh38
rs937239776 Chr.10: 50211083-50211083 on Build GRCh38
rs951803768 Chr.10: 50210858-50210858 on Build GRCh38
rs752280681 Chr.10: 50211068-50211068 on Build GRCh38
rs946172442 Chr.10: 50211150-50211150 on Build GRCh38
rs759589311 Chr.10: 50210992-50210992 on Build GRCh38
rs754491990 Chr.10: 50210927-50210927 on Build GRCh38
rs768779943 Chr.10: 50210839-50210839 on Build GRCh38
rs766246545 Chr.10: 50211182-50211183 on Build GRCh38
rs964614731 Chr.10: 50210780-50210780 on Build GRCh38

View Full Product Details