Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50218147-50218692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
UniGene
Hs.512645
Gene Chromosome Location
Chr.10: 50184861-50251564 on Build GRCh38
Transcripts
NM_001143974.1 NM_019893.2 XM_011539970.2 XM_011539971.2 XM_011539972.2 XM_011539973.2 XM_017016435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6066045 c.956A>T p.Q319L Chr.10: 50218511-50218511 on Build GRCh38
7329729 c.927C>A p.N309K Chr.10: 50218597-50218597 on Build GRCh38
6129539 c.987G>A p.E329E Chr.10: 50218537-50218537 on Build GRCh38
6431184 c.916A>G p.S306G Chr.10: 50218608-50218608 on Build GRCh38
6431185 c.859A>G p.S287G Chr.10: 50218608-50218608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763316221 Chr.10: 50218612-50218612 on Build GRCh38
rs1055540757 Chr.10: 50218591-50218591 on Build GRCh38
rs34881856 Chr.10: 50218223-50218223 on Build GRCh38
rs752292674 Chr.10: 50218484-50218485 on Build GRCh38
rs907834231 Chr.10: 50218284-50218284 on Build GRCh38
rs1021620163 Chr.10: 50218303-50218303 on Build GRCh38
rs56083155 Chr.10: 50218670-50218670 on Build GRCh38
rs990138875 Chr.10: 50218292-50218292 on Build GRCh38
rs553201514 Chr.10: 50218204-50218204 on Build GRCh38
rs1049240936 Chr.10: 50218433-50218433 on Build GRCh38
rs747036124 Chr.10: 50218525-50218525 on Build GRCh38
rs752856040 Chr.10: 50218644-50218644 on Build GRCh38
rs912606282 Chr.10: 50218381-50218381 on Build GRCh38
rs778155351 Chr.10: 50218669-50218669 on Build GRCh38
rs868755376 Chr.10: 50218274-50218274 on Build GRCh38
rs751862170 Chr.10: 50218623-50218623 on Build GRCh38
rs921541643 Chr.10: 50218400-50218400 on Build GRCh38
rs772780902 Chr.10: 50218573-50218573 on Build GRCh38
rs1002630915 Chr.10: 50218198-50218198 on Build GRCh38
rs965953512 Chr.10: 50218485-50218488 on Build GRCh38
rs777536435 Chr.10: 50218532-50218533 on Build GRCh38
rs757963221 Chr.10: 50218487-50218488 on Build GRCh38
rs755559730 Chr.10: 50218632-50218632 on Build GRCh38
rs545664569 Chr.10: 50218362-50218362 on Build GRCh38
rs967415400 Chr.10: 50218637-50218637 on Build GRCh38
rs1036125179 Chr.10: 50218543-50218543 on Build GRCh38
rs908878514 Chr.10: 50218445-50218445 on Build GRCh38
rs1019715271 Chr.10: 50218649-50218649 on Build GRCh38
rs879077132 Chr.10: 50218490-50218491 on Build GRCh38
rs1015046080 Chr.10: 50218187-50218187 on Build GRCh38
rs940316358 Chr.10: 50218498-50218498 on Build GRCh38
rs866634040 Chr.10: 50218399-50218399 on Build GRCh38
rs756082142 Chr.10: 50218656-50218656 on Build GRCh38
rs1002684609 Chr.10: 50218575-50218575 on Build GRCh38
rs965551210 Chr.10: 50218382-50218382 on Build GRCh38
rs977538663 Chr.10: 50218348-50218348 on Build GRCh38
rs967906644 Chr.10: 50218327-50218327 on Build GRCh38
rs866343887 Chr.10: 50218277-50218277 on Build GRCh38
rs894431680 Chr.10: 50218599-50218599 on Build GRCh38
rs576044126 Chr.10: 50218554-50218554 on Build GRCh38
rs775413915 Chr.10: 50218486-50218486 on Build GRCh38
rs1011445988 Chr.10: 50218607-50218607 on Build GRCh38
rs764899771 Chr.10: 50218568-50218568 on Build GRCh38
rs776252783 Chr.10: 50218547-50218547 on Build GRCh38
rs559284487 Chr.10: 50218417-50218417 on Build GRCh38
rs931544468 Chr.10: 50218424-50218424 on Build GRCh38
rs766785225 Chr.10: 50218613-50218613 on Build GRCh38
rs768445108 Chr.10: 50218534-50218534 on Build GRCh38
rs188906693 Chr.10: 50218321-50218321 on Build GRCh38
rs907040364 Chr.10: 50218195-50218195 on Build GRCh38
rs543424218 Chr.10: 50218633-50218633 on Build GRCh38

View Full Product Details