Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104269140-104269667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTO2
Entrez Gene ID
119391
Gene Name
glutathione S-transferase omega 2
Gene Aliases
GSTO 2-2, bA127L20.1
UniGene
Hs.107384
Gene Chromosome Location
Chr.10: 104267626-104304948 on Build GRCh38
Transcripts
NM_001191013.1 NM_183239.1 XM_006717624.3 XM_011539270.2 XM_011539271.1 XM_011539272.2 XM_011539273.2 XM_017015670.1 XM_017015671.1

SNP Detail

SNP ID Chromosome Location
rs186251353 Chr.10: 104269634-104269634 on Build GRCh38
rs985509294 Chr.10: 104269367-104269367 on Build GRCh38
rs1000877182 Chr.10: 104269266-104269266 on Build GRCh38
rs982825571 Chr.10: 104269598-104269598 on Build GRCh38
rs1047188454 Chr.10: 104269175-104269175 on Build GRCh38
rs1051398889 Chr.10: 104269412-104269412 on Build GRCh38
rs45605238 Chr.10: 104269602-104269602 on Build GRCh38
rs930042059 Chr.10: 104269354-104269354 on Build GRCh38
rs934276719 Chr.10: 104269390-104269390 on Build GRCh38
rs1020084816 Chr.10: 104269255-104269255 on Build GRCh38
rs968537246 Chr.10: 104269270-104269270 on Build GRCh38
rs541194964 Chr.10: 104269436-104269436 on Build GRCh38
rs948490275 Chr.10: 104269487-104269487 on Build GRCh38
rs941993406 Chr.10: 104269189-104269189 on Build GRCh38
rs909911967 Chr.10: 104269371-104269371 on Build GRCh38
rs12267156 Chr.10: 104269580-104269580 on Build GRCh38
rs533293826 Chr.10: 104269454-104269454 on Build GRCh38
rs904308751 Chr.10: 104269485-104269485 on Build GRCh38
rs749786401 Chr.10: 104269329-104269329 on Build GRCh38
rs559136536 Chr.10: 104269393-104269393 on Build GRCh38
rs374015495 Chr.10: 104269243-104269243 on Build GRCh38
rs45464593 Chr.10: 104269579-104269580 on Build GRCh38
rs189986219 Chr.10: 104269334-104269334 on Build GRCh38
rs958574517 Chr.10: 104269544-104269544 on Build GRCh38
rs568163412 Chr.10: 104269252-104269252 on Build GRCh38
rs772770713 Chr.10: 104269389-104269389 on Build GRCh38
rs995830139 Chr.10: 104269227-104269227 on Build GRCh38
rs917330301 Chr.10: 104269159-104269159 on Build GRCh38
rs1022670720 Chr.10: 104269231-104269231 on Build GRCh38
rs978587076 Chr.10: 104269285-104269285 on Build GRCh38
rs748761181 Chr.10: 104269530-104269530 on Build GRCh38
rs183193495 Chr.10: 104269596-104269596 on Build GRCh38
rs1044149628 Chr.10: 104269506-104269506 on Build GRCh38
rs1034183388 Chr.10: 104269491-104269491 on Build GRCh38
rs768941512 Chr.10: 104269398-104269398 on Build GRCh38
rs1037608017 Chr.10: 104269198-104269198 on Build GRCh38
rs1057343125 Chr.10: 104269213-104269213 on Build GRCh38
rs543742031 Chr.10: 104269206-104269206 on Build GRCh38
rs747478164 Chr.10: 104269202-104269202 on Build GRCh38
rs919197377 Chr.10: 104269399-104269399 on Build GRCh38
rs1029750971 Chr.10: 104269322-104269322 on Build GRCh38
rs578216769 Chr.10: 104269192-104269192 on Build GRCh38
rs917241354 Chr.10: 104269353-104269353 on Build GRCh38
rs45440495 Chr.10: 104269321-104269321 on Build GRCh38
rs1010406465 Chr.10: 104269253-104269253 on Build GRCh38
rs1047602677 Chr.10: 104269357-104269357 on Build GRCh38
rs45484701 Chr.10: 104269421-104269421 on Build GRCh38
rs993161593 Chr.10: 104269578-104269579 on Build GRCh38
rs973114752 Chr.10: 104269392-104269392 on Build GRCh38
rs575627610 Chr.10: 104269265-104269265 on Build GRCh38
rs1039924870 Chr.10: 104269386-104269386 on Build GRCh38
rs1056971691 Chr.10: 104269444-104269444 on Build GRCh38
rs904216631 Chr.10: 104269510-104269510 on Build GRCh38
rs1020473786 Chr.10: 104269459-104269459 on Build GRCh38
rs999976070 Chr.10: 104269486-104269486 on Build GRCh38
rs956466245 Chr.10: 104269275-104269275 on Build GRCh38
rs148931908 Chr.10: 104269542-104269542 on Build GRCh38
rs915813306 Chr.10: 104269350-104269351 on Build GRCh38
rs895569324 Chr.10: 104269225-104269225 on Build GRCh38
rs549508824 Chr.10: 104269619-104269619 on Build GRCh38
rs998367776 Chr.10: 104269287-104269287 on Build GRCh38
rs909881072 Chr.10: 104269171-104269171 on Build GRCh38
rs564106369 Chr.10: 104269449-104269449 on Build GRCh38
rs1029827687 Chr.10: 104269603-104269603 on Build GRCh38
rs897683178 Chr.10: 104269204-104269204 on Build GRCh38
rs45568844 Chr.10: 104269267-104269267 on Build GRCh38
rs1012635815 Chr.10: 104269229-104269229 on Build GRCh38
rs494431 Chr.10: 104269208-104269208 on Build GRCh38
rs773895040 Chr.10: 104269586-104269586 on Build GRCh38
rs941492592 Chr.10: 104269378-104269378 on Build GRCh38
rs769150953 Chr.10: 104269245-104269245 on Build GRCh38
rs541525080 Chr.10: 104269276-104269276 on Build GRCh38

View Full Product Details