Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104277806-104278317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTO2
Entrez Gene ID
119391
Gene Name
glutathione S-transferase omega 2
Gene Aliases
GSTO 2-2, bA127L20.1
UniGene
Hs.107384
Gene Chromosome Location
Chr.10: 104267626-104304948 on Build GRCh38
Transcripts
NM_001191013.1 NM_001191014.1 NM_001191015.1 NM_183239.1 XM_006717624.3 XM_011539270.2 XM_011539271.1 XM_011539272.2 XM_011539273.2 XM_017015670.1 XM_017015671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806401 c.282T>A p.D94E Chr.10: 104278032-104278032 on Build GRCh38
5049869 c.147T>C p.H49H Chr.10: 104277897-104277897 on Build GRCh38
5455999 c.187T>C p.Y63H Chr.10: 104277937-104277937 on Build GRCh38
1474336 c.358T>A p.F120I Chr.10: 104278108-104278108 on Build GRCh38
399269 c.336C>G p.R112R Chr.10: 104278086-104278086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs157077 Chr.10: 104278136-104278136 on Build GRCh38
rs569241863 Chr.10: 104278078-104278078 on Build GRCh38
rs779255158 Chr.10: 104278130-104278130 on Build GRCh38
rs1012548183 Chr.10: 104277833-104277833 on Build GRCh38
rs184604211 Chr.10: 104277840-104277840 on Build GRCh38
rs745641117 Chr.10: 104277910-104277910 on Build GRCh38
rs565662397 Chr.10: 104277868-104277868 on Build GRCh38
rs751068690 Chr.10: 104277979-104277979 on Build GRCh38
rs765140755 Chr.10: 104277994-104277998 on Build GRCh38
rs140989316 Chr.10: 104277922-104277922 on Build GRCh38
rs543640208 Chr.10: 104277838-104277838 on Build GRCh38
rs537213847 Chr.10: 104278263-104278263 on Build GRCh38
rs760917764 Chr.10: 104277970-104277970 on Build GRCh38
rs35233251 Chr.10: 104278199-104278200 on Build GRCh38
rs754725754 Chr.10: 104277856-104277856 on Build GRCh38
rs766202581 Chr.10: 104277994-104277994 on Build GRCh38
rs936800089 Chr.10: 104278200-104278200 on Build GRCh38
rs367989505 Chr.10: 104277968-104277968 on Build GRCh38
rs748443440 Chr.10: 104278206-104278206 on Build GRCh38
rs749484050 Chr.10: 104278001-104278001 on Build GRCh38
rs752432047 Chr.10: 104278001-104278002 on Build GRCh38
rs376506016 Chr.10: 104278084-104278084 on Build GRCh38
rs147953656 Chr.10: 104277976-104277976 on Build GRCh38
rs557171264 Chr.10: 104278276-104278276 on Build GRCh38
rs762658135 Chr.10: 104278055-104278055 on Build GRCh38
rs367969386 Chr.10: 104277896-104277896 on Build GRCh38
rs141797686 Chr.10: 104278107-104278107 on Build GRCh38
rs34289582 Chr.10: 104278155-104278155 on Build GRCh38
rs773283209 Chr.10: 104278146-104278146 on Build GRCh38
rs371150075 Chr.10: 104277850-104277850 on Build GRCh38
rs751461974 Chr.10: 104278032-104278032 on Build GRCh38
rs745711265 Chr.10: 104277987-104277987 on Build GRCh38
rs943832818 Chr.10: 104278277-104278277 on Build GRCh38
rs769505495 Chr.10: 104277989-104277989 on Build GRCh38
rs766623509 Chr.10: 104277886-104277886 on Build GRCh38
rs772745196 Chr.10: 104278031-104278031 on Build GRCh38
rs777272418 Chr.10: 104277978-104277978 on Build GRCh38
rs768931763 Chr.10: 104278003-104278003 on Build GRCh38
rs747473778 Chr.10: 104278161-104278161 on Build GRCh38
rs113205906 Chr.10: 104278222-104278222 on Build GRCh38
rs749092243 Chr.10: 104278112-104278112 on Build GRCh38
rs753315636 Chr.10: 104278127-104278127 on Build GRCh38
rs751114004 Chr.10: 104278079-104278079 on Build GRCh38
rs767017155 Chr.10: 104278085-104278085 on Build GRCh38
rs757246997 Chr.10: 104277980-104277980 on Build GRCh38
rs970858064 Chr.10: 104277933-104277933 on Build GRCh38
rs1022675374 Chr.10: 104277844-104277844 on Build GRCh38
rs750353386 Chr.10: 104278086-104278086 on Build GRCh38
rs750685329 Chr.10: 104277852-104277852 on Build GRCh38
rs374097618 Chr.10: 104277908-104277908 on Build GRCh38
rs756023576 Chr.10: 104278092-104278092 on Build GRCh38
rs772066716 Chr.10: 104278144-104278144 on Build GRCh38
rs774638750 Chr.10: 104278013-104278013 on Build GRCh38
rs199898162 Chr.10: 104277969-104277969 on Build GRCh38
rs763192301 Chr.10: 104277928-104277928 on Build GRCh38
rs371561309 Chr.10: 104277884-104277884 on Build GRCh38
rs766726986 Chr.10: 104277974-104277974 on Build GRCh38
rs754133511 Chr.10: 104277909-104277909 on Build GRCh38
rs776985540 Chr.10: 104278046-104278046 on Build GRCh38
rs761971523 Chr.10: 104278030-104278030 on Build GRCh38
rs770791064 Chr.10: 104277903-104277903 on Build GRCh38
rs765777427 Chr.10: 104277857-104277857 on Build GRCh38
rs759657759 Chr.10: 104277954-104277954 on Build GRCh38
rs761193920 Chr.10: 104278041-104278041 on Build GRCh38
rs775540190 Chr.10: 104277925-104277925 on Build GRCh38
rs534467893 Chr.10: 104277870-104277870 on Build GRCh38
rs34199276 Chr.10: 104277890-104277890 on Build GRCh38
rs375123980 Chr.10: 104278125-104278125 on Build GRCh38
rs768876856 Chr.10: 104277964-104277964 on Build GRCh38
rs779847735 Chr.10: 104277991-104277991 on Build GRCh38
rs1039877471 Chr.10: 104278280-104278280 on Build GRCh38
rs112958519 Chr.10: 104277961-104277961 on Build GRCh38

View Full Product Details