Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7995863-7996369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3F
Entrez Gene ID
8665
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
EIF3S5, eIF3-p47
UniGene
Hs.516023
Gene Chromosome Location
Chr.11: 7987320-7996171 on Build GRCh38
Transcripts
NM_003754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5759713 c.997_998insA p.D333fs*20 Chr.11: 7995945-7995946 on Build GRCh38
4680617 c.1005G>C p.L335F Chr.11: 7995953-7995953 on Build GRCh38
5759714 c.1042_1043insA p.D348fs*20 Chr.11: 7995945-7995946 on Build GRCh38
4680618 c.1050G>C p.L350F Chr.11: 7995953-7995953 on Build GRCh38
1188161 c.1008G>T p.M336I Chr.11: 7995956-7995956 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs561011232 Chr.11: 7996281-7996281 on Build GRCh38
rs1020387239 Chr.11: 7996113-7996113 on Build GRCh38
rs753508680 Chr.11: 7995925-7995925 on Build GRCh38
rs777775142 Chr.11: 7996036-7996036 on Build GRCh38
rs928242786 Chr.11: 7996195-7996195 on Build GRCh38
rs751440334 Chr.11: 7995996-7995996 on Build GRCh38
rs149594374 Chr.11: 7995948-7995948 on Build GRCh38
rs933884785 Chr.11: 7996274-7996274 on Build GRCh38
rs776275694 Chr.11: 7996051-7996051 on Build GRCh38
rs1042065939 Chr.11: 7996252-7996252 on Build GRCh38
rs186468615 Chr.11: 7996206-7996206 on Build GRCh38
rs748324069 Chr.11: 7996028-7996028 on Build GRCh38
rs547303904 Chr.11: 7996047-7996048 on Build GRCh38
rs543416260 Chr.11: 7996316-7996316 on Build GRCh38
rs547747925 Chr.11: 7996004-7996004 on Build GRCh38
rs774768076 Chr.11: 7995907-7995907 on Build GRCh38
rs764786881 Chr.11: 7996072-7996072 on Build GRCh38
rs182229070 Chr.11: 7996082-7996082 on Build GRCh38
rs760791517 Chr.11: 7996031-7996032 on Build GRCh38
rs775771292 Chr.11: 7996273-7996276 on Build GRCh38
rs375411282 Chr.11: 7996041-7996041 on Build GRCh38
rs192590368 Chr.11: 7996308-7996308 on Build GRCh38
rs1006717613 Chr.11: 7996078-7996078 on Build GRCh38
rs937186725 Chr.11: 7995942-7995942 on Build GRCh38
rs150724296 Chr.11: 7996231-7996231 on Build GRCh38
rs371566571 Chr.11: 7996149-7996149 on Build GRCh38
rs1041602174 Chr.11: 7996015-7996015 on Build GRCh38
rs749801374 Chr.11: 7996039-7996039 on Build GRCh38
rs139405488 Chr.11: 7996256-7996256 on Build GRCh38
rs572148040 Chr.11: 7996250-7996250 on Build GRCh38
rs778810915 Chr.11: 7996235-7996235 on Build GRCh38
rs144003413 Chr.11: 7995995-7995995 on Build GRCh38
rs756163024 Chr.11: 7996030-7996030 on Build GRCh38
rs763384355 Chr.11: 7995897-7995897 on Build GRCh38
rs897047637 Chr.11: 7995965-7995965 on Build GRCh38
rs556962512 Chr.11: 7996186-7996186 on Build GRCh38
rs373859618 Chr.11: 7995994-7995994 on Build GRCh38
rs982777202 Chr.11: 7996191-7996191 on Build GRCh38
rs902515288 Chr.11: 7996033-7996033 on Build GRCh38
rs770139004 Chr.11: 7996126-7996129 on Build GRCh38
rs889618518 Chr.11: 7996074-7996074 on Build GRCh38
rs767455275 Chr.11: 7995973-7995973 on Build GRCh38
rs755228761 Chr.11: 7996000-7996000 on Build GRCh38
rs1056872520 Chr.11: 7996226-7996226 on Build GRCh38
rs15879 Chr.11: 7996046-7996046 on Build GRCh38
rs7929026 Chr.11: 7996197-7996197 on Build GRCh38
rs754237401 Chr.11: 7995894-7995894 on Build GRCh38
rs761382498 Chr.11: 7996056-7996056 on Build GRCh38
rs144934973 Chr.11: 7996318-7996318 on Build GRCh38
rs900488887 Chr.11: 7996288-7996288 on Build GRCh38
rs1005041571 Chr.11: 7996329-7996329 on Build GRCh38
rs771469777 Chr.11: 7996040-7996040 on Build GRCh38
rs897155808 Chr.11: 7996270-7996270 on Build GRCh38
rs777098815 Chr.11: 7996002-7996003 on Build GRCh38
rs771275374 Chr.11: 7995906-7995906 on Build GRCh38
rs927053580 Chr.11: 7996199-7996199 on Build GRCh38
rs1010583199 Chr.11: 7995984-7995984 on Build GRCh38
rs568775312 Chr.11: 7996102-7996102 on Build GRCh38
rs957146133 Chr.11: 7996099-7996099 on Build GRCh38
rs760291930 Chr.11: 7995917-7995917 on Build GRCh38
rs963072657 Chr.11: 7996196-7996196 on Build GRCh38
rs954865282 Chr.11: 7996163-7996163 on Build GRCh38
rs991017095 Chr.11: 7996227-7996227 on Build GRCh38
rs1015620343 Chr.11: 7996337-7996337 on Build GRCh38
rs148646503 Chr.11: 7995962-7995962 on Build GRCh38

View Full Product Details