Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9142638-9143145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND5A
Entrez Gene ID
23258
Gene Name
DENN domain containing 5A
Gene Aliases
RAB6IP1
UniGene
Hs.501857
Gene Chromosome Location
Chr.11: 9138825-9265335 on Build GRCh38
Transcripts
NM_001243254.1 NM_015213.3 XM_005252832.1 XM_011519953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6642483 c.3401C>T p.T1134M Chr.11: 9142832-9142832 on Build GRCh38
6642482 c.3392G>A p.G1131D Chr.11: 9142841-9142841 on Build GRCh38
1357697 c.3475C>T p.R1159W Chr.11: 9142758-9142758 on Build GRCh38
5124340 c.3388C>T p.R1130* Chr.11: 9142845-9142845 on Build GRCh38
1203374 c.3470C>T p.S1157L Chr.11: 9142763-9142763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965133031 Chr.11: 9143114-9143114 on Build GRCh38
rs182220497 Chr.11: 9142944-9142944 on Build GRCh38
rs770405614 Chr.11: 9142786-9142786 on Build GRCh38
rs559114558 Chr.11: 9142905-9142905 on Build GRCh38
rs760415939 Chr.11: 9142921-9142921 on Build GRCh38
rs766146058 Chr.11: 9142923-9142923 on Build GRCh38
rs116742432 Chr.11: 9143111-9143111 on Build GRCh38
rs994464293 Chr.11: 9143107-9143107 on Build GRCh38
rs748779932 Chr.11: 9142774-9142774 on Build GRCh38
rs761813895 Chr.11: 9142850-9142850 on Build GRCh38
rs760907280 Chr.11: 9142860-9142861 on Build GRCh38
rs954808615 Chr.11: 9142684-9142684 on Build GRCh38
rs145302750 Chr.11: 9142762-9142762 on Build GRCh38
rs560393977 Chr.11: 9143051-9143051 on Build GRCh38
rs146355332 Chr.11: 9142867-9142867 on Build GRCh38
rs959889051 Chr.11: 9143117-9143117 on Build GRCh38
rs541099427 Chr.11: 9143007-9143007 on Build GRCh38
rs112449154 Chr.11: 9142929-9142929 on Build GRCh38
rs373435099 Chr.11: 9142696-9142696 on Build GRCh38
rs367803375 Chr.11: 9142869-9142869 on Build GRCh38
rs373053218 Chr.11: 9142704-9142704 on Build GRCh38
rs190631274 Chr.11: 9142849-9142849 on Build GRCh38
rs748911952 Chr.11: 9142879-9142879 on Build GRCh38
rs777434155 Chr.11: 9142763-9142763 on Build GRCh38
rs993404797 Chr.11: 9143008-9143008 on Build GRCh38
rs898847979 Chr.11: 9143084-9143084 on Build GRCh38
rs764355026 Chr.11: 9142687-9142687 on Build GRCh38
rs987550218 Chr.11: 9142702-9142702 on Build GRCh38
rs778436171 Chr.11: 9142887-9142887 on Build GRCh38
rs374951511 Chr.11: 9142875-9142875 on Build GRCh38
rs910600103 Chr.11: 9142706-9142706 on Build GRCh38
rs74643810 Chr.11: 9142890-9142890 on Build GRCh38
rs752486339 Chr.11: 9142683-9142683 on Build GRCh38
rs765463569 Chr.11: 9142855-9142855 on Build GRCh38
rs755329670 Chr.11: 9142874-9142874 on Build GRCh38
rs759009878 Chr.11: 9142862-9142862 on Build GRCh38
rs987268747 Chr.11: 9143090-9143090 on Build GRCh38
rs370059691 Chr.11: 9142832-9142832 on Build GRCh38
rs1026190425 Chr.11: 9143014-9143014 on Build GRCh38
rs567905657 Chr.11: 9142799-9142799 on Build GRCh38
rs750524183 Chr.11: 9142856-9142856 on Build GRCh38
rs1052029521 Chr.11: 9143024-9143024 on Build GRCh38
rs370847020 Chr.11: 9142699-9142699 on Build GRCh38
rs373302938 Chr.11: 9142735-9142735 on Build GRCh38
rs890193756 Chr.11: 9142680-9142680 on Build GRCh38
rs750904491 Chr.11: 9142755-9142755 on Build GRCh38
rs923956787 Chr.11: 9142913-9142913 on Build GRCh38
rs369257578 Chr.11: 9142677-9142677 on Build GRCh38
rs1004621428 Chr.11: 9143063-9143064 on Build GRCh38
rs774230861 Chr.11: 9142672-9142672 on Build GRCh38
rs200559278 Chr.11: 9142757-9142757 on Build GRCh38
rs551080178 Chr.11: 9142664-9142664 on Build GRCh38
rs58361373 Chr.11: 9143119-9143119 on Build GRCh38
rs1036082226 Chr.11: 9143071-9143071 on Build GRCh38
rs772019558 Chr.11: 9142896-9142896 on Build GRCh38
rs1042563836 Chr.11: 9142868-9142868 on Build GRCh38
rs766967497 Chr.11: 9142864-9142864 on Build GRCh38
rs760848249 Chr.11: 9142686-9142686 on Build GRCh38
rs371130612 Chr.11: 9142798-9142798 on Build GRCh38
rs775312488 Chr.11: 9142803-9142803 on Build GRCh38
rs779295508 Chr.11: 9142722-9142722 on Build GRCh38
rs759329139 Chr.11: 9142674-9142674 on Build GRCh38
rs780628819 Chr.11: 9142758-9142758 on Build GRCh38
rs756857795 Chr.11: 9142885-9142885 on Build GRCh38

View Full Product Details