Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10763510-10764008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTR9
Entrez Gene ID
9646
Gene Name
CTR9 homolog, Paf1/RNA polymerase II complex component
Gene Aliases
SH2BP1, TSBP, p150, p150TSP
UniGene
Hs.725151
Gene Chromosome Location
Chr.11: 10750987-10779755 on Build GRCh38
Transcripts
NM_014633.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466258 c.959A>G p.E320G Chr.11: 10763644-10763644 on Build GRCh38
1350680 c.1031T>G p.F344C Chr.11: 10763716-10763716 on Build GRCh38
922638 c.1044A>G p.G348G Chr.11: 10763729-10763729 on Build GRCh38
6637720 c.1099_1100insT p.L368fs*6 Chr.11: 10763784-10763785 on Build GRCh38
1627747 c.957+1G>A p.? Chr.11: 10763539-10763539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762845868 Chr.11: 10763693-10763693 on Build GRCh38
rs759194804 Chr.11: 10763884-10763884 on Build GRCh38
rs946353169 Chr.11: 10763618-10763618 on Build GRCh38
rs200254511 Chr.11: 10763935-10763935 on Build GRCh38
rs374903987 Chr.11: 10763927-10763927 on Build GRCh38
rs1031807350 Chr.11: 10763694-10763694 on Build GRCh38
rs541842136 Chr.11: 10763590-10763590 on Build GRCh38
rs1051614622 Chr.11: 10763835-10763835 on Build GRCh38
rs766307708 Chr.11: 10763696-10763696 on Build GRCh38
rs113823257 Chr.11: 10763766-10763766 on Build GRCh38
rs767354168 Chr.11: 10763771-10763771 on Build GRCh38
rs772656294 Chr.11: 10763691-10763691 on Build GRCh38
rs576753440 Chr.11: 10763645-10763645 on Build GRCh38
rs768325887 Chr.11: 10763576-10763576 on Build GRCh38
rs565770873 Chr.11: 10763566-10763566 on Build GRCh38
rs200587530 Chr.11: 10763580-10763580 on Build GRCh38
rs77892592 Chr.11: 10763887-10763887 on Build GRCh38
rs746242702 Chr.11: 10763629-10763629 on Build GRCh38
rs760297377 Chr.11: 10763571-10763571 on Build GRCh38
rs779466959 Chr.11: 10763844-10763844 on Build GRCh38
rs959794945 Chr.11: 10763607-10763607 on Build GRCh38
rs781560599 Chr.11: 10763608-10763608 on Build GRCh38
rs774729766 Chr.11: 10763570-10763570 on Build GRCh38
rs869312709 Chr.11: 10763811-10763811 on Build GRCh38
rs776117396 Chr.11: 10763684-10763684 on Build GRCh38
rs746306809 Chr.11: 10763848-10763848 on Build GRCh38
rs769322739 Chr.11: 10763681-10763681 on Build GRCh38
rs756358525 Chr.11: 10763810-10763810 on Build GRCh38
rs147990647 Chr.11: 10763612-10763612 on Build GRCh38
rs950796832 Chr.11: 10763977-10763977 on Build GRCh38
rs751345385 Chr.11: 10763599-10763599 on Build GRCh38
rs776194317 Chr.11: 10763579-10763579 on Build GRCh38
rs764319786 Chr.11: 10763819-10763819 on Build GRCh38
rs774358256 Chr.11: 10763914-10763914 on Build GRCh38
rs755193632 Chr.11: 10763600-10763600 on Build GRCh38
rs534755560 Chr.11: 10763569-10763569 on Build GRCh38
rs1039184387 Chr.11: 10763657-10763657 on Build GRCh38
rs113096852 Chr.11: 10763834-10763834 on Build GRCh38
rs142682548 Chr.11: 10763828-10763828 on Build GRCh38
rs959504180 Chr.11: 10763953-10763953 on Build GRCh38
rs775602651 Chr.11: 10763923-10763923 on Build GRCh38
rs1034163694 Chr.11: 10763947-10763947 on Build GRCh38
rs370226943 Chr.11: 10763902-10763902 on Build GRCh38
rs202173615 Chr.11: 10763839-10763839 on Build GRCh38
rs780352403 Chr.11: 10763866-10763866 on Build GRCh38
rs368661298 Chr.11: 10763896-10763896 on Build GRCh38
rs770832090 Chr.11: 10763909-10763909 on Build GRCh38
rs766106638 Chr.11: 10763597-10763597 on Build GRCh38
rs747728337 Chr.11: 10763651-10763651 on Build GRCh38
rs757773398 Chr.11: 10763833-10763833 on Build GRCh38
rs112427074 Chr.11: 10763913-10763913 on Build GRCh38
rs768359394 Chr.11: 10763637-10763637 on Build GRCh38
rs147353504 Chr.11: 10763867-10763867 on Build GRCh38
rs769410949 Chr.11: 10763883-10763883 on Build GRCh38
rs748825858 Chr.11: 10763907-10763907 on Build GRCh38
rs866714233 Chr.11: 10763933-10763933 on Build GRCh38
rs758762967 Chr.11: 10763856-10763856 on Build GRCh38
rs765294974 Chr.11: 10763887-10763887 on Build GRCh38
rs1028103366 Chr.11: 10763945-10763945 on Build GRCh38
rs376778984 Chr.11: 10763663-10763663 on Build GRCh38
rs773724262 Chr.11: 10763591-10763591 on Build GRCh38
rs559171057 Chr.11: 10763962-10763962 on Build GRCh38
rs373492280 Chr.11: 10763626-10763626 on Build GRCh38
rs776655388 Chr.11: 10763608-10763608 on Build GRCh38
rs199583444 Chr.11: 10763620-10763620 on Build GRCh38
rs1037662122 Chr.11: 10763895-10763895 on Build GRCh38
rs752999844 Chr.11: 10763793-10763793 on Build GRCh38
rs756266950 Chr.11: 10763614-10763614 on Build GRCh38
rs759756278 Chr.11: 10763886-10763887 on Build GRCh38
rs116866079 Chr.11: 10763596-10763596 on Build GRCh38
rs112593026 Chr.11: 10763746-10763746 on Build GRCh38
rs759360920 Chr.11: 10763714-10763714 on Build GRCh38
rs376089534 Chr.11: 10763615-10763615 on Build GRCh38
rs1033803334 Chr.11: 10763602-10763602 on Build GRCh38
rs771450084 Chr.11: 10763621-10763621 on Build GRCh38

View Full Product Details