Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13354988-13355488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
UniGene
Hs.65734
Gene Chromosome Location
Chr.11: 13276613-13387268 on Build GRCh38
Transcripts
NM_001030272.2 NM_001030273.2 NM_001178.5 NM_001297719.1 NM_001297722.1 NM_001297724.1 XM_006718233.3 XM_006718234.1 XM_011520105.2 XM_011520107.2 XM_011520108.1 XM_011520109.1 XM_011520110.1 XM_011520111.1 XM_011520112.2 XM_011520113.1 XM_017017738.1 XM_017017739.1 XM_017017740.1 XM_017017741.1 XM_017017742.1 XM_017017743.1 XM_017017744.1 XM_017017745.1 XM_017017746.1 XM_017017747.1 XM_017017748.1

SNP Detail

SNP ID Chromosome Location
rs936250374 Chr.11: 13355323-13355324 on Build GRCh38
rs760678465 Chr.11: 13355310-13355310 on Build GRCh38
rs769436602 Chr.11: 13355263-13355263 on Build GRCh38
rs552819588 Chr.11: 13355406-13355406 on Build GRCh38
rs973495954 Chr.11: 13355051-13355051 on Build GRCh38
rs1052352054 Chr.11: 13355184-13355184 on Build GRCh38
rs374284193 Chr.11: 13355261-13355261 on Build GRCh38
rs1010567285 Chr.11: 13355411-13355411 on Build GRCh38
rs550415609 Chr.11: 13355110-13355110 on Build GRCh38
rs568653185 Chr.11: 13355230-13355230 on Build GRCh38
rs1039152056 Chr.11: 13355158-13355158 on Build GRCh38
rs1023337434 Chr.11: 13355418-13355418 on Build GRCh38
rs935047635 Chr.11: 13355196-13355196 on Build GRCh38
rs1053555967 Chr.11: 13355359-13355359 on Build GRCh38
rs566831125 Chr.11: 13355187-13355187 on Build GRCh38
rs532297912 Chr.11: 13355079-13355079 on Build GRCh38
rs764646056 Chr.11: 13355102-13355102 on Build GRCh38
rs78287267 Chr.11: 13355118-13355118 on Build GRCh38
rs772439375 Chr.11: 13355064-13355064 on Build GRCh38
rs1045849604 Chr.11: 13355373-13355373 on Build GRCh38
rs772618618 Chr.11: 13355292-13355292 on Build GRCh38
rs530033646 Chr.11: 13355144-13355144 on Build GRCh38
rs1002049416 Chr.11: 13355410-13355410 on Build GRCh38
rs779917537 Chr.11: 13355248-13355248 on Build GRCh38
rs922177099 Chr.11: 13355138-13355138 on Build GRCh38
rs759866372 Chr.11: 13355289-13355289 on Build GRCh38
rs893443964 Chr.11: 13355405-13355405 on Build GRCh38
rs960387106 Chr.11: 13355037-13355037 on Build GRCh38
rs367822460 Chr.11: 13355271-13355271 on Build GRCh38
rs146287097 Chr.11: 13355436-13355436 on Build GRCh38
rs764888627 Chr.11: 13355291-13355297 on Build GRCh38
rs535295518 Chr.11: 13355122-13355122 on Build GRCh38
rs201755941 Chr.11: 13355257-13355257 on Build GRCh38
rs932276168 Chr.11: 13355183-13355183 on Build GRCh38
rs370741814 Chr.11: 13355313-13355313 on Build GRCh38
rs771383118 Chr.11: 13355280-13355280 on Build GRCh38
rs754008017 Chr.11: 13355341-13355341 on Build GRCh38
rs534429207 Chr.11: 13355286-13355286 on Build GRCh38
rs987179269 Chr.11: 13355111-13355111 on Build GRCh38
rs190717123 Chr.11: 13355442-13355442 on Build GRCh38
rs891784879 Chr.11: 13355362-13355363 on Build GRCh38
rs963296797 Chr.11: 13355018-13355018 on Build GRCh38
rs374112499 Chr.11: 13355368-13355368 on Build GRCh38
rs748479833 Chr.11: 13355015-13355015 on Build GRCh38
rs369151176 Chr.11: 13355180-13355180 on Build GRCh38
rs777512367 Chr.11: 13355264-13355264 on Build GRCh38
rs775926504 Chr.11: 13355296-13355296 on Build GRCh38
rs555042176 Chr.11: 13355186-13355186 on Build GRCh38
rs994758325 Chr.11: 13355426-13355427 on Build GRCh38
rs754924966 Chr.11: 13355260-13355260 on Build GRCh38
rs942037679 Chr.11: 13355121-13355121 on Build GRCh38
rs751409059 Chr.11: 13355252-13355252 on Build GRCh38
rs571028595 Chr.11: 13355430-13355430 on Build GRCh38
rs200621458 Chr.11: 13355099-13355099 on Build GRCh38
rs963655846 Chr.11: 13355416-13355416 on Build GRCh38
rs776762898 Chr.11: 13355236-13355236 on Build GRCh38
rs564706984 Chr.11: 13355026-13355026 on Build GRCh38

View Full Product Details